Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150368650-150369145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REPIN1
Entrez Gene ID
29803
Gene Name
replication initiator 1
Gene Aliases
AP4, RIP60, ZNF464, Zfp464
UniGene
Hs.647086
Gene Chromosome Location
Chr.7: 150368228-150374044 on Build GRCh38
Transcripts
NM_001099695.1 NM_001099696.2 NM_013400.3 XM_005249985.1 XM_006715947.3 XM_006715948.3 XM_006715949.3 XM_006715950.3 XM_006715951.2 XM_006715952.2 XM_006715953.2 XM_017012081.1 XM_017012082.1

SNP Detail

SNP ID Chromosome Location
rs892061035 Chr.7: 150368684-150368684 on Build GRCh38
rs1049874033 Chr.7: 150368672-150368672 on Build GRCh38
rs991835799 Chr.7: 150368784-150368784 on Build GRCh38
rs930740487 Chr.7: 150368805-150368805 on Build GRCh38
rs3735166 Chr.7: 150368919-150368919 on Build GRCh38
rs1026908636 Chr.7: 150369008-150369008 on Build GRCh38
rs985911367 Chr.7: 150368824-150368824 on Build GRCh38
rs964896249 Chr.7: 150368838-150368838 on Build GRCh38
rs947257526 Chr.7: 150368978-150368987 on Build GRCh38
rs1017701546 Chr.7: 150368774-150368774 on Build GRCh38
rs547300047 Chr.7: 150369043-150369044 on Build GRCh38
rs13233102 Chr.7: 150368941-150368941 on Build GRCh38
rs958991443 Chr.7: 150368755-150368755 on Build GRCh38
rs542020890 Chr.7: 150368704-150368704 on Build GRCh38
rs914640388 Chr.7: 150368942-150368942 on Build GRCh38
rs868165673 Chr.7: 150368746-150368746 on Build GRCh38
rs1043353465 Chr.7: 150368888-150368888 on Build GRCh38
rs185062943 Chr.7: 150369038-150369038 on Build GRCh38
rs911316321 Chr.7: 150368825-150368825 on Build GRCh38
rs938705274 Chr.7: 150369093-150369093 on Build GRCh38
rs914321186 Chr.7: 150368959-150368979 on Build GRCh38
rs1001020169 Chr.7: 150368744-150368744 on Build GRCh38
rs988965666 Chr.7: 150368877-150368877 on Build GRCh38
rs961062952 Chr.7: 150368978-150368978 on Build GRCh38
rs865809414 Chr.7: 150369015-150369015 on Build GRCh38
rs561404569 Chr.7: 150368705-150368705 on Build GRCh38
rs572119381 Chr.7: 150368795-150368795 on Build GRCh38
rs919748779 Chr.7: 150368812-150368812 on Build GRCh38
rs913227203 Chr.7: 150369074-150369074 on Build GRCh38
rs997725357 Chr.7: 150368851-150368851 on Build GRCh38
rs549211263 Chr.7: 150369124-150369124 on Build GRCh38
rs946278695 Chr.7: 150368878-150368878 on Build GRCh38
rs1033436790 Chr.7: 150368750-150368750 on Build GRCh38
rs976834121 Chr.7: 150369050-150369050 on Build GRCh38
rs931204174 Chr.7: 150368819-150368819 on Build GRCh38
rs1041248711 Chr.7: 150368682-150368682 on Build GRCh38
rs927290234 Chr.7: 150369083-150369083 on Build GRCh38
rs1010337323 Chr.7: 150368702-150368702 on Build GRCh38
rs927317975 Chr.7: 150368687-150368687 on Build GRCh38
rs1030577856 Chr.7: 150368854-150368854 on Build GRCh38
rs563623776 Chr.7: 150368959-150368959 on Build GRCh38
rs1024872650 Chr.7: 150368797-150368797 on Build GRCh38
rs961264874 Chr.7: 150368803-150368803 on Build GRCh38
rs540640306 Chr.7: 150368799-150368799 on Build GRCh38
rs985325333 Chr.7: 150369030-150369030 on Build GRCh38
rs201711107 Chr.7: 150368705-150368710 on Build GRCh38
rs550087796 Chr.7: 150368958-150368958 on Build GRCh38
rs965416568 Chr.7: 150369044-150369044 on Build GRCh38
rs576225628 Chr.7: 150368676-150368676 on Build GRCh38
rs13232770 Chr.7: 150368940-150368940 on Build GRCh38
rs564085661 Chr.7: 150368907-150368907 on Build GRCh38
rs373945522 Chr.7: 150368989-150368989 on Build GRCh38
rs529054255 Chr.7: 150369046-150369046 on Build GRCh38
rs956197471 Chr.7: 150368863-150368863 on Build GRCh38
rs1041043575 Chr.7: 150368850-150368850 on Build GRCh38
rs1043667309 Chr.7: 150369099-150369099 on Build GRCh38
rs972701908 Chr.7: 150368806-150368806 on Build GRCh38
rs1049196554 Chr.7: 150368836-150368836 on Build GRCh38
rs952620907 Chr.7: 150369011-150369011 on Build GRCh38
rs866931318 Chr.7: 150368992-150368992 on Build GRCh38
rs994491651 Chr.7: 150368987-150368987 on Build GRCh38

View Full Product Details