Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 30474515-30475021 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBPMS
Entrez Gene ID
11030
Gene Name
RNA binding protein with multiple splicing
Gene Aliases
HERMES
UniGene
Hs.334587
Gene Chromosome Location
Chr.8: 30384501-30572261 on Build GRCh38
Transcripts
NM_001008710.2 NM_001008711.2 NM_001008712.2 NM_006867.3 XM_017012980.1 XM_017012981.1 XM_017012982.1 XM_017012983.1 XM_017012984.1 XM_017012986.1 XM_017012987.1 XM_017012988.1 XM_017012989.1 XM_017012990.1 XM_017012991.1 XM_017012992.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5922701 c.67-4C>T p.? Chr.8: 30474775-30474775 on Build GRCh38
3367315 c.71G>T p.R24L Chr.8: 30474783-30474783 on Build GRCh38
236556 c.130T>A p.F44I Chr.8: 30474842-30474842 on Build GRCh38
1456595 c.71G>A p.R24Q Chr.8: 30474783-30474783 on Build GRCh38
3648200 c.116A>G p.E39G Chr.8: 30474828-30474828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769102908 Chr.8: 30474836-30474836 on Build GRCh38
rs532445912 Chr.8: 30474854-30474854 on Build GRCh38
rs1015931387 Chr.8: 30474957-30474958 on Build GRCh38
rs577437212 Chr.8: 30474825-30474825 on Build GRCh38
rs572351601 Chr.8: 30474760-30474760 on Build GRCh38
rs373319357 Chr.8: 30474833-30474833 on Build GRCh38
rs770557248 Chr.8: 30474790-30474790 on Build GRCh38
rs935322315 Chr.8: 30474610-30474610 on Build GRCh38
rs149499598 Chr.8: 30474649-30474649 on Build GRCh38
rs1004664955 Chr.8: 30474915-30474915 on Build GRCh38
rs190682746 Chr.8: 30474652-30474652 on Build GRCh38
rs902619284 Chr.8: 30474715-30474715 on Build GRCh38
rs755921303 Chr.8: 30474903-30474903 on Build GRCh38
rs1036895395 Chr.8: 30474607-30474607 on Build GRCh38
rs145996898 Chr.8: 30474701-30474701 on Build GRCh38
rs540978589 Chr.8: 30474782-30474782 on Build GRCh38
rs777192339 Chr.8: 30474861-30474861 on Build GRCh38
rs987539421 Chr.8: 30474586-30474586 on Build GRCh38
rs35731243 Chr.8: 30474568-30474569 on Build GRCh38
rs564361009 Chr.8: 30474794-30474794 on Build GRCh38
rs939831901 Chr.8: 30474604-30474604 on Build GRCh38
rs773444466 Chr.8: 30474881-30474881 on Build GRCh38
rs539989416 Chr.8: 30474547-30474547 on Build GRCh38
rs185717851 Chr.8: 30474643-30474643 on Build GRCh38
rs754815994 Chr.8: 30474743-30474743 on Build GRCh38
rs372974209 Chr.8: 30474977-30474977 on Build GRCh38
rs113920678 Chr.8: 30474969-30474969 on Build GRCh38
rs750613237 Chr.8: 30474742-30474742 on Build GRCh38
rs199664065 Chr.8: 30474755-30474755 on Build GRCh38
rs1049720974 Chr.8: 30474654-30474654 on Build GRCh38
rs780800880 Chr.8: 30474748-30474748 on Build GRCh38
rs752375191 Chr.8: 30474902-30474902 on Build GRCh38
rs763121434 Chr.8: 30474899-30474899 on Build GRCh38
rs889794651 Chr.8: 30474659-30474659 on Build GRCh38
rs201462610 Chr.8: 30474866-30474866 on Build GRCh38
rs1022552692 Chr.8: 30474892-30474892 on Build GRCh38
rs745472931 Chr.8: 30474805-30474805 on Build GRCh38
rs947295370 Chr.8: 30474718-30474718 on Build GRCh38
rs1056861346 Chr.8: 30474863-30474863 on Build GRCh38
rs924033470 Chr.8: 30474590-30474590 on Build GRCh38
rs764028194 Chr.8: 30474861-30474862 on Build GRCh38
rs772600046 Chr.8: 30474821-30474821 on Build GRCh38
rs765278914 Chr.8: 30474874-30474874 on Build GRCh38
rs1041525575 Chr.8: 30474703-30474703 on Build GRCh38
rs368733156 Chr.8: 30474784-30474784 on Build GRCh38

View Full Product Details