Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 5085894-5086452 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG1
Entrez Gene ID
56052
Gene Name
ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase
Gene Aliases
CDG1K, HMAT1, HMT-1, HMT1, MT-1, Mat-1, hMat-1
UniGene
Hs.592086
Gene Chromosome Location
Chr.16: 5071809-5087379 on Build GRCh38
Transcripts
NM_019109.4 NM_001289029.1 NM_201400.3 NM_201598.3 XM_005255157.4 XM_005255158.4 XM_011522404.2

Gene Symbol
EEF2KMT
Entrez Gene ID
196483
Gene Name
eukaryotic elongation factor 2 lysine methyltransferase
Gene Aliases
FAM86A, SB153, eEF2-KMT
UniGene
Hs.406461
Gene Chromosome Location
Chr.16: 5084298-5097820 on Build GRCh38
Transcripts
NM_019109.4 NM_001289029.1 NM_201400.3 NM_201598.3 XM_005255157.4 XM_005255158.4 XM_011522404.2

SNP Detail

SNP ID Chromosome Location
rs753027372 Chr.16: 5086334-5086338 on Build GRCh38
rs12922304 Chr.16: 5086319-5086319 on Build GRCh38
rs147517741 Chr.16: 5086419-5086419 on Build GRCh38
rs886598702 Chr.16: 5086058-5086058 on Build GRCh38
rs773075103 Chr.16: 5086058-5086058 on Build GRCh38
rs537130220 Chr.16: 5086111-5086111 on Build GRCh38
rs921932816 Chr.16: 5086332-5086332 on Build GRCh38
rs374671583 Chr.16: 5086335-5086338 on Build GRCh38
rs559311174 Chr.16: 5086029-5086029 on Build GRCh38
rs552179410 Chr.16: 5086000-5086000 on Build GRCh38
rs1038980625 Chr.16: 5086025-5086025 on Build GRCh38
rs186602069 Chr.16: 5086185-5086185 on Build GRCh38
rs947406796 Chr.16: 5085986-5085986 on Build GRCh38
rs1051976603 Chr.16: 5086057-5086057 on Build GRCh38
rs191507482 Chr.16: 5086199-5086199 on Build GRCh38
rs571206182 Chr.16: 5086054-5086054 on Build GRCh38
rs183691393 Chr.16: 5086248-5086248 on Build GRCh38
rs532464961 Chr.16: 5085999-5085999 on Build GRCh38
rs543489333 Chr.16: 5086380-5086380 on Build GRCh38
rs899391905 Chr.16: 5086152-5086152 on Build GRCh38
rs773283628 Chr.16: 5086334-5086337 on Build GRCh38
rs1022626268 Chr.16: 5086182-5086182 on Build GRCh38
rs376537060 Chr.16: 5086043-5086043 on Build GRCh38
rs4786593 Chr.16: 5085955-5085955 on Build GRCh38
rs556079793 Chr.16: 5086336-5086339 on Build GRCh38
rs62036246 Chr.16: 5085983-5085983 on Build GRCh38
rs968591368 Chr.16: 5086288-5086288 on Build GRCh38
rs907913813 Chr.16: 5086393-5086393 on Build GRCh38
rs62036247 Chr.16: 5086375-5086375 on Build GRCh38
rs770971453 Chr.16: 5086336-5086338 on Build GRCh38
rs4786594 Chr.16: 5086084-5086084 on Build GRCh38
rs989128586 Chr.16: 5085973-5085973 on Build GRCh38
rs995185688 Chr.16: 5086162-5086162 on Build GRCh38
rs576923101 Chr.16: 5086406-5086406 on Build GRCh38
rs541521482 Chr.16: 5085954-5085954 on Build GRCh38
rs928440795 Chr.16: 5085950-5085950 on Build GRCh38
rs1022739375 Chr.16: 5085928-5085928 on Build GRCh38
rs1012487377 Chr.16: 5086432-5086432 on Build GRCh38
rs189184478 Chr.16: 5086404-5086404 on Build GRCh38
rs572405748 Chr.16: 5086234-5086234 on Build GRCh38
rs570262270 Chr.16: 5086190-5086190 on Build GRCh38
rs956077603 Chr.16: 5086224-5086224 on Build GRCh38
rs543177308 Chr.16: 5085977-5085977 on Build GRCh38
rs4238853 Chr.16: 5085978-5085978 on Build GRCh38
rs568129805 Chr.16: 5086107-5086107 on Build GRCh38
rs574313691 Chr.16: 5085957-5085957 on Build GRCh38
rs899445734 Chr.16: 5086425-5086425 on Build GRCh38
rs4483845 Chr.16: 5085946-5085946 on Build GRCh38
rs558941209 Chr.16: 5086229-5086229 on Build GRCh38
rs371997547 Chr.16: 5086374-5086385 on Build GRCh38
rs1016529262 Chr.16: 5086088-5086088 on Build GRCh38
rs760492544 Chr.16: 5086066-5086066 on Build GRCh38
rs111263245 Chr.16: 5086293-5086293 on Build GRCh38
rs969767356 Chr.16: 5085934-5085934 on Build GRCh38

View Full Product Details