Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20475659-20476176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM2A
Entrez Gene ID
123876
Gene Name
acyl-CoA synthetase medium-chain family member 2A
Gene Aliases
A-923A4.1, ACSM2
UniGene
Hs.298252
Gene Chromosome Location
Chr.16: 20451461-20487669 on Build GRCh38
Transcripts
NM_001308169.1 NM_001308172.1 NM_001308954.1 XM_017022923.1 XM_017022924.1 XM_017022925.1 XM_017022926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4521079 c.1093G>T p.E365* Chr.16: 20475768-20475768 on Build GRCh38
3506959 c.984C>T p.F328F Chr.16: 20475659-20475659 on Build GRCh38
4515401 c.1086_1087CC>AT p.Q363* Chr.16: 20475761-20475762 on Build GRCh38
1478600 c.1098G>A p.T366T Chr.16: 20475773-20475773 on Build GRCh38
5794362 c.1003G>C p.V335L Chr.16: 20475678-20475678 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376985838 Chr.16: 20475774-20475774 on Build GRCh38
rs1055683278 Chr.16: 20476007-20476007 on Build GRCh38
rs974398507 Chr.16: 20475920-20475920 on Build GRCh38
rs921607272 Chr.16: 20475929-20475929 on Build GRCh38
rs755090254 Chr.16: 20475793-20475793 on Build GRCh38
rs140718178 Chr.16: 20476063-20476063 on Build GRCh38
rs375807855 Chr.16: 20475789-20475789 on Build GRCh38
rs373701058 Chr.16: 20475807-20475807 on Build GRCh38
rs4643305 Chr.16: 20475678-20475678 on Build GRCh38
rs1022155623 Chr.16: 20476045-20476045 on Build GRCh38
rs7194525 Chr.16: 20476068-20476068 on Build GRCh38
rs545155456 Chr.16: 20476084-20476084 on Build GRCh38
rs567611041 Chr.16: 20475898-20475898 on Build GRCh38
rs568717334 Chr.16: 20475844-20475844 on Build GRCh38
rs770321710 Chr.16: 20475894-20475894 on Build GRCh38
rs765780796 Chr.16: 20475694-20475694 on Build GRCh38
rs150888398 Chr.16: 20475772-20475772 on Build GRCh38
rs529706655 Chr.16: 20475891-20475891 on Build GRCh38
rs1049361067 Chr.16: 20475937-20475937 on Build GRCh38
rs377370065 Chr.16: 20475870-20475870 on Build GRCh38
rs149973694 Chr.16: 20475711-20475711 on Build GRCh38
rs760212019 Chr.16: 20475772-20475773 on Build GRCh38
rs765828342 Chr.16: 20475708-20475708 on Build GRCh38
rs908015632 Chr.16: 20475948-20475948 on Build GRCh38
rs762511273 Chr.16: 20475700-20475700 on Build GRCh38
rs540468572 Chr.16: 20476012-20476012 on Build GRCh38
rs940834671 Chr.16: 20475959-20475959 on Build GRCh38
rs7194172 Chr.16: 20475893-20475893 on Build GRCh38
rs556155291 Chr.16: 20475931-20475931 on Build GRCh38
rs1629314 Chr.16: 20475803-20475803 on Build GRCh38
rs763530400 Chr.16: 20475767-20475767 on Build GRCh38
rs911613694 Chr.16: 20475908-20475908 on Build GRCh38
rs1045937017 Chr.16: 20475970-20475970 on Build GRCh38
rs752355476 Chr.16: 20475775-20475775 on Build GRCh38
rs386789671 Chr.16: 20475844-20475845 on Build GRCh38
rs777625321 Chr.16: 20475743-20475743 on Build GRCh38
rs907306931 Chr.16: 20475861-20475861 on Build GRCh38
rs753684318 Chr.16: 20475790-20475790 on Build GRCh38
rs754557094 Chr.16: 20475724-20475724 on Build GRCh38
rs377092188 Chr.16: 20475811-20475811 on Build GRCh38
rs571237988 Chr.16: 20475947-20475947 on Build GRCh38
rs141193766 Chr.16: 20475761-20475761 on Build GRCh38
rs370692642 Chr.16: 20475930-20475930 on Build GRCh38
rs4586421 Chr.16: 20475685-20475685 on Build GRCh38
rs763971345 Chr.16: 20475773-20475774 on Build GRCh38
rs768743436 Chr.16: 20475754-20475754 on Build GRCh38
rs771329190 Chr.16: 20475816-20475816 on Build GRCh38
rs776920544 Chr.16: 20475757-20475757 on Build GRCh38
rs965861366 Chr.16: 20475910-20475910 on Build GRCh38
rs145974879 Chr.16: 20475736-20475736 on Build GRCh38
rs900753104 Chr.16: 20475797-20475797 on Build GRCh38
rs182151770 Chr.16: 20476099-20476099 on Build GRCh38
rs756937801 Chr.16: 20475810-20475810 on Build GRCh38
rs186674723 Chr.16: 20475967-20475967 on Build GRCh38
rs563682940 Chr.16: 20476044-20476044 on Build GRCh38
rs766991867 Chr.16: 20475687-20475687 on Build GRCh38
rs759143815 Chr.16: 20475773-20475773 on Build GRCh38
rs190097007 Chr.16: 20476030-20476030 on Build GRCh38
rs188648395 Chr.16: 20475748-20475748 on Build GRCh38
rs921577894 Chr.16: 20476145-20476145 on Build GRCh38
rs867104027 Chr.16: 20476093-20476093 on Build GRCh38
rs149105084 Chr.16: 20475792-20475792 on Build GRCh38
rs148631132 Chr.16: 20475747-20475747 on Build GRCh38
rs562548761 Chr.16: 20475804-20475804 on Build GRCh38
rs775021492 Chr.16: 20475694-20475694 on Build GRCh38
rs753830038 Chr.16: 20475797-20475797 on Build GRCh38
rs752421976 Chr.16: 20475728-20475728 on Build GRCh38
rs781230612 Chr.16: 20475798-20475798 on Build GRCh38
rs780965176 Chr.16: 20475726-20475726 on Build GRCh38
rs1010780618 Chr.16: 20475881-20475881 on Build GRCh38
rs762322900 Chr.16: 20475690-20475690 on Build GRCh38
rs1018814065 Chr.16: 20476081-20476081 on Build GRCh38
rs755867496 Chr.16: 20475778-20475778 on Build GRCh38
rs199759251 Chr.16: 20475855-20475855 on Build GRCh38
rs371399781 Chr.16: 20475764-20475764 on Build GRCh38
rs184343147 Chr.16: 20475695-20475695 on Build GRCh38
rs146721513 Chr.16: 20475875-20475875 on Build GRCh38
rs373306567 Chr.16: 20475766-20475766 on Build GRCh38
rs562513616 Chr.16: 20476067-20476067 on Build GRCh38
rs974494792 Chr.16: 20476103-20476103 on Build GRCh38
rs145344329 Chr.16: 20475976-20475976 on Build GRCh38
rs776851912 Chr.16: 20475689-20475689 on Build GRCh38
rs551872058 Chr.16: 20475845-20475845 on Build GRCh38
rs375321510 Chr.16: 20476001-20476001 on Build GRCh38

View Full Product Details