Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 70331815-70332339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPFIA1
Entrez Gene ID
8500
Gene Name
PTPRF interacting protein alpha 1
Gene Aliases
LIP.1, LIP1, LIPRIN
UniGene
Hs.530749
Gene Chromosome Location
Chr.11: 70270687-70384501 on Build GRCh38
Transcripts
NM_003626.3 NM_177423.2 NR_045286.1 XM_006718716.3 XM_011545306.2 XM_011545307.2 XM_011545308.2 XM_011545309.2 XM_011545310.2 XM_011545311.2 XM_011545312.2 XM_011545313.2 XM_011545314.2 XM_011545315.2 XM_011545316.2 XM_011545317.2 XM_011545319.1 XM_017018448.1 XM_017018449.1 XM_017018450.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7154033 c.1191G>C p.Q397H Chr.11: 70332073-70332073 on Build GRCh38
4652058 c.1095C>T p.R365R Chr.11: 70331977-70331977 on Build GRCh38
690166 c.1198G>C p.A400P Chr.11: 70332080-70332080 on Build GRCh38
3870081 c.1208C>T p.S403F Chr.11: 70332090-70332090 on Build GRCh38
6753295 c.1094G>A p.R365H Chr.11: 70331976-70331976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143350650 Chr.11: 70331967-70331967 on Build GRCh38
rs769570871 Chr.11: 70332137-70332138 on Build GRCh38
rs185874729 Chr.11: 70332290-70332290 on Build GRCh38
rs778785947 Chr.11: 70332037-70332037 on Build GRCh38
rs1024904894 Chr.11: 70331948-70331948 on Build GRCh38
rs768098552 Chr.11: 70332121-70332121 on Build GRCh38
rs976209163 Chr.11: 70331841-70331841 on Build GRCh38
rs749147605 Chr.11: 70332019-70332019 on Build GRCh38
rs776719992 Chr.11: 70332057-70332057 on Build GRCh38
rs749886519 Chr.11: 70332004-70332004 on Build GRCh38
rs544092093 Chr.11: 70332210-70332210 on Build GRCh38
rs1017438545 Chr.11: 70332161-70332161 on Build GRCh38
rs763353351 Chr.11: 70331972-70331972 on Build GRCh38
rs770971864 Chr.11: 70332126-70332126 on Build GRCh38
rs770050858 Chr.11: 70332061-70332061 on Build GRCh38
rs757300030 Chr.11: 70332080-70332080 on Build GRCh38
rs574282374 Chr.11: 70332280-70332280 on Build GRCh38
rs772831089 Chr.11: 70332034-70332034 on Build GRCh38
rs765399361 Chr.11: 70331944-70331944 on Build GRCh38
rs774523775 Chr.11: 70332111-70332111 on Build GRCh38
rs924817783 Chr.11: 70331875-70331875 on Build GRCh38
rs763293373 Chr.11: 70332094-70332094 on Build GRCh38
rs997629750 Chr.11: 70332311-70332311 on Build GRCh38
rs201954774 Chr.11: 70331968-70331968 on Build GRCh38
rs576599150 Chr.11: 70332209-70332209 on Build GRCh38
rs376830476 Chr.11: 70331991-70331991 on Build GRCh38
rs769049622 Chr.11: 70332103-70332103 on Build GRCh38
rs79783439 Chr.11: 70332008-70332008 on Build GRCh38
rs553765591 Chr.11: 70331976-70331976 on Build GRCh38
rs771835705 Chr.11: 70332049-70332049 on Build GRCh38
rs756776453 Chr.11: 70331989-70331989 on Build GRCh38
rs977536582 Chr.11: 70332281-70332281 on Build GRCh38
rs755862919 Chr.11: 70332009-70332009 on Build GRCh38
rs760244042 Chr.11: 70332123-70332123 on Build GRCh38
rs779808412 Chr.11: 70332016-70332016 on Build GRCh38
rs202040644 Chr.11: 70332043-70332043 on Build GRCh38
rs1014399286 Chr.11: 70331916-70331916 on Build GRCh38
rs567556678 Chr.11: 70331894-70331894 on Build GRCh38
rs138104472 Chr.11: 70332252-70332252 on Build GRCh38
rs764547561 Chr.11: 70331975-70331975 on Build GRCh38
rs207471999 Chr.11: 70331922-70331922 on Build GRCh38
rs774288844 Chr.11: 70331881-70331881 on Build GRCh38
rs939094451 Chr.11: 70332093-70332093 on Build GRCh38
rs946135535 Chr.11: 70332266-70332266 on Build GRCh38
rs200901690 Chr.11: 70332060-70332060 on Build GRCh38
rs767149740 Chr.11: 70331990-70331990 on Build GRCh38
rs762388882 Chr.11: 70332115-70332115 on Build GRCh38
rs775657756 Chr.11: 70332085-70332085 on Build GRCh38
rs557914953 Chr.11: 70332122-70332122 on Build GRCh38
rs754774570 Chr.11: 70332026-70332026 on Build GRCh38
rs776631972 Chr.11: 70331927-70331927 on Build GRCh38

View Full Product Details