Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219246534-219247076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STK16
Entrez Gene ID
8576
Gene Name
serine/threonine kinase 16
Gene Aliases
KRCT, MPSK, PKL12, TSF1
UniGene
Hs.153003
Gene Chromosome Location
Chr.2: 219245469-219250337 on Build GRCh38
Transcripts
NM_001008910.2 NR_026909.1 XM_011512067.2 XM_017005160.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6499084 c.305_306+19del21 p.? Chr.2: 219246875-219246895 on Build GRCh38
5107375 c.176G>A p.R59Q Chr.2: 219246746-219246746 on Build GRCh38
3743604 c.189G>A p.Q63Q Chr.2: 219246759-219246759 on Build GRCh38
3044762 c.209G>A p.R70H Chr.2: 219246779-219246779 on Build GRCh38
4627988 c.239T>C p.L80P Chr.2: 219246809-219246809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754595646 Chr.2: 219246572-219246573 on Build GRCh38
rs752479408 Chr.2: 219246766-219246768 on Build GRCh38
rs751944503 Chr.2: 219246654-219246654 on Build GRCh38
rs371178580 Chr.2: 219246746-219246746 on Build GRCh38
rs780851050 Chr.2: 219246702-219246702 on Build GRCh38
rs1055448357 Chr.2: 219246650-219246650 on Build GRCh38
rs902311240 Chr.2: 219246969-219246969 on Build GRCh38
rs548466076 Chr.2: 219246719-219246719 on Build GRCh38
rs761797547 Chr.2: 219246605-219246605 on Build GRCh38
rs34799131 Chr.2: 219246692-219246692 on Build GRCh38
rs970538809 Chr.2: 219246820-219246820 on Build GRCh38
rs757109237 Chr.2: 219246904-219246904 on Build GRCh38
rs577198736 Chr.2: 219247032-219247032 on Build GRCh38
rs777644163 Chr.2: 219246880-219246883 on Build GRCh38
rs200058316 Chr.2: 219246779-219246779 on Build GRCh38
rs777487052 Chr.2: 219246685-219246685 on Build GRCh38
rs190018521 Chr.2: 219246660-219246660 on Build GRCh38
rs772033553 Chr.2: 219246860-219246860 on Build GRCh38
rs374274331 Chr.2: 219246585-219246585 on Build GRCh38
rs775434668 Chr.2: 219246893-219246893 on Build GRCh38
rs200267587 Chr.2: 219246638-219246638 on Build GRCh38
rs755419224 Chr.2: 219246575-219246575 on Build GRCh38
rs779716259 Chr.2: 219246920-219246920 on Build GRCh38
rs757094373 Chr.2: 219246788-219246788 on Build GRCh38
rs767787780 Chr.2: 219246667-219246667 on Build GRCh38
rs893708636 Chr.2: 219246927-219246927 on Build GRCh38
rs1055078111 Chr.2: 219246916-219246916 on Build GRCh38
rs375565820 Chr.2: 219246760-219246760 on Build GRCh38
rs371377253 Chr.2: 219246792-219246792 on Build GRCh38
rs748581772 Chr.2: 219246821-219246821 on Build GRCh38
rs529660363 Chr.2: 219246806-219246806 on Build GRCh38
rs201945391 Chr.2: 219246665-219246665 on Build GRCh38
rs937944720 Chr.2: 219246649-219246649 on Build GRCh38
rs759691946 Chr.2: 219246891-219246891 on Build GRCh38
rs778342281 Chr.2: 219246763-219246763 on Build GRCh38
rs559500604 Chr.2: 219246708-219246708 on Build GRCh38
rs777429921 Chr.2: 219246598-219246598 on Build GRCh38
rs538331418 Chr.2: 219246832-219246832 on Build GRCh38
rs188193002 Chr.2: 219246965-219246965 on Build GRCh38
rs764437002 Chr.2: 219246641-219246641 on Build GRCh38
rs377106870 Chr.2: 219246818-219246818 on Build GRCh38
rs558730425 Chr.2: 219246864-219246864 on Build GRCh38
rs200977899 Chr.2: 219246705-219246705 on Build GRCh38
rs752722187 Chr.2: 219246681-219246681 on Build GRCh38
rs200301017 Chr.2: 219246916-219246917 on Build GRCh38
rs758350335 Chr.2: 219246684-219246684 on Build GRCh38
rs1029825821 Chr.2: 219247040-219247040 on Build GRCh38
rs185901280 Chr.2: 219246940-219246940 on Build GRCh38
rs756287095 Chr.2: 219246674-219246674 on Build GRCh38
rs34282267 Chr.2: 219246799-219246799 on Build GRCh38
rs1038745089 Chr.2: 219247000-219247000 on Build GRCh38
rs201550308 Chr.2: 219246811-219246811 on Build GRCh38
rs781040637 Chr.2: 219246746-219246746 on Build GRCh38
rs751560096 Chr.2: 219246898-219246898 on Build GRCh38
rs780260514 Chr.2: 219246675-219246675 on Build GRCh38
rs770564404 Chr.2: 219246696-219246696 on Build GRCh38
rs769251333 Chr.2: 219246636-219246636 on Build GRCh38
rs35947471 Chr.2: 219246733-219246733 on Build GRCh38
rs763424055 Chr.2: 219246640-219246640 on Build GRCh38
rs889495053 Chr.2: 219247056-219247056 on Build GRCh38
rs929419920 Chr.2: 219246886-219246886 on Build GRCh38
rs543764276 Chr.2: 219247005-219247005 on Build GRCh38
rs61733614 Chr.2: 219246732-219246732 on Build GRCh38
rs754566157 Chr.2: 219246928-219246928 on Build GRCh38
rs202239732 Chr.2: 219246718-219246718 on Build GRCh38
rs751933081 Chr.2: 219246569-219246569 on Build GRCh38
rs746705600 Chr.2: 219246583-219246583 on Build GRCh38
rs372905234 Chr.2: 219246810-219246810 on Build GRCh38
rs374247901 Chr.2: 219246668-219246668 on Build GRCh38
rs557061469 Chr.2: 219247016-219247016 on Build GRCh38
rs180854978 Chr.2: 219246905-219246905 on Build GRCh38
rs751169442 Chr.2: 219246573-219246573 on Build GRCh38
rs368802233 Chr.2: 219246836-219246836 on Build GRCh38
rs898920348 Chr.2: 219247045-219247045 on Build GRCh38
rs919402776 Chr.2: 219246838-219246838 on Build GRCh38
rs764096412 Chr.2: 219246778-219246778 on Build GRCh38
rs546314239 Chr.2: 219246706-219246706 on Build GRCh38
rs755842078 Chr.2: 219246918-219246918 on Build GRCh38
rs780940834 Chr.2: 219246789-219246789 on Build GRCh38
rs754954645 Chr.2: 219246768-219246768 on Build GRCh38
rs369674665 Chr.2: 219246805-219246805 on Build GRCh38
rs756010846 Chr.2: 219246769-219246770 on Build GRCh38
rs200583155 Chr.2: 219246887-219246887 on Build GRCh38
rs372257860 Chr.2: 219246892-219246892 on Build GRCh38
rs770602003 Chr.2: 219246589-219246589 on Build GRCh38
rs761072103 Chr.2: 219246926-219246926 on Build GRCh38
rs35916728 Chr.2: 219246761-219246761 on Build GRCh38
rs750235342 Chr.2: 219246914-219246914 on Build GRCh38
rs777591979 Chr.2: 219246576-219246576 on Build GRCh38

View Full Product Details