Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 96332318-96332743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR2F2
Entrez Gene ID
7026
Gene Name
nuclear receptor subfamily 2 group F member 2
Gene Aliases
ARP1, CHTD4, COUPTFB, COUPTFII, NF-E3, NR2F1, SVP40, TFCOUP2
UniGene
Hs.347991
Gene Chromosome Location
Chr.15: 96325928-96340263 on Build GRCh38
Transcripts
NM_001145155.1 NM_001145156.1 NM_021005.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
198670 c.322C>T p.R108W Chr.15: 96332427-96332427 on Build GRCh38
4058055 c.222G>A p.Q74Q Chr.15: 96332327-96332327 on Build GRCh38
5807356 c.289G>A p.E97K Chr.15: 96332394-96332394 on Build GRCh38
7194223 c.323G>T p.R108L Chr.15: 96332428-96332428 on Build GRCh38
2141209 c.225A>G p.Q75Q Chr.15: 96332330-96332330 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781377128 Chr.15: 96332576-96332576 on Build GRCh38
rs757445345 Chr.15: 96332369-96332369 on Build GRCh38
rs765228973 Chr.15: 96332553-96332553 on Build GRCh38
rs1006257933 Chr.15: 96332653-96332653 on Build GRCh38
rs753715153 Chr.15: 96332339-96332339 on Build GRCh38
rs1017084842 Chr.15: 96332665-96332665 on Build GRCh38
rs767059013 Chr.15: 96332454-96332454 on Build GRCh38
rs972481631 Chr.15: 96332375-96332375 on Build GRCh38
rs761260932 Chr.15: 96332447-96332447 on Build GRCh38
rs770026575 Chr.15: 96332387-96332387 on Build GRCh38
rs961553941 Chr.15: 96332680-96332680 on Build GRCh38
rs1017555984 Chr.15: 96332351-96332351 on Build GRCh38
rs945423018 Chr.15: 96332599-96332599 on Build GRCh38
rs774866322 Chr.15: 96332444-96332444 on Build GRCh38
rs137861279 Chr.15: 96332462-96332462 on Build GRCh38
rs756420335 Chr.15: 96332593-96332593 on Build GRCh38
rs1050199711 Chr.15: 96332626-96332626 on Build GRCh38
rs372651988 Chr.15: 96332372-96332372 on Build GRCh38
rs766073630 Chr.15: 96332471-96332471 on Build GRCh38
rs761415286 Chr.15: 96332703-96332703 on Build GRCh38
rs775993510 Chr.15: 96332426-96332426 on Build GRCh38
rs916901985 Chr.15: 96332379-96332379 on Build GRCh38
rs373598393 Chr.15: 96332354-96332354 on Build GRCh38
rs750610514 Chr.15: 96332588-96332588 on Build GRCh38
rs371267056 Chr.15: 96332468-96332468 on Build GRCh38
rs998148245 Chr.15: 96332619-96332619 on Build GRCh38
rs199765586 Chr.15: 96332438-96332438 on Build GRCh38
rs769096071 Chr.15: 96332440-96332440 on Build GRCh38
rs905319292 Chr.15: 96332677-96332677 on Build GRCh38
rs901269508 Chr.15: 96332603-96332603 on Build GRCh38
rs1045116361 Chr.15: 96332657-96332657 on Build GRCh38
rs150966894 Chr.15: 96332602-96332602 on Build GRCh38
rs369494475 Chr.15: 96332342-96332342 on Build GRCh38
rs369256802 Chr.15: 96332366-96332366 on Build GRCh38
rs1011053780 Chr.15: 96332631-96332631 on Build GRCh38
rs752703752 Chr.15: 96332561-96332561 on Build GRCh38
rs935924408 Chr.15: 96332510-96332510 on Build GRCh38
rs550952784 Chr.15: 96332621-96332621 on Build GRCh38
rs759411357 Chr.15: 96332552-96332552 on Build GRCh38
rs1003496700 Chr.15: 96332691-96332691 on Build GRCh38
rs760443433 Chr.15: 96332465-96332465 on Build GRCh38
rs890038161 Chr.15: 96332639-96332639 on Build GRCh38
rs994309490 Chr.15: 96332681-96332681 on Build GRCh38
rs746152342 Chr.15: 96332378-96332378 on Build GRCh38
rs757391827 Chr.15: 96332575-96332575 on Build GRCh38
rs1023985917 Chr.15: 96332719-96332719 on Build GRCh38

View Full Product Details