Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55470599-55471198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VOPP1
Entrez Gene ID
81552
Gene Name
vesicular, overexpressed in cancer, prosurvival protein 1
Gene Aliases
ECOP, GASP
UniGene
Hs.488307
Gene Chromosome Location
Chr.7: 55434406-55572520 on Build GRCh38
Transcripts
NM_001284282.1 NM_001284283.1 NM_001284284.1 NM_001321242.1 NM_001321243.1 NM_001321244.1 NM_001321246.1 NM_001321247.1 NM_001321248.1 NM_001321249.1 NM_001321250.1 NM_001321251.1 NM_030796.4 XM_011515539.1 XM_011515540.2 XM_011515541.2 XM_011515546.2

SNP Detail

SNP ID Chromosome Location
rs538636164 Chr.7: 55471019-55471019 on Build GRCh38
rs929209524 Chr.7: 55470795-55470795 on Build GRCh38
rs6620 Chr.7: 55470847-55470847 on Build GRCh38
rs3894996 Chr.7: 55470984-55470984 on Build GRCh38
rs1012564762 Chr.7: 55470855-55470855 on Build GRCh38
rs192022943 Chr.7: 55470965-55470965 on Build GRCh38
rs563883730 Chr.7: 55471045-55471045 on Build GRCh38
rs372107807 Chr.7: 55471008-55471008 on Build GRCh38
rs3211244 Chr.7: 55470851-55470851 on Build GRCh38
rs986111076 Chr.7: 55470716-55470716 on Build GRCh38
rs967980795 Chr.7: 55470690-55470690 on Build GRCh38
rs576289191 Chr.7: 55470752-55470752 on Build GRCh38
rs777965978 Chr.7: 55470747-55470747 on Build GRCh38
rs976229170 Chr.7: 55471161-55471161 on Build GRCh38
rs919138543 Chr.7: 55470788-55470788 on Build GRCh38
rs3894999 Chr.7: 55470683-55470683 on Build GRCh38
rs986083089 Chr.7: 55471072-55471072 on Build GRCh38
rs185438958 Chr.7: 55470954-55470954 on Build GRCh38
rs3894997 Chr.7: 55470744-55470744 on Build GRCh38
rs906932635 Chr.7: 55470938-55470938 on Build GRCh38
rs1048938025 Chr.7: 55470803-55470803 on Build GRCh38
rs1040594558 Chr.7: 55470761-55470761 on Build GRCh38
rs577310075 Chr.7: 55471007-55471007 on Build GRCh38
rs545230178 Chr.7: 55471016-55471016 on Build GRCh38
rs369812404 Chr.7: 55470969-55470969 on Build GRCh38
rs34224092 Chr.7: 55470743-55470744 on Build GRCh38
rs1044111307 Chr.7: 55470930-55470930 on Build GRCh38
rs1035590108 Chr.7: 55470945-55470945 on Build GRCh38
rs966099620 Chr.7: 55471157-55471157 on Build GRCh38
rs1002662770 Chr.7: 55470939-55470939 on Build GRCh38
rs558945623 Chr.7: 55471049-55471049 on Build GRCh38
rs530984064 Chr.7: 55471135-55471135 on Build GRCh38
rs537375847 Chr.7: 55470753-55470753 on Build GRCh38
rs34003501 Chr.7: 55470800-55470801 on Build GRCh38
rs910605957 Chr.7: 55470748-55470748 on Build GRCh38
rs367632994 Chr.7: 55470719-55470719 on Build GRCh38
rs3894998 Chr.7: 55470714-55470714 on Build GRCh38
rs910578142 Chr.7: 55471149-55471149 on Build GRCh38

View Full Product Details