Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 15470449-15470991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSIP1
Entrez Gene ID
11168
Gene Name
PC4 and SFRS1 interacting protein 1
Gene Aliases
DFS70, LEDGF, PAIP, PSIP2, p52, p75
UniGene
Hs.658434
Gene Chromosome Location
Chr.9: 15464066-15511019 on Build GRCh38
Transcripts
NM_001128217.2 NM_001317898.1 NM_001317900.1 NM_021144.3 NM_033222.4

SNP Detail

SNP ID Chromosome Location
rs911242228 Chr.9: 15470821-15470821 on Build GRCh38
rs893415270 Chr.9: 15470946-15470946 on Build GRCh38
rs538356038 Chr.9: 15470604-15470604 on Build GRCh38
rs570041063 Chr.9: 15470477-15470477 on Build GRCh38
rs900274845 Chr.9: 15470656-15470657 on Build GRCh38
rs931749007 Chr.9: 15470878-15470878 on Build GRCh38
rs756095575 Chr.9: 15470935-15470937 on Build GRCh38
rs673611 Chr.9: 15470854-15470854 on Build GRCh38
rs753768892 Chr.9: 15470607-15470607 on Build GRCh38
rs913171638 Chr.9: 15470825-15470825 on Build GRCh38
rs938692133 Chr.9: 15470814-15470814 on Build GRCh38
rs920543662 Chr.9: 15470860-15470860 on Build GRCh38
rs775882604 Chr.9: 15470643-15470643 on Build GRCh38
rs928648936 Chr.9: 15470635-15470638 on Build GRCh38
rs1047618951 Chr.9: 15470863-15470863 on Build GRCh38
rs673665 Chr.9: 15470886-15470886 on Build GRCh38
rs573641623 Chr.9: 15470935-15470936 on Build GRCh38
rs970109680 Chr.9: 15470764-15470764 on Build GRCh38
rs137933815 Chr.9: 15470605-15470605 on Build GRCh38
rs185641879 Chr.9: 15470645-15470645 on Build GRCh38
rs886259894 Chr.9: 15470879-15470879 on Build GRCh38
rs930439214 Chr.9: 15470558-15470558 on Build GRCh38
rs905498185 Chr.9: 15470626-15470626 on Build GRCh38
rs1010477083 Chr.9: 15470954-15470954 on Build GRCh38
rs897397984 Chr.9: 15470957-15470957 on Build GRCh38
rs749868843 Chr.9: 15470759-15470759 on Build GRCh38
rs988707237 Chr.9: 15470820-15470820 on Build GRCh38
rs1020425490 Chr.9: 15470955-15470955 on Build GRCh38
rs191851 Chr.9: 15470788-15470788 on Build GRCh38
rs673645 Chr.9: 15470875-15470875 on Build GRCh38
rs1027183025 Chr.9: 15470753-15470756 on Build GRCh38
rs961515151 Chr.9: 15470728-15470728 on Build GRCh38
rs543615582 Chr.9: 15470724-15470724 on Build GRCh38
rs571291323 Chr.9: 15470527-15470527 on Build GRCh38
rs169330 Chr.9: 15470686-15470686 on Build GRCh38
rs995876640 Chr.9: 15470494-15470494 on Build GRCh38
rs748575925 Chr.9: 15470936-15470937 on Build GRCh38
rs192441472 Chr.9: 15470707-15470707 on Build GRCh38
rs990915046 Chr.9: 15470959-15470959 on Build GRCh38
rs996357353 Chr.9: 15470696-15470698 on Build GRCh38
rs190509312 Chr.9: 15470646-15470646 on Build GRCh38
rs930575215 Chr.9: 15470862-15470862 on Build GRCh38
rs113391732 Chr.9: 15470778-15470778 on Build GRCh38
rs2821532 Chr.9: 15470960-15470960 on Build GRCh38
rs537098900 Chr.9: 15470721-15470722 on Build GRCh38
rs938606919 Chr.9: 15470635-15470635 on Build GRCh38
rs986820859 Chr.9: 15470799-15470799 on Build GRCh38
rs1015825181 Chr.9: 15470953-15470953 on Build GRCh38
rs890157958 Chr.9: 15470718-15470718 on Build GRCh38
rs556773913 Chr.9: 15470892-15470894 on Build GRCh38
rs928642906 Chr.9: 15470806-15470806 on Build GRCh38
rs1032669767 Chr.9: 15470945-15470945 on Build GRCh38
rs537230418 Chr.9: 15470513-15470513 on Build GRCh38
rs530014453 Chr.9: 15470898-15470898 on Build GRCh38
rs905529611 Chr.9: 15470930-15470930 on Build GRCh38
rs567145334 Chr.9: 15470957-15470958 on Build GRCh38
rs995780752 Chr.9: 15470958-15470958 on Build GRCh38
rs1013995549 Chr.9: 15470623-15470623 on Build GRCh38
rs989416205 Chr.9: 15470746-15470746 on Build GRCh38
rs1027150521 Chr.9: 15470964-15470964 on Build GRCh38
rs558078032 Chr.9: 15470542-15470542 on Build GRCh38
rs955637981 Chr.9: 15470519-15470519 on Build GRCh38
rs150302178 Chr.9: 15470523-15470523 on Build GRCh38
rs947226413 Chr.9: 15470645-15470646 on Build GRCh38
rs193362 Chr.9: 15470743-15470743 on Build GRCh38
rs942658627 Chr.9: 15470849-15470849 on Build GRCh38
rs869056513 Chr.9: 15470954-15470958 on Build GRCh38
rs890087301 Chr.9: 15470947-15470948 on Build GRCh38
rs1004973790 Chr.9: 15470950-15470950 on Build GRCh38
rs1053478541 Chr.9: 15470636-15470636 on Build GRCh38
rs1029980760 Chr.9: 15470703-15470703 on Build GRCh38
rs757937225 Chr.9: 15470937-15470946 on Build GRCh38
rs980112100 Chr.9: 15470621-15470621 on Build GRCh38
rs961545397 Chr.9: 15470552-15470552 on Build GRCh38
rs899002376 Chr.9: 15470546-15470546 on Build GRCh38
rs778011596 Chr.9: 15470729-15470729 on Build GRCh38
rs1039342064 Chr.9: 15470541-15470541 on Build GRCh38
rs2737834 Chr.9: 15470624-15470624 on Build GRCh38
rs764936328 Chr.9: 15470700-15470700 on Build GRCh38
rs577225262 Chr.9: 15470797-15470797 on Build GRCh38
rs541156527 Chr.9: 15470817-15470817 on Build GRCh38
rs562156713 Chr.9: 15470730-15470733 on Build GRCh38
rs1042858751 Chr.9: 15470847-15470847 on Build GRCh38
rs949794455 Chr.9: 15470884-15470884 on Build GRCh38
rs184442409 Chr.9: 15470830-15470830 on Build GRCh38
rs565677691 Chr.9: 15470629-15470629 on Build GRCh38
rs2665508 Chr.9: 15470549-15470549 on Build GRCh38
rs541853029 Chr.9: 15470935-15470935 on Build GRCh38
rs374899833 Chr.9: 15470502-15470502 on Build GRCh38
rs974697213 Chr.9: 15470850-15470850 on Build GRCh38
rs948671713 Chr.9: 15470612-15470612 on Build GRCh38
rs58019501 Chr.9: 15470958-15470958 on Build GRCh38

View Full Product Details