Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 167792493-167793058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINI1
Entrez Gene ID
5274
Gene Name
serpin family I member 1
Gene Aliases
PI12, neuroserpin
UniGene
Hs.478153
Gene Chromosome Location
Chr.3: 167735335-167825569 on Build GRCh38
Transcripts
NM_001122752.1 NM_005025.4 XM_017006618.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4763421 c.493G>T p.D165Y Chr.3: 167792601-167792601 on Build GRCh38
6163994 c.562G>T p.G188W Chr.3: 167792670-167792670 on Build GRCh38
164270 c.622G>A p.D208N Chr.3: 167792730-167792730 on Build GRCh38
729215 c.653A>G p.Y218C Chr.3: 167792761-167792761 on Build GRCh38
1632965 c.584G>T p.R195M Chr.3: 167792692-167792692 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774989939 Chr.3: 167792616-167792616 on Build GRCh38
rs745438717 Chr.3: 167792716-167792716 on Build GRCh38
rs907746140 Chr.3: 167792639-167792639 on Build GRCh38
rs762427588 Chr.3: 167792634-167792634 on Build GRCh38
rs191476872 Chr.3: 167792829-167792829 on Build GRCh38
rs746082671 Chr.3: 167792569-167792569 on Build GRCh38
rs370446701 Chr.3: 167792690-167792690 on Build GRCh38
rs746668637 Chr.3: 167792744-167792744 on Build GRCh38
rs1023900014 Chr.3: 167792934-167792934 on Build GRCh38
rs61761892 Chr.3: 167792704-167792704 on Build GRCh38
rs766752335 Chr.3: 167792879-167792879 on Build GRCh38
rs778458057 Chr.3: 167792670-167792670 on Build GRCh38
rs182990298 Chr.3: 167792964-167792964 on Build GRCh38
rs750601672 Chr.3: 167792657-167792657 on Build GRCh38
rs755188629 Chr.3: 167792679-167792679 on Build GRCh38
rs773900198 Chr.3: 167792584-167792584 on Build GRCh38
rs971680054 Chr.3: 167792949-167792949 on Build GRCh38
rs965855763 Chr.3: 167793013-167793013 on Build GRCh38
rs772199277 Chr.3: 167792574-167792575 on Build GRCh38
rs9790152 Chr.3: 167792915-167792915 on Build GRCh38
rs148829024 Chr.3: 167792989-167792989 on Build GRCh38
rs2229697 Chr.3: 167792684-167792684 on Build GRCh38
rs765811460 Chr.3: 167792638-167792638 on Build GRCh38
rs905417561 Chr.3: 167792963-167792963 on Build GRCh38
rs778816229 Chr.3: 167792710-167792710 on Build GRCh38
rs749255048 Chr.3: 167792569-167792569 on Build GRCh38
rs2420038 Chr.3: 167792570-167792570 on Build GRCh38
rs868562091 Chr.3: 167792976-167792976 on Build GRCh38
rs901775636 Chr.3: 167792649-167792649 on Build GRCh38
rs747025290 Chr.3: 167792792-167792792 on Build GRCh38
rs976025763 Chr.3: 167792693-167792693 on Build GRCh38
rs763453312 Chr.3: 167792801-167792801 on Build GRCh38
rs768902962 Chr.3: 167792620-167792620 on Build GRCh38
rs142316110 Chr.3: 167792731-167792731 on Build GRCh38
rs745350666 Chr.3: 167792594-167792594 on Build GRCh38
rs958440759 Chr.3: 167792967-167792967 on Build GRCh38
rs768301457 Chr.3: 167792755-167792755 on Build GRCh38
rs879780984 Chr.3: 167792962-167792962 on Build GRCh38
rs763836063 Chr.3: 167792820-167792820 on Build GRCh38
rs1019616009 Chr.3: 167793000-167793000 on Build GRCh38
rs76236854 Chr.3: 167792855-167792855 on Build GRCh38
rs780104028 Chr.3: 167792583-167792583 on Build GRCh38
rs773872160 Chr.3: 167792793-167792793 on Build GRCh38
rs1027425377 Chr.3: 167792975-167792975 on Build GRCh38
rs769165272 Chr.3: 167792814-167792815 on Build GRCh38
rs755876538 Chr.3: 167792555-167792555 on Build GRCh38
rs771822255 Chr.3: 167792607-167792607 on Build GRCh38
rs777559077 Chr.3: 167792558-167792558 on Build GRCh38
rs777027183 Chr.3: 167792898-167792898 on Build GRCh38
rs762267952 Chr.3: 167792778-167792778 on Build GRCh38
rs777179641 Chr.3: 167792771-167792771 on Build GRCh38
rs61735309 Chr.3: 167792626-167792626 on Build GRCh38
rs199813213 Chr.3: 167792831-167792831 on Build GRCh38
rs766402832 Chr.3: 167792805-167792805 on Build GRCh38
rs751609680 Chr.3: 167792811-167792811 on Build GRCh38
rs566381455 Chr.3: 167792674-167792674 on Build GRCh38
rs1014132720 Chr.3: 167792928-167792928 on Build GRCh38
rs989770824 Chr.3: 167792533-167792533 on Build GRCh38
rs866041653 Chr.3: 167792770-167792770 on Build GRCh38
rs770484017 Chr.3: 167792783-167792783 on Build GRCh38
rs763115467 Chr.3: 167792659-167792659 on Build GRCh38
rs903501917 Chr.3: 167792854-167792854 on Build GRCh38
rs760314901 Chr.3: 167792618-167792618 on Build GRCh38
rs879704918 Chr.3: 167792940-167792940 on Build GRCh38
rs549900839 Chr.3: 167792545-167792545 on Build GRCh38
rs889837897 Chr.3: 167792727-167792727 on Build GRCh38
rs568173360 Chr.3: 167792546-167792546 on Build GRCh38
rs777452195 Chr.3: 167792683-167792683 on Build GRCh38
rs1052177511 Chr.3: 167792896-167792896 on Build GRCh38

View Full Product Details