Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 19060446-19060947 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRGPRX2
Entrez Gene ID
117194
Gene Name
MAS related GPR family member X2
Gene Aliases
MGRG3, MRGX2
UniGene
Hs.350566
Gene Chromosome Location
Chr.11: 19054455-19060681 on Build GRCh38
Transcripts
NM_054030.3

SNP Detail

SNP ID Chromosome Location
rs926465921 Chr.11: 19060890-19060891 on Build GRCh38
rs1045190367 Chr.11: 19060557-19060557 on Build GRCh38
rs184164639 Chr.11: 19060896-19060896 on Build GRCh38
rs982433729 Chr.11: 19060917-19060917 on Build GRCh38
rs1503503 Chr.11: 19060482-19060482 on Build GRCh38
rs545541414 Chr.11: 19060819-19060819 on Build GRCh38
rs141058362 Chr.11: 19060838-19060838 on Build GRCh38
rs560781806 Chr.11: 19060909-19060909 on Build GRCh38
rs869230698 Chr.11: 19060570-19060573 on Build GRCh38
rs548641324 Chr.11: 19060506-19060506 on Build GRCh38
rs949119632 Chr.11: 19060549-19060549 on Build GRCh38
rs917692247 Chr.11: 19060516-19060516 on Build GRCh38
rs369810845 Chr.11: 19060910-19060910 on Build GRCh38
rs753317631 Chr.11: 19060863-19060863 on Build GRCh38
rs956596261 Chr.11: 19060484-19060484 on Build GRCh38
rs754383674 Chr.11: 19060499-19060499 on Build GRCh38
rs571798773 Chr.11: 19060705-19060705 on Build GRCh38
rs201667509 Chr.11: 19060570-19060570 on Build GRCh38
rs200677900 Chr.11: 19060572-19060572 on Build GRCh38
rs1054046702 Chr.11: 19060644-19060644 on Build GRCh38
rs75352951 Chr.11: 19060677-19060677 on Build GRCh38
rs71987524 Chr.11: 19060569-19060572 on Build GRCh38
rs190881633 Chr.11: 19060895-19060895 on Build GRCh38
rs939207842 Chr.11: 19060899-19060899 on Build GRCh38
rs982405647 Chr.11: 19060507-19060507 on Build GRCh38
rs113460758 Chr.11: 19060880-19060880 on Build GRCh38
rs915120903 Chr.11: 19060884-19060884 on Build GRCh38
rs538252500 Chr.11: 19060471-19060471 on Build GRCh38
rs1029845330 Chr.11: 19060864-19060864 on Build GRCh38
rs561754718 Chr.11: 19060616-19060616 on Build GRCh38
rs1044879346 Chr.11: 19060581-19060581 on Build GRCh38
rs987775407 Chr.11: 19060883-19060883 on Build GRCh38
rs939445801 Chr.11: 19060637-19060637 on Build GRCh38
rs966640294 Chr.11: 19060852-19060852 on Build GRCh38
rs759998313 Chr.11: 19060676-19060676 on Build GRCh38
rs764246289 Chr.11: 19060925-19060925 on Build GRCh38
rs567037899 Chr.11: 19060604-19060604 on Build GRCh38
rs143283907 Chr.11: 19060848-19060848 on Build GRCh38
rs974310764 Chr.11: 19060886-19060886 on Build GRCh38
rs147510458 Chr.11: 19060885-19060885 on Build GRCh38
rs535272527 Chr.11: 19060666-19060666 on Build GRCh38
rs1017778112 Chr.11: 19060851-19060851 on Build GRCh38
rs1049306688 Chr.11: 19060807-19060807 on Build GRCh38
rs915172762 Chr.11: 19060521-19060521 on Build GRCh38
rs974024268 Chr.11: 19060472-19060472 on Build GRCh38
rs993507737 Chr.11: 19060775-19060775 on Build GRCh38

View Full Product Details