Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 18338323-18338772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIK3C2G
Entrez Gene ID
5288
Gene Name
phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 gamma
Gene Aliases
PI3K-C2-gamma, PI3K-C2GAMMA
UniGene
Hs.22500
Gene Chromosome Location
Chr.12: 18242946-18716861 on Build GRCh38
Transcripts
NM_001288772.1 NM_001288774.1 NM_004570.5 XM_011520696.2 XM_011520697.2 XM_011520700.2 XM_011520701.2 XM_017019470.1 XM_017019471.1 XM_017019472.1 XM_017019473.1 XM_017019474.1 XM_017019475.1 XM_017019476.1 XM_017019477.1 XM_017019478.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6922851 c.1273G>A p.E425K Chr.12: 18338426-18338426 on Build GRCh38
6924870 c.1349C>A p.T450K Chr.12: 18338502-18338502 on Build GRCh38
6938286 c.1314C>G p.C438W Chr.12: 18338467-18338467 on Build GRCh38
3458756 c.1332G>T p.V444V Chr.12: 18338485-18338485 on Build GRCh38
4040637 c.1342C>T p.Q448* Chr.12: 18338495-18338495 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780792167 Chr.12: 18338575-18338575 on Build GRCh38
rs7484847 Chr.12: 18338663-18338663 on Build GRCh38
rs773854324 Chr.12: 18338598-18338601 on Build GRCh38
rs1047916770 Chr.12: 18338711-18338711 on Build GRCh38
rs528096287 Chr.12: 18338630-18338630 on Build GRCh38
rs535051237 Chr.12: 18338572-18338572 on Build GRCh38
rs747504141 Chr.12: 18338553-18338553 on Build GRCh38
rs957340169 Chr.12: 18338734-18338746 on Build GRCh38
rs534166409 Chr.12: 18338687-18338687 on Build GRCh38
rs367935508 Chr.12: 18338453-18338453 on Build GRCh38
rs368062288 Chr.12: 18338544-18338544 on Build GRCh38
rs754123698 Chr.12: 18338537-18338537 on Build GRCh38
rs745701167 Chr.12: 18338598-18338599 on Build GRCh38
rs531110246 Chr.12: 18338519-18338519 on Build GRCh38
rs769096176 Chr.12: 18338593-18338594 on Build GRCh38
rs190750617 Chr.12: 18338445-18338445 on Build GRCh38
rs565415797 Chr.12: 18338652-18338652 on Build GRCh38
rs749176032 Chr.12: 18338502-18338502 on Build GRCh38
rs1040512752 Chr.12: 18338688-18338689 on Build GRCh38
rs780145674 Chr.12: 18338349-18338349 on Build GRCh38
rs772909001 Chr.12: 18338662-18338667 on Build GRCh38
rs371052357 Chr.12: 18338393-18338393 on Build GRCh38
rs751221657 Chr.12: 18338566-18338566 on Build GRCh38
rs199613361 Chr.12: 18338664-18338665 on Build GRCh38
rs781421703 Chr.12: 18338611-18338611 on Build GRCh38
rs17847784 Chr.12: 18338431-18338431 on Build GRCh38
rs920761680 Chr.12: 18338677-18338677 on Build GRCh38
rs574503132 Chr.12: 18338654-18338685 on Build GRCh38
rs528621475 Chr.12: 18338414-18338414 on Build GRCh38
rs745430174 Chr.12: 18338595-18338595 on Build GRCh38
rs200273120 Chr.12: 18338525-18338525 on Build GRCh38
rs571849674 Chr.12: 18338486-18338486 on Build GRCh38
rs767606750 Chr.12: 18338390-18338390 on Build GRCh38
rs758728342 Chr.12: 18338492-18338496 on Build GRCh38
rs780601851 Chr.12: 18338521-18338522 on Build GRCh38
rs748140688 Chr.12: 18338665-18338665 on Build GRCh38
rs765546183 Chr.12: 18338565-18338565 on Build GRCh38
rs756781973 Chr.12: 18338432-18338432 on Build GRCh38
rs761927345 Chr.12: 18338376-18338376 on Build GRCh38
rs886719307 Chr.12: 18338748-18338748 on Build GRCh38
rs368644064 Chr.12: 18338421-18338421 on Build GRCh38
rs747980002 Chr.12: 18338491-18338494 on Build GRCh38
rs7484881 Chr.12: 18338669-18338669 on Build GRCh38
rs544423946 Chr.12: 18338666-18338669 on Build GRCh38
rs183348350 Chr.12: 18338643-18338643 on Build GRCh38
rs780884296 Chr.12: 18338435-18338435 on Build GRCh38
rs756613926 Chr.12: 18338395-18338395 on Build GRCh38
rs762053615 Chr.12: 18338508-18338508 on Build GRCh38
rs113984200 Chr.12: 18338373-18338373 on Build GRCh38
rs756975860 Chr.12: 18338574-18338574 on Build GRCh38
rs777524318 Chr.12: 18338424-18338424 on Build GRCh38
rs867884709 Chr.12: 18338661-18338661 on Build GRCh38
rs748712401 Chr.12: 18338595-18338596 on Build GRCh38
rs886686606 Chr.12: 18338377-18338377 on Build GRCh38
rs915481555 Chr.12: 18338558-18338558 on Build GRCh38
rs774582477 Chr.12: 18338507-18338507 on Build GRCh38
rs371333450 Chr.12: 18338464-18338464 on Build GRCh38
rs768736965 Chr.12: 18338503-18338503 on Build GRCh38
rs987520045 Chr.12: 18338609-18338609 on Build GRCh38
rs955465244 Chr.12: 18338582-18338582 on Build GRCh38
rs770640477 Chr.12: 18338596-18338597 on Build GRCh38
rs777345500 Chr.12: 18338595-18338598 on Build GRCh38
rs372044072 Chr.12: 18338664-18338667 on Build GRCh38
rs1049464643 Chr.12: 18338358-18338358 on Build GRCh38
rs773290534 Chr.12: 18338524-18338524 on Build GRCh38
rs375386705 Chr.12: 18338419-18338419 on Build GRCh38
rs770529329 Chr.12: 18338664-18338669 on Build GRCh38
rs7484882 Chr.12: 18338671-18338671 on Build GRCh38
rs756351769 Chr.12: 18338663-18338666 on Build GRCh38

View Full Product Details