Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169594531-169595102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SELP
Entrez Gene ID
6403
Gene Name
selectin P
Gene Aliases
CD62, CD62P, GMP140, GRMP, LECAM3, PADGEM, PSEL
UniGene
Hs.73800
Gene Chromosome Location
Chr.1: 169588849-169630139 on Build GRCh38
Transcripts
NM_003005.3 XM_005245435.1 XM_005245436.3 XM_005245438.1 XM_005245439.1 XM_005245440.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
899311 c.2151C>A p.C717* Chr.1: 169594828-169594828 on Build GRCh38
1472959 c.2126T>C p.V709A Chr.1: 169594853-169594853 on Build GRCh38
899310 c.2201G>A p.C734Y Chr.1: 169594778-169594778 on Build GRCh38
899309 c.2203C>A p.L735I Chr.1: 169594776-169594776 on Build GRCh38
6774438 c.2221A>G p.N741D Chr.1: 169594758-169594758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765608567 Chr.1: 169594761-169594761 on Build GRCh38
rs138392958 Chr.1: 169594722-169594722 on Build GRCh38
rs546626416 Chr.1: 169595010-169595010 on Build GRCh38
rs1040694906 Chr.1: 169594608-169594608 on Build GRCh38
rs1569471 Chr.1: 169594892-169594892 on Build GRCh38
rs748117954 Chr.1: 169594795-169594795 on Build GRCh38
rs984320501 Chr.1: 169595060-169595060 on Build GRCh38
rs563750271 Chr.1: 169594952-169594952 on Build GRCh38
rs1009582799 Chr.1: 169594657-169594657 on Build GRCh38
rs932318960 Chr.1: 169595041-169595041 on Build GRCh38
rs201718876 Chr.1: 169594777-169594777 on Build GRCh38
rs368392089 Chr.1: 169594751-169594751 on Build GRCh38
rs765811479 Chr.1: 169594843-169594843 on Build GRCh38
rs6136 Chr.1: 169594713-169594713 on Build GRCh38
rs751766138 Chr.1: 169594774-169594774 on Build GRCh38
rs758728260 Chr.1: 169594768-169594768 on Build GRCh38
rs779149033 Chr.1: 169594888-169594888 on Build GRCh38
rs757489907 Chr.1: 169594838-169594838 on Build GRCh38
rs546755660 Chr.1: 169594580-169594580 on Build GRCh38
rs751734112 Chr.1: 169594771-169594772 on Build GRCh38
rs954221578 Chr.1: 169594742-169594742 on Build GRCh38
rs1044232974 Chr.1: 169594564-169594564 on Build GRCh38
rs375732703 Chr.1: 169594708-169594708 on Build GRCh38
rs1031426266 Chr.1: 169595073-169595073 on Build GRCh38
rs1000048231 Chr.1: 169594877-169594877 on Build GRCh38
rs998357449 Chr.1: 169594965-169594965 on Build GRCh38
rs528713470 Chr.1: 169594987-169594987 on Build GRCh38
rs745515214 Chr.1: 169594816-169594816 on Build GRCh38
rs13306840 Chr.1: 169594611-169594611 on Build GRCh38
rs755241647 Chr.1: 169594876-169594876 on Build GRCh38
rs750724894 Chr.1: 169594647-169594647 on Build GRCh38
rs904266604 Chr.1: 169595061-169595061 on Build GRCh38
rs772243819 Chr.1: 169594909-169594909 on Build GRCh38
rs199982828 Chr.1: 169594823-169594823 on Build GRCh38
rs759169239 Chr.1: 169594723-169594723 on Build GRCh38
rs1002281710 Chr.1: 169594588-169594588 on Build GRCh38
rs767043661 Chr.1: 169594741-169594741 on Build GRCh38
rs139661252 Chr.1: 169594730-169594730 on Build GRCh38
rs111873260 Chr.1: 169594978-169594978 on Build GRCh38
rs763364922 Chr.1: 169594848-169594848 on Build GRCh38
rs774208816 Chr.1: 169594810-169594810 on Build GRCh38
rs767586113 Chr.1: 169594881-169594881 on Build GRCh38
rs368587128 Chr.1: 169594897-169594897 on Build GRCh38
rs1024223611 Chr.1: 169594770-169594770 on Build GRCh38
rs1000474664 Chr.1: 169595069-169595069 on Build GRCh38
rs747146843 Chr.1: 169594947-169594947 on Build GRCh38
rs742127 Chr.1: 169594929-169594929 on Build GRCh38
rs781336253 Chr.1: 169594794-169594794 on Build GRCh38
rs553177637 Chr.1: 169594783-169594783 on Build GRCh38
rs780979317 Chr.1: 169594612-169594612 on Build GRCh38
rs569148833 Chr.1: 169594688-169594688 on Build GRCh38
rs752853885 Chr.1: 169594887-169594887 on Build GRCh38
rs750814671 Chr.1: 169594762-169594762 on Build GRCh38
rs748112541 Chr.1: 169594696-169594696 on Build GRCh38
rs965382640 Chr.1: 169594957-169594957 on Build GRCh38
rs755060112 Chr.1: 169594784-169594784 on Build GRCh38
rs905735340 Chr.1: 169594585-169594585 on Build GRCh38
rs780207476 Chr.1: 169594675-169594675 on Build GRCh38
rs144853111 Chr.1: 169594799-169594799 on Build GRCh38
rs749226849 Chr.1: 169594911-169594911 on Build GRCh38
rs1001143937 Chr.1: 169594749-169594749 on Build GRCh38
rs539869908 Chr.1: 169594701-169594701 on Build GRCh38
rs1040746909 Chr.1: 169594982-169594982 on Build GRCh38
rs766724007 Chr.1: 169594856-169594856 on Build GRCh38
rs777813301 Chr.1: 169594801-169594801 on Build GRCh38
rs372425190 Chr.1: 169594807-169594807 on Build GRCh38
rs1048472642 Chr.1: 169594573-169594573 on Build GRCh38
rs773685474 Chr.1: 169594846-169594846 on Build GRCh38
rs747129326 Chr.1: 169594678-169594678 on Build GRCh38
rs868212587 Chr.1: 169594959-169594959 on Build GRCh38
rs534934269 Chr.1: 169594755-169594755 on Build GRCh38
rs566756291 Chr.1: 169594707-169594707 on Build GRCh38
rs779335207 Chr.1: 169594645-169594645 on Build GRCh38
rs780301097 Chr.1: 169594769-169594769 on Build GRCh38
rs535510371 Chr.1: 169594621-169594621 on Build GRCh38
rs866453397 Chr.1: 169594770-169594771 on Build GRCh38
rs755078983 Chr.1: 169594790-169594790 on Build GRCh38
rs766575380 Chr.1: 169594678-169594679 on Build GRCh38
rs1028178302 Chr.1: 169594673-169594673 on Build GRCh38
rs754086574 Chr.1: 169594802-169594802 on Build GRCh38
rs1036688172 Chr.1: 169594773-169594773 on Build GRCh38
rs551063011 Chr.1: 169594644-169594644 on Build GRCh38
rs768931017 Chr.1: 169594951-169594951 on Build GRCh38
rs768699281 Chr.1: 169594684-169594684 on Build GRCh38
rs751854308 Chr.1: 169594863-169594863 on Build GRCh38
rs138760541 Chr.1: 169594837-169594837 on Build GRCh38
rs758638080 Chr.1: 169594659-169594659 on Build GRCh38

View Full Product Details