Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 165309058-165309539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCN2A
Entrez Gene ID
6326
Gene Name
sodium voltage-gated channel alpha subunit 2
Gene Aliases
BFIC3, BFIS3, BFNIS, EIEE11, HBA, HBSCI, HBSCII, NAC2, Na(v)1.2, Nav1.2, SCN2A1, SCN2A2
UniGene
Hs.93485
Gene Chromosome Location
Chr.2: 165208056-165392310 on Build GRCh38
Transcripts
NM_001040142.1 NM_001040143.1 NM_021007.2 XM_005246753.3 XM_005246755.4 XM_017004655.1 XM_017004656.1 XM_017004657.1 XM_017004658.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5761404 c.667C>T p.R223* Chr.2: 165309413-165309413 on Build GRCh38
6054356 c.675G>C p.L225F Chr.2: 165309234-165309234 on Build GRCh38
6772993 c.632G>A p.G211D Chr.2: 165309378-165309378 on Build GRCh38
164209 c.607T>C p.Y203H Chr.2: 165309166-165309166 on Build GRCh38
1008567 c.664C>A p.L222I Chr.2: 165309410-165309410 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202186740 Chr.2: 165309121-165309121 on Build GRCh38
rs772746212 Chr.2: 165309489-165309489 on Build GRCh38
rs750284423 Chr.2: 165309329-165309330 on Build GRCh38
rs1043787498 Chr.2: 165309328-165309328 on Build GRCh38
rs149024364 Chr.2: 165309390-165309390 on Build GRCh38
rs778696534 Chr.2: 165309167-165309167 on Build GRCh38
rs267598957 Chr.2: 165309405-165309405 on Build GRCh38
rs761152525 Chr.2: 165309330-165309330 on Build GRCh38
rs2228987 Chr.2: 165309427-165309427 on Build GRCh38
rs879098149 Chr.2: 165309273-165309273 on Build GRCh38
rs1057517854 Chr.2: 165309386-165309386 on Build GRCh38
rs796053203 Chr.2: 165309393-165309393 on Build GRCh38
rs1028102001 Chr.2: 165309114-165309114 on Build GRCh38
rs1028484708 Chr.2: 165309509-165309509 on Build GRCh38
rs372214702 Chr.2: 165309465-165309465 on Build GRCh38
rs773412222 Chr.2: 165309271-165309271 on Build GRCh38
rs558474027 Chr.2: 165309208-165309208 on Build GRCh38
rs754960917 Chr.2: 165309446-165309446 on Build GRCh38
rs2228982 Chr.2: 165309358-165309358 on Build GRCh38
rs781207717 Chr.2: 165309450-165309450 on Build GRCh38
rs116336413 Chr.2: 165309502-165309502 on Build GRCh38
rs763159391 Chr.2: 165309285-165309285 on Build GRCh38
rs1033531496 Chr.2: 165309501-165309501 on Build GRCh38
rs370724112 Chr.2: 165309391-165309391 on Build GRCh38
rs377388579 Chr.2: 165309109-165309109 on Build GRCh38
rs767880177 Chr.2: 165309292-165309292 on Build GRCh38
rs775823242 Chr.2: 165309476-165309476 on Build GRCh38
rs770992144 Chr.2: 165309456-165309456 on Build GRCh38
rs746859046 Chr.2: 165309437-165309437 on Build GRCh38
rs2228986 Chr.2: 165309418-165309418 on Build GRCh38
rs373545585 Chr.2: 165309464-165309464 on Build GRCh38
rs774572427 Chr.2: 165309289-165309289 on Build GRCh38
rs193054811 Chr.2: 165309129-165309129 on Build GRCh38
rs768762425 Chr.2: 165309268-165309268 on Build GRCh38
rs376381203 Chr.2: 165309269-165309269 on Build GRCh38
rs376203810 Chr.2: 165309494-165309494 on Build GRCh38
rs764647930 Chr.2: 165309374-165309374 on Build GRCh38
rs745514055 Chr.2: 165309169-165309169 on Build GRCh38
rs121917752 Chr.2: 165309414-165309414 on Build GRCh38
rs1050583868 Chr.2: 165309209-165309209 on Build GRCh38
rs528200849 Chr.2: 165309507-165309507 on Build GRCh38
rs2228983 Chr.2: 165309364-165309364 on Build GRCh38
rs770918241 Chr.2: 165309286-165309286 on Build GRCh38
rs772333935 Chr.2: 165309466-165309466 on Build GRCh38
rs771844443 Chr.2: 165309194-165309194 on Build GRCh38
rs751863902 Chr.2: 165309120-165309120 on Build GRCh38
rs759696674 Chr.2: 165309290-165309290 on Build GRCh38
rs926762361 Chr.2: 165309345-165309345 on Build GRCh38
rs1038389455 Chr.2: 165309392-165309392 on Build GRCh38
rs777237813 Chr.2: 165309136-165309136 on Build GRCh38
rs756849813 Chr.2: 165309148-165309148 on Build GRCh38
rs995244494 Chr.2: 165309096-165309096 on Build GRCh38
rs369779411 Chr.2: 165309301-165309301 on Build GRCh38
rs779972161 Chr.2: 165309433-165309433 on Build GRCh38
rs748046296 Chr.2: 165309453-165309453 on Build GRCh38
rs746904068 Chr.2: 165309246-165309246 on Build GRCh38
rs947245904 Chr.2: 165309306-165309306 on Build GRCh38
rs761097228 Chr.2: 165309485-165309485 on Build GRCh38
rs769322683 Chr.2: 165309110-165309110 on Build GRCh38
rs753398618 Chr.2: 165309140-165309140 on Build GRCh38
rs112639117 Chr.2: 165309483-165309483 on Build GRCh38
rs1004482447 Chr.2: 165309482-165309482 on Build GRCh38
rs942312047 Chr.2: 165309387-165309387 on Build GRCh38
rs753150536 Chr.2: 165309126-165309126 on Build GRCh38

View Full Product Details