Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18878167-18878766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERS1
Entrez Gene ID
10715
Gene Name
ceramide synthase 1
Gene Aliases
EPM8, LAG1, LASS1, UOG1
UniGene
Hs.412355
Gene Chromosome Location
Chr.19: 18868546-18896727 on Build GRCh38
Transcripts
NM_001290265.1 NM_021267.4 NM_198207.2 NM_001492.5

Gene Symbol
GDF1
Entrez Gene ID
2657
Gene Name
growth differentiation factor 1
Gene Aliases
DORV, DTGA3, RAI
UniGene
Hs.412355
Gene Chromosome Location
Chr.19: 18868546-18896144 on Build GRCh38
Transcripts
NM_001290265.1 NM_021267.4 NM_198207.2 NM_001492.5

SNP Detail

SNP ID Chromosome Location
rs1030287490 Chr.19: 18878279-18878279 on Build GRCh38
rs564002546 Chr.19: 18878333-18878333 on Build GRCh38
rs559657143 Chr.19: 18878324-18878324 on Build GRCh38
rs890670309 Chr.19: 18878325-18878325 on Build GRCh38
rs1011586623 Chr.19: 18878379-18878379 on Build GRCh38
rs947769869 Chr.19: 18878197-18878197 on Build GRCh38
rs551798078 Chr.19: 18878482-18878482 on Build GRCh38
rs4808164 Chr.19: 18878646-18878646 on Build GRCh38
rs891161198 Chr.19: 18878295-18878295 on Build GRCh38
rs1009159170 Chr.19: 18878339-18878339 on Build GRCh38
rs1055584786 Chr.19: 18878223-18878223 on Build GRCh38
rs893113774 Chr.19: 18878565-18878565 on Build GRCh38
rs995048844 Chr.19: 18878641-18878641 on Build GRCh38
rs188033775 Chr.19: 18878442-18878442 on Build GRCh38
rs185468299 Chr.19: 18878746-18878746 on Build GRCh38
rs193060861 Chr.19: 18878457-18878457 on Build GRCh38
rs946112134 Chr.19: 18878272-18878272 on Build GRCh38
rs540900923 Chr.19: 18878392-18878392 on Build GRCh38
rs994977151 Chr.19: 18878430-18878430 on Build GRCh38
rs571678687 Chr.19: 18878491-18878491 on Build GRCh38
rs570006292 Chr.19: 18878711-18878711 on Build GRCh38
rs1034624873 Chr.19: 18878698-18878698 on Build GRCh38
rs958859804 Chr.19: 18878680-18878680 on Build GRCh38
rs1038534561 Chr.19: 18878233-18878233 on Build GRCh38
rs971815153 Chr.19: 18878733-18878733 on Build GRCh38
rs115623466 Chr.19: 18878479-18878479 on Build GRCh38
rs73526937 Chr.19: 18878681-18878681 on Build GRCh38
rs572083411 Chr.19: 18878299-18878299 on Build GRCh38
rs1052186354 Chr.19: 18878232-18878232 on Build GRCh38
rs1043171742 Chr.19: 18878275-18878275 on Build GRCh38
rs556414544 Chr.19: 18878220-18878220 on Build GRCh38
rs927919222 Chr.19: 18878225-18878225 on Build GRCh38
rs768519231 Chr.19: 18878483-18878483 on Build GRCh38
rs967142683 Chr.19: 18878401-18878401 on Build GRCh38
rs962221165 Chr.19: 18878417-18878417 on Build GRCh38
rs926101425 Chr.19: 18878514-18878514 on Build GRCh38
rs1039691748 Chr.19: 18878553-18878553 on Build GRCh38
rs1009128636 Chr.19: 18878580-18878580 on Build GRCh38
rs4808870 Chr.19: 18878511-18878511 on Build GRCh38
rs897300152 Chr.19: 18878234-18878234 on Build GRCh38
rs904511296 Chr.19: 18878276-18878276 on Build GRCh38
rs1043528535 Chr.19: 18878512-18878512 on Build GRCh38
rs4808869 Chr.19: 18878461-18878461 on Build GRCh38
rs928537506 Chr.19: 18878420-18878420 on Build GRCh38
rs887525491 Chr.19: 18878227-18878228 on Build GRCh38
rs960386458 Chr.19: 18878710-18878710 on Build GRCh38
rs913750924 Chr.19: 18878257-18878257 on Build GRCh38
rs897982662 Chr.19: 18878606-18878606 on Build GRCh38
rs960356267 Chr.19: 18878363-18878363 on Build GRCh38
rs1036636925 Chr.19: 18878602-18878602 on Build GRCh38
rs981081884 Chr.19: 18878222-18878222 on Build GRCh38
rs1020289709 Chr.19: 18878631-18878631 on Build GRCh38
rs1051390015 Chr.19: 18878499-18878499 on Build GRCh38
rs1035638212 Chr.19: 18878666-18878666 on Build GRCh38
rs183667458 Chr.19: 18878287-18878287 on Build GRCh38
rs561149924 Chr.19: 18878440-18878440 on Build GRCh38
rs1006919387 Chr.19: 18878522-18878522 on Build GRCh38
rs969125779 Chr.19: 18878659-18878659 on Build GRCh38
rs932063570 Chr.19: 18878558-18878558 on Build GRCh38
rs369729899 Chr.19: 18878576-18878576 on Build GRCh38
rs913720054 Chr.19: 18878507-18878507 on Build GRCh38
rs913857259 Chr.19: 18878715-18878715 on Build GRCh38
rs547540442 Chr.19: 18878563-18878563 on Build GRCh38
rs187001958 Chr.19: 18878230-18878230 on Build GRCh38
rs963000996 Chr.19: 18878699-18878699 on Build GRCh38
rs191513907 Chr.19: 18878246-18878246 on Build GRCh38

View Full Product Details