Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 127282053-127282573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF146
Entrez Gene ID
81847
Gene Name
ring finger protein 146
Gene Aliases
-
UniGene
Hs.267120
Gene Chromosome Location
Chr.6: 127266340-127289893 on Build GRCh38
Transcripts
NM_001242844.1 NM_001242845.1 NM_001242846.1 NM_001242847.1 NM_001242848.1 NM_001242849.1 NM_001242850.1 NM_001242851.1 NM_001242852.1 NM_030963.3 XM_006715571.3 XM_011536161.2 XM_011536162.2 XM_011536163.2 XM_011536164.2 XM_017011336.1 XM_017011337.1 XM_017011338.1 XM_017011339.1 XM_017011340.1 XM_017011341.1 XM_017011342.1 XM_017011343.1

SNP Detail

SNP ID Chromosome Location
rs543173743 Chr.6: 127282422-127282422 on Build GRCh38
rs937310813 Chr.6: 127282167-127282167 on Build GRCh38
rs747664326 Chr.6: 127282233-127282233 on Build GRCh38
rs1012658640 Chr.6: 127282314-127282314 on Build GRCh38
rs975063758 Chr.6: 127282186-127282186 on Build GRCh38
rs757001232 Chr.6: 127282149-127282149 on Build GRCh38
rs537978731 Chr.6: 127282134-127282134 on Build GRCh38
rs1043886455 Chr.6: 127282200-127282200 on Build GRCh38
rs995839716 Chr.6: 127282457-127282457 on Build GRCh38
rs986522496 Chr.6: 127282094-127282094 on Build GRCh38
rs796530091 Chr.6: 127282517-127282518 on Build GRCh38
rs963792537 Chr.6: 127282520-127282520 on Build GRCh38
rs898422995 Chr.6: 127282332-127282332 on Build GRCh38
rs922182325 Chr.6: 127282187-127282187 on Build GRCh38
rs975494172 Chr.6: 127282542-127282542 on Build GRCh38
rs896518361 Chr.6: 127282296-127282296 on Build GRCh38
rs746174935 Chr.6: 127282454-127282454 on Build GRCh38
rs944291300 Chr.6: 127282367-127282367 on Build GRCh38
rs187790740 Chr.6: 127282330-127282330 on Build GRCh38
rs981298374 Chr.6: 127282158-127282158 on Build GRCh38
rs928650187 Chr.6: 127282161-127282161 on Build GRCh38
rs991538390 Chr.6: 127282225-127282225 on Build GRCh38
rs1056939808 Chr.6: 127282295-127282295 on Build GRCh38
rs1015283247 Chr.6: 127282120-127282120 on Build GRCh38
rs778827268 Chr.6: 127282324-127282324 on Build GRCh38
rs1008008974 Chr.6: 127282438-127282438 on Build GRCh38
rs991443342 Chr.6: 127282545-127282545 on Build GRCh38
rs545888086 Chr.6: 127282472-127282472 on Build GRCh38
rs538994556 Chr.6: 127282323-127282323 on Build GRCh38
rs946938439 Chr.6: 127282193-127282193 on Build GRCh38
rs770155008 Chr.6: 127282471-127282471 on Build GRCh38
rs757773519 Chr.6: 127282353-127282353 on Build GRCh38
rs753549334 Chr.6: 127282093-127282093 on Build GRCh38
rs964043290 Chr.6: 127282122-127282122 on Build GRCh38
rs555524113 Chr.6: 127282502-127282502 on Build GRCh38
rs1025499260 Chr.6: 127282369-127282369 on Build GRCh38
rs754500394 Chr.6: 127282081-127282081 on Build GRCh38
rs1029192900 Chr.6: 127282463-127282463 on Build GRCh38
rs781611513 Chr.6: 127282415-127282415 on Build GRCh38
rs961823076 Chr.6: 127282493-127282495 on Build GRCh38
rs1041877737 Chr.6: 127282298-127282298 on Build GRCh38
rs887520353 Chr.6: 127282466-127282466 on Build GRCh38
rs575807718 Chr.6: 127282399-127282399 on Build GRCh38
rs761830618 Chr.6: 127282169-127282172 on Build GRCh38
rs557432332 Chr.6: 127282343-127282343 on Build GRCh38
rs935442975 Chr.6: 127282280-127282280 on Build GRCh38
rs958740592 Chr.6: 127282533-127282533 on Build GRCh38
rs1041289589 Chr.6: 127282414-127282414 on Build GRCh38
rs1035535147 Chr.6: 127282524-127282524 on Build GRCh38
rs912252328 Chr.6: 127282117-127282117 on Build GRCh38
rs961967695 Chr.6: 127282139-127282139 on Build GRCh38

View Full Product Details