Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 119796544-119797036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP5F
Entrez Gene ID
22876
Gene Name
inositol polyphosphate-5-phosphatase F
Gene Aliases
MSTP007, MSTPO47, SAC2, hSAC2
UniGene
Hs.369755
Gene Chromosome Location
Chr.10: 119704395-119829151 on Build GRCh38
Transcripts
NM_014937.3 XM_006717720.3 XM_011539524.2 XM_011539525.2 XM_011539527.2 XM_011539528.2 XM_017015935.1 XM_017015936.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4919127 c.847C>T p.R283C Chr.10: 119796892-119796892 on Build GRCh38
915395 c.817C>T p.H273Y Chr.10: 119796862-119796862 on Build GRCh38
1321505 c.705C>G p.I235M Chr.10: 119796750-119796750 on Build GRCh38
6460925 c.773G>T p.S258I Chr.10: 119796818-119796818 on Build GRCh38
6434960 c.684C>A p.D228E Chr.10: 119796729-119796729 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772877605 Chr.10: 119796950-119796950 on Build GRCh38
rs374135735 Chr.10: 119796702-119796702 on Build GRCh38
rs889388292 Chr.10: 119796868-119796868 on Build GRCh38
rs34239637 Chr.10: 119796747-119796747 on Build GRCh38
rs766806991 Chr.10: 119796876-119796876 on Build GRCh38
rs370751233 Chr.10: 119796992-119796992 on Build GRCh38
rs750419826 Chr.10: 119796826-119796827 on Build GRCh38
rs746923029 Chr.10: 119796721-119796721 on Build GRCh38
rs750294807 Chr.10: 119796907-119796907 on Build GRCh38
rs1036550520 Chr.10: 119796569-119796569 on Build GRCh38
rs749003085 Chr.10: 119796925-119796925 on Build GRCh38
rs373887437 Chr.10: 119796928-119796928 on Build GRCh38
rs202163857 Chr.10: 119796954-119796954 on Build GRCh38
rs149889046 Chr.10: 119796904-119796904 on Build GRCh38
rs753033639 Chr.10: 119796892-119796892 on Build GRCh38
rs748475671 Chr.10: 119796856-119796856 on Build GRCh38
rs968460222 Chr.10: 119796646-119796646 on Build GRCh38
rs770799425 Chr.10: 119796723-119796723 on Build GRCh38
rs755024875 Chr.10: 119796696-119796696 on Build GRCh38
rs745636185 Chr.10: 119796739-119796739 on Build GRCh38
rs796460433 Chr.10: 119796748-119796748 on Build GRCh38
rs747466506 Chr.10: 119796953-119796953 on Build GRCh38
rs565201983 Chr.10: 119796829-119796829 on Build GRCh38
rs766859366 Chr.10: 119796793-119796793 on Build GRCh38
rs769421563 Chr.10: 119796896-119796896 on Build GRCh38
rs898040096 Chr.10: 119796571-119796571 on Build GRCh38
rs199989664 Chr.10: 119796796-119796796 on Build GRCh38
rs35384980 Chr.10: 119796938-119796938 on Build GRCh38
rs761649756 Chr.10: 119796703-119796703 on Build GRCh38
rs370097956 Chr.10: 119796695-119796695 on Build GRCh38
rs778999179 Chr.10: 119796800-119796800 on Build GRCh38
rs774091071 Chr.10: 119796840-119796840 on Build GRCh38
rs182068908 Chr.10: 119796576-119796576 on Build GRCh38
rs754172929 Chr.10: 119796881-119796881 on Build GRCh38
rs1034490269 Chr.10: 119796587-119796587 on Build GRCh38
rs180849016 Chr.10: 119796766-119796766 on Build GRCh38
rs942095523 Chr.10: 119796782-119796782 on Build GRCh38
rs370105314 Chr.10: 119796955-119796955 on Build GRCh38
rs781106571 Chr.10: 119796815-119796815 on Build GRCh38
rs768169121 Chr.10: 119796834-119796834 on Build GRCh38
rs765436662 Chr.10: 119796895-119796895 on Build GRCh38
rs759818620 Chr.10: 119796964-119796964 on Build GRCh38
rs780420022 Chr.10: 119796694-119796694 on Build GRCh38
rs765348873 Chr.10: 119796889-119796889 on Build GRCh38
rs761107054 Chr.10: 119796792-119796792 on Build GRCh38
rs775111243 Chr.10: 119796760-119796760 on Build GRCh38
rs766816219 Chr.10: 119796697-119796697 on Build GRCh38
rs925038062 Chr.10: 119796745-119796745 on Build GRCh38
rs758493779 Chr.10: 119796966-119796966 on Build GRCh38
rs1007256137 Chr.10: 119796640-119796640 on Build GRCh38
rs915852391 Chr.10: 119796724-119796724 on Build GRCh38
rs759996227 Chr.10: 119796884-119796884 on Build GRCh38
rs992700708 Chr.10: 119796709-119796709 on Build GRCh38
rs771765473 Chr.10: 119796846-119796846 on Build GRCh38
rs998555624 Chr.10: 119796996-119796996 on Build GRCh38
rs993079046 Chr.10: 119796606-119796606 on Build GRCh38
rs186266329 Chr.10: 119796650-119796650 on Build GRCh38
rs778859851 Chr.10: 119796701-119796701 on Build GRCh38
rs977694514 Chr.10: 119796743-119796743 on Build GRCh38
rs756077795 Chr.10: 119796921-119796921 on Build GRCh38
rs779777503 Chr.10: 119796924-119796924 on Build GRCh38
rs761433014 Chr.10: 119796773-119796773 on Build GRCh38
rs768104728 Chr.10: 119796778-119796778 on Build GRCh38
rs761223167 Chr.10: 119796842-119796842 on Build GRCh38
rs73355843 Chr.10: 119796604-119796604 on Build GRCh38
rs758399126 Chr.10: 119796903-119796903 on Build GRCh38
rs758548836 Chr.10: 119796813-119796813 on Build GRCh38
rs377678035 Chr.10: 119796832-119796832 on Build GRCh38
rs752699934 Chr.10: 119796808-119796808 on Build GRCh38
rs781159229 Chr.10: 119796732-119796732 on Build GRCh38
rs778416088 Chr.10: 119796934-119796934 on Build GRCh38
rs771638410 Chr.10: 119796942-119796942 on Build GRCh38
rs756585793 Chr.10: 119796690-119796690 on Build GRCh38
rs1014955838 Chr.10: 119796948-119796948 on Build GRCh38
rs370583284 Chr.10: 119796869-119796869 on Build GRCh38
rs748932452 Chr.10: 119796833-119796833 on Build GRCh38
rs953144669 Chr.10: 119796635-119796635 on Build GRCh38
rs767374469 Chr.10: 119796775-119796776 on Build GRCh38
rs760620057 Chr.10: 119796858-119796859 on Build GRCh38
rs533796075 Chr.10: 119796589-119796589 on Build GRCh38
rs768147780 Chr.10: 119796768-119796768 on Build GRCh38
rs143032805 Chr.10: 119796643-119796643 on Build GRCh38
rs533134849 Chr.10: 119796893-119796893 on Build GRCh38
rs754240615 Chr.10: 119796744-119796744 on Build GRCh38
rs751037494 Chr.10: 119796780-119796780 on Build GRCh38

View Full Product Details