Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 159319478-159320042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAZ2B
Entrez Gene ID
29994
Gene Name
bromodomain adjacent to zinc finger domain 2B
Gene Aliases
WALp4
UniGene
Hs.470369
Gene Chromosome Location
Chr.2: 159315312-159712439 on Build GRCh38
Transcripts
NM_001289975.1 NM_013450.3 XM_005246488.2 XM_005246489.4 XM_005246492.4 XM_006712461.3 XM_011511038.2 XM_011511039.1 XM_011511042.1 XM_011511043.1 XM_011511044.1 XM_011511045.1 XM_011511047.1 XM_011511048.2 XM_011511050.1 XM_011511051.1 XM_011511052.1 XM_011511056.1 XM_017003918.1 XM_017003919.1 XM_017003920.1 XM_017003921.1 XM_017003922.1 XM_017003923.1 XM_017003924.1 XM_017003925.1 XM_017003926.1 XM_017003927.1 XM_017003928.1 XM_017003929.1 XM_017003930.1 XM_017003931.1 XM_017003932.1 XM_017003933.1 XM_017003934.1 XM_017003935.1 XM_017003936.1 XM_017003937.1

SNP Detail

SNP ID Chromosome Location
rs904748690 Chr.2: 159319924-159319924 on Build GRCh38
rs552579249 Chr.2: 159319577-159319577 on Build GRCh38
rs536113536 Chr.2: 159319900-159319900 on Build GRCh38
rs899741592 Chr.2: 159319965-159319965 on Build GRCh38
rs530127900 Chr.2: 159319746-159319746 on Build GRCh38
rs1010388036 Chr.2: 159319797-159319797 on Build GRCh38
rs748998531 Chr.2: 159319887-159319887 on Build GRCh38
rs1039740205 Chr.2: 159319710-159319710 on Build GRCh38
rs941308861 Chr.2: 159319679-159319679 on Build GRCh38
rs878934995 Chr.2: 159319916-159319916 on Build GRCh38
rs992946967 Chr.2: 159319593-159319593 on Build GRCh38
rs1034755501 Chr.2: 159319971-159319971 on Build GRCh38
rs920027207 Chr.2: 159319765-159319765 on Build GRCh38
rs759272525 Chr.2: 159319985-159319985 on Build GRCh38
rs931357765 Chr.2: 159319786-159319786 on Build GRCh38
rs753600430 Chr.2: 159319766-159319766 on Build GRCh38
rs768118619 Chr.2: 159319636-159319636 on Build GRCh38
rs982684670 Chr.2: 159319669-159319669 on Build GRCh38
rs979141873 Chr.2: 159319536-159319536 on Build GRCh38
rs929819786 Chr.2: 159319612-159319612 on Build GRCh38
rs890603386 Chr.2: 159319776-159319776 on Build GRCh38
rs979580938 Chr.2: 159319508-159319508 on Build GRCh38
rs917085950 Chr.2: 159319604-159319604 on Build GRCh38
rs1000455662 Chr.2: 159319934-159319934 on Build GRCh38
rs567988013 Chr.2: 159319650-159319650 on Build GRCh38
rs933732342 Chr.2: 159319737-159319737 on Build GRCh38
rs899503068 Chr.2: 159319714-159319714 on Build GRCh38
rs886979881 Chr.2: 159319894-159319894 on Build GRCh38
rs924069420 Chr.2: 159319513-159319513 on Build GRCh38
rs992860784 Chr.2: 159319553-159319554 on Build GRCh38
rs1041388530 Chr.2: 159319948-159319948 on Build GRCh38
rs909762983 Chr.2: 159319675-159319675 on Build GRCh38
rs948067623 Chr.2: 159319667-159319671 on Build GRCh38
rs544868064 Chr.2: 159319963-159319963 on Build GRCh38
rs1041902087 Chr.2: 159319804-159319804 on Build GRCh38
rs958744831 Chr.2: 159319590-159319590 on Build GRCh38
rs956981186 Chr.2: 159319533-159319536 on Build GRCh38
rs968733873 Chr.2: 159319531-159319531 on Build GRCh38
rs1034372240 Chr.2: 159319589-159319589 on Build GRCh38

View Full Product Details