Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101398269-101398767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLA
Entrez Gene ID
2717
Gene Name
galactosidase alpha
Gene Aliases
GALA
UniGene
Hs.69089
Gene Chromosome Location
Chr.X: 101397791-101408013 on Build GRCh38
Transcripts
NM_000169.2 NM_001199973.1 NM_001199974.1

Gene Symbol
RPL36A-HNRNPH2
Entrez Gene ID
100529097
Gene Name
RPL36A-HNRNPH2 readthrough
Gene Aliases
-
UniGene
Hs.432485
Gene Chromosome Location
Chr.X: 101390890-101414140 on Build GRCh38
Transcripts
NM_000169.2 NM_001199973.1 NM_001199974.1

SNP Detail

SNP ID Chromosome Location
rs869312424 Chr.X: 101398636-101398636 on Build GRCh38
rs869312459 Chr.X: 101398330-101398330 on Build GRCh38
rs869312439 Chr.X: 101398498-101398498 on Build GRCh38
rs869312430 Chr.X: 101398551-101398551 on Build GRCh38
rs869312442 Chr.X: 101398482-101398482 on Build GRCh38
rs104894839 Chr.X: 101398508-101398508 on Build GRCh38
rs111812846 Chr.X: 101398495-101398495 on Build GRCh38
rs869312420 Chr.X: 101398668-101398668 on Build GRCh38
rs797044499 Chr.X: 101398569-101398569 on Build GRCh38
rs104894846 Chr.X: 101398481-101398481 on Build GRCh38
rs869312415 Chr.X: 101398713-101398713 on Build GRCh38
rs869312157 Chr.X: 101398422-101398422 on Build GRCh38
rs869312427 Chr.X: 101398564-101398564 on Build GRCh38
rs782811400 Chr.X: 101398361-101398361 on Build GRCh38
rs797044769 Chr.X: 101398489-101398490 on Build GRCh38
rs28935495 Chr.X: 101398554-101398554 on Build GRCh38
rs869312454 Chr.X: 101398410-101398410 on Build GRCh38
rs782529392 Chr.X: 101398573-101398573 on Build GRCh38
rs28935492 Chr.X: 101398386-101398386 on Build GRCh38
rs398123228 Chr.X: 101398395-101398395 on Build GRCh38
rs869312444 Chr.X: 101398475-101398475 on Build GRCh38
rs869312422 Chr.X: 101398596-101398597 on Build GRCh38
rs869312447 Chr.X: 101398449-101398449 on Build GRCh38
rs782250167 Chr.X: 101398454-101398454 on Build GRCh38
rs886044860 Chr.X: 101398368-101398368 on Build GRCh38
rs28935491 Chr.X: 101398390-101398390 on Build GRCh38
rs782122397 Chr.X: 101398567-101398568 on Build GRCh38
rs375538532 Chr.X: 101398501-101398501 on Build GRCh38
rs797044768 Chr.X: 101398453-101398453 on Build GRCh38
rs140329381 Chr.X: 101398504-101398504 on Build GRCh38
rs869312446 Chr.X: 101398455-101398455 on Build GRCh38
rs869312417 Chr.X: 101398711-101398711 on Build GRCh38
rs869312453 Chr.X: 101398420-101398421 on Build GRCh38
rs782196174 Chr.X: 101398538-101398538 on Build GRCh38
rs142240841 Chr.X: 101398342-101398342 on Build GRCh38
rs869312437 Chr.X: 101398506-101398506 on Build GRCh38
rs151208856 Chr.X: 101398424-101398424 on Build GRCh38
rs869312429 Chr.X: 101398557-101398557 on Build GRCh38
rs869312432 Chr.X: 101398543-101398543 on Build GRCh38
rs727504372 Chr.X: 101398540-101398540 on Build GRCh38
rs730880452 Chr.X: 101398522-101398522 on Build GRCh38
rs869312451 Chr.X: 101398403-101398423 on Build GRCh38
rs398123225 Chr.X: 101398407-101398410 on Build GRCh38
rs104894828 Chr.X: 101398467-101398467 on Build GRCh38
rs869312419 Chr.X: 101398701-101398701 on Build GRCh38
rs782649832 Chr.X: 101398439-101398439 on Build GRCh38
rs869312414 Chr.X: 101398721-101398721 on Build GRCh38
rs869312160 Chr.X: 101398389-101398389 on Build GRCh38
rs398123223 Chr.X: 101398470-101398470 on Build GRCh38
rs869312456 Chr.X: 101398398-101398398 on Build GRCh38
rs398123224 Chr.X: 101398468-101398468 on Build GRCh38
rs869312426 Chr.X: 101398563-101398566 on Build GRCh38
rs104894830 Chr.X: 101398483-101398483 on Build GRCh38
rs922821899 Chr.X: 101398714-101398714 on Build GRCh38
rs869312435 Chr.X: 101398520-101398531 on Build GRCh38
rs398123227 Chr.X: 101398396-101398396 on Build GRCh38
rs782383870 Chr.X: 101398600-101398600 on Build GRCh38
rs398123229 Chr.X: 101398370-101398373 on Build GRCh38
rs869312159 Chr.X: 101398401-101398401 on Build GRCh38
rs869312434 Chr.X: 101398531-101398531 on Build GRCh38
rs398123226 Chr.X: 101398403-101398403 on Build GRCh38
rs28935489 Chr.X: 101398479-101398479 on Build GRCh38
rs869312421 Chr.X: 101398591-101398591 on Build GRCh38
rs926075804 Chr.X: 101398599-101398599 on Build GRCh38
rs869312452 Chr.X: 101398421-101398422 on Build GRCh38
rs1001403373 Chr.X: 101398406-101398406 on Build GRCh38
rs869312409 Chr.X: 101398683-101398683 on Build GRCh38
rs142217511 Chr.X: 101398423-101398423 on Build GRCh38
rs782126889 Chr.X: 101398374-101398374 on Build GRCh38
rs376623208 Chr.X: 101398385-101398385 on Build GRCh38
rs964170797 Chr.X: 101398679-101398679 on Build GRCh38
rs782096403 Chr.X: 101398735-101398735 on Build GRCh38
rs869312445 Chr.X: 101398462-101398462 on Build GRCh38
rs730880437 Chr.X: 101398378-101398378 on Build GRCh38
rs782032720 Chr.X: 101398535-101398535 on Build GRCh38
rs869312425 Chr.X: 101398663-101398663 on Build GRCh38
rs869312450 Chr.X: 101398423-101398423 on Build GRCh38
rs869312411 Chr.X: 101398743-101398743 on Build GRCh38
rs869312423 Chr.X: 101398651-101398651 on Build GRCh38
rs869312441 Chr.X: 101398491-101398491 on Build GRCh38
rs869312416 Chr.X: 101398719-101398719 on Build GRCh38
rs886044766 Chr.X: 101398539-101398539 on Build GRCh38
rs797044498 Chr.X: 101398569-101398570 on Build GRCh38
rs869312156 Chr.X: 101398426-101398426 on Build GRCh38
rs727504773 Chr.X: 101398565-101398570 on Build GRCh38
rs570042882 Chr.X: 101398433-101398433 on Build GRCh38
rs781884596 Chr.X: 101398370-101398370 on Build GRCh38
rs730880439 Chr.X: 101398408-101398408 on Build GRCh38
rs782056296 Chr.X: 101398616-101398617 on Build GRCh38
rs1057518537 Chr.X: 101398374-101398375 on Build GRCh38
rs104894832 Chr.X: 101398387-101398387 on Build GRCh38
rs869312431 Chr.X: 101398545-101398545 on Build GRCh38
rs869312458 Chr.X: 101398375-101398375 on Build GRCh38
rs782238586 Chr.X: 101398634-101398634 on Build GRCh38
rs782298737 Chr.X: 101398494-101398494 on Build GRCh38
rs869312443 Chr.X: 101398477-101398478 on Build GRCh38
rs869312457 Chr.X: 101398331-101398331 on Build GRCh38
rs28935490 Chr.X: 101398432-101398432 on Build GRCh38
rs869312158 Chr.X: 101398419-101398419 on Build GRCh38
rs869312433 Chr.X: 101398533-101398533 on Build GRCh38
rs782757545 Chr.X: 101398375-101398375 on Build GRCh38
rs398123222 Chr.X: 101398546-101398546 on Build GRCh38
rs869312161 Chr.X: 101398380-101398380 on Build GRCh38
rs782575087 Chr.X: 101398478-101398478 on Build GRCh38
rs869025435 Chr.X: 101398546-101398546 on Build GRCh38
rs28935485 Chr.X: 101398534-101398534 on Build GRCh38
rs869312436 Chr.X: 101398507-101398507 on Build GRCh38
rs869312428 Chr.X: 101398511-101398511 on Build GRCh38
rs147737890 Chr.X: 101398402-101398402 on Build GRCh38
rs188036247 Chr.X: 101398609-101398609 on Build GRCh38
rs28935488 Chr.X: 101398563-101398563 on Build GRCh38
rs781931935 Chr.X: 101398326-101398326 on Build GRCh38
rs369055986 Chr.X: 101398425-101398425 on Build GRCh38
rs151195362 Chr.X: 101398391-101398391 on Build GRCh38
rs869312449 Chr.X: 101398438-101398438 on Build GRCh38
rs869312455 Chr.X: 101398405-101398406 on Build GRCh38
rs869312155 Chr.X: 101398443-101398443 on Build GRCh38
rs869312413 Chr.X: 101398725-101398725 on Build GRCh38
rs869312418 Chr.X: 101398710-101398710 on Build GRCh38
rs869312460 Chr.X: 101398302-101398302 on Build GRCh38
rs869312440 Chr.X: 101398492-101398492 on Build GRCh38

View Full Product Details