Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75087348-75087918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL12A1
Entrez Gene ID
1303
Gene Name
collagen type XII alpha 1 chain
Gene Aliases
BA209D8.1, BTHLM2, COL12A1L, DJ234P15.1, UCMD2
UniGene
Hs.101302
Gene Chromosome Location
Chr.6: 75084326-75206262 on Build GRCh38
Transcripts
NM_004370.5 NM_080645.2 XM_011535434.1 XM_011535435.1 XM_011535436.2 XM_017010252.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1081633 c.9022G>A p.E3008K Chr.6: 75087736-75087736 on Build GRCh38
5431011 c.9121C>T p.P3041S Chr.6: 75087637-75087637 on Build GRCh38
3875466 c.9153C>A p.S3051R Chr.6: 75087605-75087605 on Build GRCh38
1081631 c.9103C>T p.R3035* Chr.6: 75087655-75087655 on Build GRCh38
122265 c.9064C>A p.P3022T Chr.6: 75087694-75087694 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560180665 Chr.6: 75087685-75087685 on Build GRCh38
rs74616265 Chr.6: 75087416-75087416 on Build GRCh38
rs763341868 Chr.6: 75087557-75087557 on Build GRCh38
rs201412621 Chr.6: 75087649-75087649 on Build GRCh38
rs879090705 Chr.6: 75087472-75087472 on Build GRCh38
rs768155096 Chr.6: 75087532-75087532 on Build GRCh38
rs747009147 Chr.6: 75087526-75087526 on Build GRCh38
rs778727603 Chr.6: 75087771-75087771 on Build GRCh38
rs116059284 Chr.6: 75087524-75087524 on Build GRCh38
rs547597960 Chr.6: 75087845-75087845 on Build GRCh38
rs35292916 Chr.6: 75087596-75087596 on Build GRCh38
rs988363717 Chr.6: 75087802-75087802 on Build GRCh38
rs777425494 Chr.6: 75087650-75087650 on Build GRCh38
rs746192829 Chr.6: 75087777-75087777 on Build GRCh38
rs747656891 Chr.6: 75087634-75087634 on Build GRCh38
rs781013677 Chr.6: 75087633-75087633 on Build GRCh38
rs573530946 Chr.6: 75087658-75087658 on Build GRCh38
rs867032927 Chr.6: 75087559-75087559 on Build GRCh38
rs146873644 Chr.6: 75087876-75087876 on Build GRCh38
rs565801419 Chr.6: 75087523-75087523 on Build GRCh38
rs754761310 Chr.6: 75087719-75087719 on Build GRCh38
rs370683755 Chr.6: 75087755-75087755 on Build GRCh38
rs752591741 Chr.6: 75087752-75087752 on Build GRCh38
rs765185429 Chr.6: 75087426-75087426 on Build GRCh38
rs1002433115 Chr.6: 75087589-75087589 on Build GRCh38
rs926692503 Chr.6: 75087843-75087843 on Build GRCh38
rs376161354 Chr.6: 75087720-75087720 on Build GRCh38
rs369315573 Chr.6: 75087543-75087543 on Build GRCh38
rs2273264 Chr.6: 75087833-75087833 on Build GRCh38
rs752857440 Chr.6: 75087513-75087513 on Build GRCh38
rs374037516 Chr.6: 75087710-75087710 on Build GRCh38
rs376189614 Chr.6: 75087682-75087682 on Build GRCh38
rs774985994 Chr.6: 75087573-75087573 on Build GRCh38
rs563604000 Chr.6: 75087427-75087427 on Build GRCh38
rs776054700 Chr.6: 75087794-75087794 on Build GRCh38
rs1004806251 Chr.6: 75087449-75087449 on Build GRCh38
rs779895718 Chr.6: 75087615-75087615 on Build GRCh38
rs200178350 Chr.6: 75087711-75087711 on Build GRCh38
rs751471471 Chr.6: 75087624-75087624 on Build GRCh38
rs564552134 Chr.6: 75087769-75087769 on Build GRCh38
rs41266761 Chr.6: 75087675-75087675 on Build GRCh38
rs369646700 Chr.6: 75087593-75087593 on Build GRCh38
rs547541301 Chr.6: 75087522-75087522 on Build GRCh38
rs756395971 Chr.6: 75087514-75087514 on Build GRCh38
rs769439797 Chr.6: 75087544-75087544 on Build GRCh38
rs777215475 Chr.6: 75087768-75087768 on Build GRCh38
rs374384579 Chr.6: 75087770-75087770 on Build GRCh38
rs57396313 Chr.6: 75087614-75087614 on Build GRCh38
rs572512247 Chr.6: 75087686-75087686 on Build GRCh38
rs550350661 Chr.6: 75087400-75087400 on Build GRCh38
rs751306147 Chr.6: 75087717-75087717 on Build GRCh38
rs978214339 Chr.6: 75087864-75087864 on Build GRCh38
rs767461073 Chr.6: 75087587-75087587 on Build GRCh38
rs749005420 Chr.6: 75087654-75087654 on Build GRCh38
rs370090007 Chr.6: 75087648-75087648 on Build GRCh38
rs140011165 Chr.6: 75087433-75087433 on Build GRCh38
rs878856640 Chr.6: 75087799-75087799 on Build GRCh38
rs771434132 Chr.6: 75087569-75087569 on Build GRCh38
rs772326311 Chr.6: 75087779-75087779 on Build GRCh38
rs923406031 Chr.6: 75087375-75087375 on Build GRCh38
rs1046409074 Chr.6: 75087840-75087840 on Build GRCh38
rs936882149 Chr.6: 75087785-75087785 on Build GRCh38
rs970547 Chr.6: 75087586-75087586 on Build GRCh38
rs752866217 Chr.6: 75087588-75087588 on Build GRCh38
rs267601120 Chr.6: 75087621-75087621 on Build GRCh38
rs769193105 Chr.6: 75087644-75087644 on Build GRCh38
rs754958196 Chr.6: 75087630-75087630 on Build GRCh38
rs760094440 Chr.6: 75087677-75087677 on Build GRCh38
rs887628569 Chr.6: 75087695-75087695 on Build GRCh38
rs200901687 Chr.6: 75087694-75087694 on Build GRCh38
rs913880092 Chr.6: 75087829-75087829 on Build GRCh38
rs767762945 Chr.6: 75087508-75087508 on Build GRCh38
rs751344618 Chr.6: 75087592-75087592 on Build GRCh38
rs758519964 Chr.6: 75087572-75087572 on Build GRCh38
rs371725823 Chr.6: 75087550-75087550 on Build GRCh38

View Full Product Details