Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 8751486-8752057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NXPH1
Entrez Gene ID
30010
Gene Name
neurexophilin 1
Gene Aliases
NPH1, Nbla00697
UniGene
Hs.487564
Gene Chromosome Location
Chr.7: 8433955-8752963 on Build GRCh38
Transcripts
NM_152745.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1092726 c.535G>A p.A179T Chr.7: 8751488-8751488 on Build GRCh38
3412454 c.535G>C p.A179P Chr.7: 8751488-8751488 on Build GRCh38
1092734 c.695C>T p.S232F Chr.7: 8751648-8751648 on Build GRCh38
3412456 c.772G>A p.D258N Chr.7: 8751725-8751725 on Build GRCh38
3033240 c.810G>C p.S270S Chr.7: 8751763-8751763 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774123742 Chr.7: 8751521-8751521 on Build GRCh38
rs371180162 Chr.7: 8751537-8751537 on Build GRCh38
rs768909829 Chr.7: 8751649-8751649 on Build GRCh38
rs749493668 Chr.7: 8751648-8751648 on Build GRCh38
rs771372049 Chr.7: 8751789-8751789 on Build GRCh38
rs893379237 Chr.7: 8751872-8751872 on Build GRCh38
rs762280995 Chr.7: 8751507-8751507 on Build GRCh38
rs369091443 Chr.7: 8751784-8751784 on Build GRCh38
rs932773129 Chr.7: 8751782-8751782 on Build GRCh38
rs189108272 Chr.7: 8751873-8751873 on Build GRCh38
rs267601614 Chr.7: 8751613-8751613 on Build GRCh38
rs779110302 Chr.7: 8751766-8751766 on Build GRCh38
rs772389139 Chr.7: 8751663-8751663 on Build GRCh38
rs908416911 Chr.7: 8751977-8751977 on Build GRCh38
rs759311254 Chr.7: 8751561-8751561 on Build GRCh38
rs1011748147 Chr.7: 8751883-8751883 on Build GRCh38
rs969165436 Chr.7: 8751573-8751573 on Build GRCh38
rs757697842 Chr.7: 8752035-8752035 on Build GRCh38
rs969111698 Chr.7: 8751907-8751907 on Build GRCh38
rs768088761 Chr.7: 8751514-8751514 on Build GRCh38
rs983177692 Chr.7: 8751972-8751972 on Build GRCh38
rs368010668 Chr.7: 8751676-8751676 on Build GRCh38
rs964126640 Chr.7: 8751830-8751830 on Build GRCh38
rs73244893 Chr.7: 8751842-8751842 on Build GRCh38
rs751247477 Chr.7: 8751637-8751637 on Build GRCh38
rs1022012930 Chr.7: 8751548-8751548 on Build GRCh38
rs750253607 Chr.7: 8751762-8751762 on Build GRCh38
rs375393297 Chr.7: 8751646-8751646 on Build GRCh38
rs777173431 Chr.7: 8751682-8751682 on Build GRCh38
rs908470270 Chr.7: 8751744-8751744 on Build GRCh38
rs752198705 Chr.7: 8751715-8751715 on Build GRCh38
rs1056181055 Chr.7: 8752024-8752024 on Build GRCh38
rs1023171077 Chr.7: 8751886-8751886 on Build GRCh38
rs556554313 Chr.7: 8751801-8751801 on Build GRCh38
rs183316469 Chr.7: 8751594-8751594 on Build GRCh38
rs778996609 Chr.7: 8752036-8752036 on Build GRCh38
rs757071991 Chr.7: 8751643-8751643 on Build GRCh38
rs769504277 Chr.7: 8751802-8751802 on Build GRCh38
rs765004578 Chr.7: 8751706-8751706 on Build GRCh38
rs777781873 Chr.7: 8751636-8751636 on Build GRCh38
rs570857247 Chr.7: 8751779-8751779 on Build GRCh38
rs982865547 Chr.7: 8751973-8751973 on Build GRCh38
rs140953357 Chr.7: 8752025-8752025 on Build GRCh38
rs773760900 Chr.7: 8751667-8751667 on Build GRCh38
rs930019126 Chr.7: 8751960-8751960 on Build GRCh38
rs753526886 Chr.7: 8751549-8751549 on Build GRCh38
rs758363505 Chr.7: 8751630-8751630 on Build GRCh38
rs560119091 Chr.7: 8751926-8751926 on Build GRCh38
rs77545591 Chr.7: 8751833-8751833 on Build GRCh38
rs761143146 Chr.7: 8751671-8751671 on Build GRCh38
rs759223293 Chr.7: 8751697-8751697 on Build GRCh38
rs922681514 Chr.7: 8751925-8751925 on Build GRCh38
rs764807888 Chr.7: 8751604-8751604 on Build GRCh38
rs764623572 Chr.7: 8751948-8751948 on Build GRCh38
rs763579524 Chr.7: 8751745-8751745 on Build GRCh38
rs1045239319 Chr.7: 8752003-8752003 on Build GRCh38
rs888680518 Chr.7: 8751603-8751603 on Build GRCh38
rs941566875 Chr.7: 8751716-8751716 on Build GRCh38
rs910149239 Chr.7: 8751981-8751981 on Build GRCh38
rs891104086 Chr.7: 8751834-8751834 on Build GRCh38
rs748432098 Chr.7: 8751771-8751771 on Build GRCh38
rs980423098 Chr.7: 8751621-8751621 on Build GRCh38
rs757195157 Chr.7: 8751757-8751757 on Build GRCh38
rs773388495 Chr.7: 8751515-8751515 on Build GRCh38
rs777763157 Chr.7: 8751780-8751780 on Build GRCh38
rs567005105 Chr.7: 8751655-8751655 on Build GRCh38
rs1047350505 Chr.7: 8751542-8751542 on Build GRCh38
rs1034654502 Chr.7: 8751747-8751747 on Build GRCh38
rs543758572 Chr.7: 8752034-8752034 on Build GRCh38
rs960375757 Chr.7: 8751964-8751964 on Build GRCh38
rs3779355 Chr.7: 8751988-8751988 on Build GRCh38
rs748237878 Chr.7: 8751661-8751661 on Build GRCh38
rs146456118 Chr.7: 8752016-8752016 on Build GRCh38
rs752284747 Chr.7: 8751616-8751616 on Build GRCh38
rs780936787 Chr.7: 8751758-8751758 on Build GRCh38
rs921126080 Chr.7: 8752018-8752018 on Build GRCh38
rs374966340 Chr.7: 8751763-8751763 on Build GRCh38
rs372390499 Chr.7: 8751805-8751805 on Build GRCh38
rs568384804 Chr.7: 8751841-8751841 on Build GRCh38
rs553984828 Chr.7: 8751866-8751866 on Build GRCh38
rs200983135 Chr.7: 8751718-8751718 on Build GRCh38
rs751504857 Chr.7: 8751754-8751754 on Build GRCh38
rs1001980405 Chr.7: 8751910-8751910 on Build GRCh38

View Full Product Details