Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 129716913-129717506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NRF1
Entrez Gene ID
4899
Gene Name
nuclear respiratory factor 1
Gene Aliases
ALPHA-PAL
UniGene
Hs.654363
Gene Chromosome Location
Chr.7: 129611702-129757082 on Build GRCh38
Transcripts
NM_001040110.1 NM_001293163.1 NM_001293164.1 NM_005011.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2152922 c.1147G>A p.A383T Chr.7: 129717300-129717300 on Build GRCh38
6108652 c.1185G>T p.E395D Chr.7: 129717338-129717338 on Build GRCh38
4359721 c.1143G>A p.A381A Chr.7: 129717296-129717296 on Build GRCh38
3411582 c.1152G>A p.S384S Chr.7: 129717305-129717305 on Build GRCh38
3411580 c.1147G>A p.A383T Chr.7: 129717300-129717300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941980890 Chr.7: 129717088-129717088 on Build GRCh38
rs78877132 Chr.7: 129717328-129717328 on Build GRCh38
rs1050440359 Chr.7: 129717072-129717072 on Build GRCh38
rs750986793 Chr.7: 129716969-129716969 on Build GRCh38
rs781533688 Chr.7: 129716960-129716960 on Build GRCh38
rs781567672 Chr.7: 129717427-129717427 on Build GRCh38
rs761276231 Chr.7: 129717296-129717296 on Build GRCh38
rs994078705 Chr.7: 129717102-129717102 on Build GRCh38
rs768152680 Chr.7: 129717286-129717286 on Build GRCh38
rs893523967 Chr.7: 129717472-129717472 on Build GRCh38
rs1029713560 Chr.7: 129717459-129717459 on Build GRCh38
rs531109667 Chr.7: 129717465-129717465 on Build GRCh38
rs1047031627 Chr.7: 129717428-129717428 on Build GRCh38
rs767037669 Chr.7: 129717305-129717305 on Build GRCh38
rs766286938 Chr.7: 129717317-129717317 on Build GRCh38
rs910542741 Chr.7: 129717078-129717078 on Build GRCh38
rs180884027 Chr.7: 129717455-129717455 on Build GRCh38
rs1054806538 Chr.7: 129717468-129717468 on Build GRCh38
rs757704626 Chr.7: 129717425-129717425 on Build GRCh38
rs779076542 Chr.7: 129717227-129717227 on Build GRCh38
rs758466637 Chr.7: 129717401-129717401 on Build GRCh38
rs111612142 Chr.7: 129716937-129716937 on Build GRCh38
rs933418502 Chr.7: 129717070-129717070 on Build GRCh38
rs373325490 Chr.7: 129717368-129717368 on Build GRCh38
rs979456427 Chr.7: 129717044-129717044 on Build GRCh38
rs773347057 Chr.7: 129717306-129717306 on Build GRCh38
rs1020836360 Chr.7: 129717229-129717229 on Build GRCh38
rs187399737 Chr.7: 129716971-129716971 on Build GRCh38
rs895569706 Chr.7: 129717239-129717239 on Build GRCh38
rs1033290751 Chr.7: 129716941-129716941 on Build GRCh38
rs755190001 Chr.7: 129717200-129717200 on Build GRCh38
rs774023509 Chr.7: 129717295-129717295 on Build GRCh38
rs190838970 Chr.7: 129717272-129717272 on Build GRCh38
rs866386749 Chr.7: 129717028-129717028 on Build GRCh38
rs2293629 Chr.7: 129717458-129717458 on Build GRCh38
rs942010655 Chr.7: 129717393-129717393 on Build GRCh38
rs777293839 Chr.7: 129717460-129717460 on Build GRCh38
rs766270472 Chr.7: 129717190-129717190 on Build GRCh38
rs767393668 Chr.7: 129717176-129717176 on Build GRCh38
rs919533262 Chr.7: 129717419-129717419 on Build GRCh38
rs575094240 Chr.7: 129716976-129716976 on Build GRCh38
rs954494655 Chr.7: 129717125-129717125 on Build GRCh38
rs75550852 Chr.7: 129717456-129717456 on Build GRCh38
rs71529414 Chr.7: 129717456-129717458 on Build GRCh38
rs754113727 Chr.7: 129717191-129717191 on Build GRCh38
rs1044561380 Chr.7: 129717390-129717390 on Build GRCh38
rs748827747 Chr.7: 129717406-129717406 on Build GRCh38
rs200050482 Chr.7: 129717171-129717171 on Build GRCh38
rs750384099 Chr.7: 129717188-129717188 on Build GRCh38
rs922697753 Chr.7: 129717121-129717121 on Build GRCh38
rs928987072 Chr.7: 129717234-129717234 on Build GRCh38
rs778195422 Chr.7: 129717254-129717254 on Build GRCh38
rs760618119 Chr.7: 129717315-129717315 on Build GRCh38
rs879842050 Chr.7: 129716951-129716951 on Build GRCh38
rs752945543 Chr.7: 129717394-129717394 on Build GRCh38
rs17161218 Chr.7: 129717096-129717096 on Build GRCh38
rs12154342 Chr.7: 129717099-129717099 on Build GRCh38
rs748140517 Chr.7: 129717238-129717238 on Build GRCh38
rs1023596968 Chr.7: 129717039-129717039 on Build GRCh38
rs910416932 Chr.7: 129717187-129717187 on Build GRCh38
rs765419489 Chr.7: 129717379-129717379 on Build GRCh38
rs758475449 Chr.7: 129717248-129717248 on Build GRCh38
rs771354813 Chr.7: 129717269-129717269 on Build GRCh38
rs377376736 Chr.7: 129717409-129717409 on Build GRCh38
rs923385254 Chr.7: 129717050-129717050 on Build GRCh38
rs756028484 Chr.7: 129717179-129717179 on Build GRCh38
rs970331623 Chr.7: 129717040-129717040 on Build GRCh38
rs574876934 Chr.7: 129717424-129717424 on Build GRCh38
rs541431392 Chr.7: 129717259-129717259 on Build GRCh38
rs1038045168 Chr.7: 129717208-129717208 on Build GRCh38
rs750350902 Chr.7: 129717177-129717177 on Build GRCh38
rs1033002940 Chr.7: 129717087-129717087 on Build GRCh38
rs541252387 Chr.7: 129717173-129717173 on Build GRCh38

View Full Product Details