Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 23546207-23546661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOX5
Entrez Gene ID
6660
Gene Name
SRY-box 5
Gene Aliases
L-SOX5, L-SOX5B, L-SOX5F, LAMSHF
UniGene
Hs.657542
Gene Chromosome Location
Chr.12: 23529499-24562701 on Build GRCh38
Transcripts
NM_001261414.1 NM_001261415.1 NM_006940.4 NM_152989.3 NM_178010.2 XM_011520831.2 XM_011520832.2 XM_011520833.2 XM_011520834.2 XM_011520835.2 XM_011520837.2 XM_011520838.2 XM_011520842.2 XM_017019888.1 XM_017019889.1 XM_017019890.1 XM_017019891.1 XM_017019892.1 XM_017019893.1 XM_017019894.1 XM_017019895.1 XM_017019896.1 XM_017019897.1 XM_017019898.1 XM_017019899.1 XM_017019900.1 XM_017019901.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6386403 c.404C>T p.A135V Chr.12: 23546351-23546351 on Build GRCh38
7293133 c.390A>G p.G130G Chr.12: 23546365-23546365 on Build GRCh38
5012508 c.1504G>T p.E502* Chr.12: 23546409-23546409 on Build GRCh38
7258726 c.331-5delT p.? Chr.12: 23546429-23546429 on Build GRCh38
5825733 c.335A>C p.K112T Chr.12: 23546420-23546420 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053569497 Chr.12: 23546602-23546602 on Build GRCh38
rs778013687 Chr.12: 23546353-23546353 on Build GRCh38
rs866277021 Chr.12: 23546598-23546598 on Build GRCh38
rs1034544485 Chr.12: 23546559-23546559 on Build GRCh38
rs755614685 Chr.12: 23546545-23546545 on Build GRCh38
rs766239567 Chr.12: 23546438-23546440 on Build GRCh38
rs774119544 Chr.12: 23546289-23546289 on Build GRCh38
rs547581821 Chr.12: 23546596-23546596 on Build GRCh38
rs35132403 Chr.12: 23546581-23546582 on Build GRCh38
rs758550572 Chr.12: 23546349-23546349 on Build GRCh38
rs534151269 Chr.12: 23546592-23546592 on Build GRCh38
rs770367334 Chr.12: 23546467-23546467 on Build GRCh38
rs1051792865 Chr.12: 23546454-23546454 on Build GRCh38
rs750162465 Chr.12: 23546254-23546254 on Build GRCh38
rs766355368 Chr.12: 23546308-23546308 on Build GRCh38
rs759396603 Chr.12: 23546336-23546336 on Build GRCh38
rs377289233 Chr.12: 23546294-23546294 on Build GRCh38
rs917228851 Chr.12: 23546509-23546509 on Build GRCh38
rs967666286 Chr.12: 23546606-23546606 on Build GRCh38
rs202222877 Chr.12: 23546419-23546419 on Build GRCh38
rs770252719 Chr.12: 23546287-23546287 on Build GRCh38
rs376491286 Chr.12: 23546608-23546608 on Build GRCh38
rs765000530 Chr.12: 23546341-23546341 on Build GRCh38
rs899380076 Chr.12: 23546547-23546547 on Build GRCh38
rs766004790 Chr.12: 23546563-23546563 on Build GRCh38
rs184667789 Chr.12: 23546623-23546623 on Build GRCh38
rs993250226 Chr.12: 23546489-23546489 on Build GRCh38
rs780287740 Chr.12: 23546471-23546471 on Build GRCh38
rs769873993 Chr.12: 23546428-23546429 on Build GRCh38
rs936137755 Chr.12: 23546577-23546577 on Build GRCh38
rs199957729 Chr.12: 23546418-23546418 on Build GRCh38
rs549078060 Chr.12: 23546485-23546485 on Build GRCh38
rs746372775 Chr.12: 23546459-23546459 on Build GRCh38
rs1024666604 Chr.12: 23546238-23546238 on Build GRCh38
rs200992905 Chr.12: 23546413-23546413 on Build GRCh38
rs528856011 Chr.12: 23546439-23546439 on Build GRCh38
rs992747324 Chr.12: 23546238-23546239 on Build GRCh38
rs760130581 Chr.12: 23546295-23546295 on Build GRCh38
rs767131610 Chr.12: 23546291-23546291 on Build GRCh38
rs977664096 Chr.12: 23546520-23546520 on Build GRCh38
rs940956648 Chr.12: 23546488-23546488 on Build GRCh38
rs140106320 Chr.12: 23546388-23546388 on Build GRCh38
rs757387985 Chr.12: 23546385-23546385 on Build GRCh38
rs565623858 Chr.12: 23546553-23546553 on Build GRCh38
rs751611995 Chr.12: 23546361-23546361 on Build GRCh38
rs1036451546 Chr.12: 23546504-23546504 on Build GRCh38
rs112285652 Chr.12: 23546320-23546320 on Build GRCh38
rs946289953 Chr.12: 23546516-23546516 on Build GRCh38
rs752475410 Chr.12: 23546345-23546345 on Build GRCh38
rs560846657 Chr.12: 23546460-23546460 on Build GRCh38

View Full Product Details