Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 113022642-113023144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL36B
Entrez Gene ID
27177
Gene Name
interleukin 36, beta
Gene Aliases
FIL1, FIL1-(ETA), FIL1H, FILI-(ETA), IL-1F8, IL-1H2, IL1-ETA, IL1F8, IL1H2
UniGene
Hs.278909
Gene Chromosome Location
Chr.2: 113022091-113052867 on Build GRCh38
Transcripts
NM_014438.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3565447 c.399G>A p.K133K Chr.2: 113022770-113022770 on Build GRCh38
3406800 c.404G>A p.W135* Chr.2: 113022765-113022765 on Build GRCh38
1690920 c.476G>A p.G159E Chr.2: 113022693-113022693 on Build GRCh38
5687712 c.435G>A p.R145R Chr.2: 113022734-113022734 on Build GRCh38
3565445 c.480G>A p.M160I Chr.2: 113022689-113022689 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893402936 Chr.2: 113022980-113022980 on Build GRCh38
rs867016988 Chr.2: 113022775-113022775 on Build GRCh38
rs1046963039 Chr.2: 113023040-113023040 on Build GRCh38
rs770269491 Chr.2: 113022754-113022754 on Build GRCh38
rs763458820 Chr.2: 113022778-113022778 on Build GRCh38
rs750344480 Chr.2: 113022816-113022816 on Build GRCh38
rs935110594 Chr.2: 113022947-113022947 on Build GRCh38
rs779061101 Chr.2: 113022714-113022714 on Build GRCh38
rs1029448381 Chr.2: 113023039-113023039 on Build GRCh38
rs772685813 Chr.2: 113022728-113022730 on Build GRCh38
rs923674867 Chr.2: 113022938-113022938 on Build GRCh38
rs976072941 Chr.2: 113022833-113022833 on Build GRCh38
rs1053425541 Chr.2: 113022974-113022975 on Build GRCh38
rs1001680166 Chr.2: 113022871-113022871 on Build GRCh38
rs918584789 Chr.2: 113023115-113023115 on Build GRCh38
rs781518301 Chr.2: 113022696-113022696 on Build GRCh38
rs147745025 Chr.2: 113022720-113022720 on Build GRCh38
rs529822898 Chr.2: 113022706-113022706 on Build GRCh38
rs768808533 Chr.2: 113022732-113022732 on Build GRCh38
rs745930242 Chr.2: 113022716-113022716 on Build GRCh38
rs779327578 Chr.2: 113022678-113022678 on Build GRCh38
rs763062737 Chr.2: 113022860-113022860 on Build GRCh38
rs140063120 Chr.2: 113023035-113023035 on Build GRCh38
rs745411769 Chr.2: 113022722-113022723 on Build GRCh38
rs770900472 Chr.2: 113023046-113023046 on Build GRCh38
rs374123046 Chr.2: 113022803-113022803 on Build GRCh38
rs911095330 Chr.2: 113023085-113023085 on Build GRCh38
rs532149478 Chr.2: 113022902-113022902 on Build GRCh38
rs371409243 Chr.2: 113022873-113022873 on Build GRCh38
rs918627482 Chr.2: 113022794-113022794 on Build GRCh38
rs142880845 Chr.2: 113022694-113022694 on Build GRCh38
rs200218645 Chr.2: 113022702-113022702 on Build GRCh38
rs955530307 Chr.2: 113022876-113022876 on Build GRCh38
rs766977479 Chr.2: 113022780-113022780 on Build GRCh38
rs752541934 Chr.2: 113022949-113022949 on Build GRCh38
rs887164645 Chr.2: 113023060-113023060 on Build GRCh38
rs752174564 Chr.2: 113022782-113022782 on Build GRCh38
rs1021966667 Chr.2: 113022991-113022991 on Build GRCh38
rs774405900 Chr.2: 113022983-113022983 on Build GRCh38
rs765770037 Chr.2: 113022670-113022670 on Build GRCh38
rs370421207 Chr.2: 113022666-113022666 on Build GRCh38
rs746077689 Chr.2: 113022728-113022728 on Build GRCh38
rs564845239 Chr.2: 113023042-113023042 on Build GRCh38
rs767208011 Chr.2: 113023108-113023108 on Build GRCh38
rs778854716 Chr.2: 113022814-113022814 on Build GRCh38
rs749087744 Chr.2: 113022705-113022705 on Build GRCh38
rs112276977 Chr.2: 113022841-113022841 on Build GRCh38
rs760355797 Chr.2: 113022785-113022785 on Build GRCh38
rs146086675 Chr.2: 113022893-113022893 on Build GRCh38
rs767032395 Chr.2: 113022693-113022693 on Build GRCh38
rs371066689 Chr.2: 113022805-113022805 on Build GRCh38
rs773647007 Chr.2: 113022762-113022762 on Build GRCh38
rs1005658388 Chr.2: 113023072-113023072 on Build GRCh38
rs964399942 Chr.2: 113023103-113023103 on Build GRCh38
rs777020603 Chr.2: 113022734-113022734 on Build GRCh38
rs758431269 Chr.2: 113022821-113022821 on Build GRCh38
rs113419369 Chr.2: 113022832-113022832 on Build GRCh38
rs754536213 Chr.2: 113022684-113022684 on Build GRCh38
rs549594008 Chr.2: 113022742-113022742 on Build GRCh38
rs1003592949 Chr.2: 113022712-113022712 on Build GRCh38
rs577125105 Chr.2: 113023079-113023079 on Build GRCh38
rs964304753 Chr.2: 113022797-113022797 on Build GRCh38
rs966818994 Chr.2: 113023028-113023028 on Build GRCh38
rs962660853 Chr.2: 113023086-113023086 on Build GRCh38
rs775728629 Chr.2: 113022837-113022837 on Build GRCh38
rs866451511 Chr.2: 113022789-113022789 on Build GRCh38
rs780311808 Chr.2: 113022726-113022726 on Build GRCh38
rs1045393734 Chr.2: 113022987-113022987 on Build GRCh38
rs893710089 Chr.2: 113022685-113022685 on Build GRCh38
rs141111717 Chr.2: 113022697-113022697 on Build GRCh38
rs1045825575 Chr.2: 113022842-113022842 on Build GRCh38
rs985303472 Chr.2: 113023083-113023083 on Build GRCh38
rs763696214 Chr.2: 113022790-113022790 on Build GRCh38

View Full Product Details