Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 40776046-40776431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NSUN7
Entrez Gene ID
79730
Gene Name
NOP2/Sun RNA methyltransferase family member 7
Gene Aliases
-
UniGene
Hs.570821
Gene Chromosome Location
Chr.4: 40749874-40810601 on Build GRCh38
Transcripts
NM_024677.4 XM_017008611.1 XM_017008612.1 XM_017008613.1 XM_017008614.1 XM_017008615.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3781158 c.838A>C p.S280R Chr.4: 40776061-40776061 on Build GRCh38
3781157 c.838A>C p.S280R Chr.4: 40776061-40776061 on Build GRCh38
1694264 c.862G>A p.A288T Chr.4: 40776085-40776085 on Build GRCh38
3825844 c.977T>C p.V326A Chr.4: 40776200-40776200 on Build GRCh38
4778153 c.922T>G p.S308A Chr.4: 40776145-40776145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145496822 Chr.4: 40776333-40776333 on Build GRCh38
rs746759353 Chr.4: 40776184-40776184 on Build GRCh38
rs150491454 Chr.4: 40776178-40776178 on Build GRCh38
rs776016399 Chr.4: 40776079-40776079 on Build GRCh38
rs556136647 Chr.4: 40776260-40776260 on Build GRCh38
rs775092872 Chr.4: 40776099-40776099 on Build GRCh38
rs750593395 Chr.4: 40776141-40776141 on Build GRCh38
rs768290847 Chr.4: 40776254-40776254 on Build GRCh38
rs544742106 Chr.4: 40776325-40776329 on Build GRCh38
rs1039775240 Chr.4: 40776407-40776407 on Build GRCh38
rs76338075 Chr.4: 40776246-40776246 on Build GRCh38
rs770830213 Chr.4: 40776203-40776203 on Build GRCh38
rs766728880 Chr.4: 40776305-40776305 on Build GRCh38
rs116478242 Chr.4: 40776225-40776225 on Build GRCh38
rs79812417 Chr.4: 40776248-40776248 on Build GRCh38
rs2437323 Chr.4: 40776145-40776145 on Build GRCh38
rs376365842 Chr.4: 40776218-40776218 on Build GRCh38
rs749539912 Chr.4: 40776150-40776150 on Build GRCh38
rs184478751 Chr.4: 40776315-40776315 on Build GRCh38
rs777790005 Chr.4: 40776361-40776361 on Build GRCh38
rs763357014 Chr.4: 40776113-40776113 on Build GRCh38
rs77596230 Chr.4: 40776239-40776239 on Build GRCh38
rs764389943 Chr.4: 40776119-40776119 on Build GRCh38
rs972412686 Chr.4: 40776349-40776349 on Build GRCh38
rs1025838555 Chr.4: 40776359-40776359 on Build GRCh38
rs572194795 Chr.4: 40776391-40776391 on Build GRCh38
rs780469058 Chr.4: 40776149-40776149 on Build GRCh38
rs866159976 Chr.4: 40776092-40776092 on Build GRCh38
rs751914094 Chr.4: 40776120-40776120 on Build GRCh38
rs750933153 Chr.4: 40776281-40776281 on Build GRCh38
rs755184425 Chr.4: 40776164-40776164 on Build GRCh38
rs764936168 Chr.4: 40776090-40776090 on Build GRCh38
rs761211206 Chr.4: 40776299-40776299 on Build GRCh38
rs371547373 Chr.4: 40776217-40776217 on Build GRCh38
rs992523156 Chr.4: 40776261-40776261 on Build GRCh38
rs763252579 Chr.4: 40776292-40776294 on Build GRCh38
rs376456213 Chr.4: 40776086-40776086 on Build GRCh38
rs77518783 Chr.4: 40776250-40776250 on Build GRCh38
rs767979060 Chr.4: 40776278-40776278 on Build GRCh38
rs2465570 Chr.4: 40776129-40776129 on Build GRCh38
rs776312450 Chr.4: 40776148-40776149 on Build GRCh38
rs781102326 Chr.4: 40776211-40776211 on Build GRCh38
rs762227614 Chr.4: 40776276-40776276 on Build GRCh38
rs979304063 Chr.4: 40776338-40776338 on Build GRCh38
rs576228289 Chr.4: 40776314-40776314 on Build GRCh38
rs1033992712 Chr.4: 40776231-40776231 on Build GRCh38
rs757711903 Chr.4: 40776122-40776122 on Build GRCh38
rs768257043 Chr.4: 40776137-40776138 on Build GRCh38

View Full Product Details