Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140668107-140668663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR55
Entrez Gene ID
54853
Gene Name
WD repeat domain 55
Gene Aliases
-
UniGene
Hs.286261
Gene Chromosome Location
Chr.5: 140664799-140674119 on Build GRCh38
Transcripts
NM_017706.4 XM_005268469.2 XM_017009600.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4616324 c.271G>A p.A91T Chr.5: 140668313-140668313 on Build GRCh38
6821958 c.206G>A p.C69Y Chr.5: 140668248-140668248 on Build GRCh38
1567480 c.278C>G p.S93C Chr.5: 140668320-140668320 on Build GRCh38
1433652 c.227A>T p.E76V Chr.5: 140668269-140668269 on Build GRCh38
7111206 c.252C>T p.L84L Chr.5: 140668294-140668294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs996061514 Chr.5: 140668354-140668354 on Build GRCh38
rs777062455 Chr.5: 140668315-140668315 on Build GRCh38
rs755130325 Chr.5: 140668629-140668629 on Build GRCh38
rs753555227 Chr.5: 140668580-140668580 on Build GRCh38
rs750499741 Chr.5: 140668494-140668494 on Build GRCh38
rs768951470 Chr.5: 140668526-140668526 on Build GRCh38
rs760336966 Chr.5: 140668477-140668477 on Build GRCh38
rs758618498 Chr.5: 140668507-140668507 on Build GRCh38
rs2251860 Chr.5: 140668624-140668624 on Build GRCh38
rs375815187 Chr.5: 140668484-140668484 on Build GRCh38
rs901047389 Chr.5: 140668320-140668320 on Build GRCh38
rs751110141 Chr.5: 140668356-140668356 on Build GRCh38
rs373321646 Chr.5: 140668345-140668345 on Build GRCh38
rs115157171 Chr.5: 140668399-140668399 on Build GRCh38
rs770166210 Chr.5: 140668541-140668541 on Build GRCh38
rs764420575 Chr.5: 140668501-140668503 on Build GRCh38
rs761710744 Chr.5: 140668595-140668595 on Build GRCh38
rs759083730 Chr.5: 140668473-140668473 on Build GRCh38
rs746012633 Chr.5: 140668268-140668268 on Build GRCh38
rs775536564 Chr.5: 140668269-140668269 on Build GRCh38
rs149703566 Chr.5: 140668561-140668561 on Build GRCh38
rs879803600 Chr.5: 140668597-140668597 on Build GRCh38
rs905908039 Chr.5: 140668432-140668432 on Build GRCh38
rs780922529 Chr.5: 140668217-140668217 on Build GRCh38
rs753701912 Chr.5: 140668483-140668483 on Build GRCh38
rs77675161 Chr.5: 140668182-140668182 on Build GRCh38
rs747374313 Chr.5: 140668416-140668416 on Build GRCh38
rs748591255 Chr.5: 140668535-140668535 on Build GRCh38
rs750444381 Chr.5: 140668608-140668608 on Build GRCh38
rs202074788 Chr.5: 140668414-140668414 on Build GRCh38
rs765738739 Chr.5: 140668350-140668350 on Build GRCh38
rs778960323 Chr.5: 140668491-140668491 on Build GRCh38
rs767871680 Chr.5: 140668574-140668574 on Build GRCh38
rs760642220 Chr.5: 140668304-140668304 on Build GRCh38
rs770977496 Chr.5: 140668255-140668255 on Build GRCh38
rs770354288 Chr.5: 140668462-140668462 on Build GRCh38
rs983768060 Chr.5: 140668176-140668176 on Build GRCh38
rs185373111 Chr.5: 140668231-140668231 on Build GRCh38
rs939783259 Chr.5: 140668198-140668198 on Build GRCh38
rs980402582 Chr.5: 140668139-140668139 on Build GRCh38
rs116171138 Chr.5: 140668160-140668160 on Build GRCh38
rs772402262 Chr.5: 140668406-140668406 on Build GRCh38
rs749126482 Chr.5: 140668380-140668380 on Build GRCh38
rs780240150 Chr.5: 140668618-140668618 on Build GRCh38
rs747161685 Chr.5: 140668519-140668519 on Build GRCh38
rs757227249 Chr.5: 140668384-140668384 on Build GRCh38
rs777607611 Chr.5: 140668234-140668234 on Build GRCh38
rs747955941 Chr.5: 140668218-140668218 on Build GRCh38
rs565515996 Chr.5: 140668605-140668608 on Build GRCh38
rs202204644 Chr.5: 140668273-140668273 on Build GRCh38
rs771516564 Chr.5: 140668563-140668563 on Build GRCh38
rs978996326 Chr.5: 140668557-140668557 on Build GRCh38
rs145791745 Chr.5: 140668577-140668577 on Build GRCh38
rs751113054 Chr.5: 140668185-140668185 on Build GRCh38
rs764665086 Chr.5: 140668305-140668305 on Build GRCh38
rs200455085 Chr.5: 140668400-140668400 on Build GRCh38
rs762183873 Chr.5: 140668459-140668459 on Build GRCh38
rs34170825 Chr.5: 140668140-140668140 on Build GRCh38
rs112617106 Chr.5: 140668366-140668366 on Build GRCh38
rs769083158 Chr.5: 140668425-140668425 on Build GRCh38
rs952297794 Chr.5: 140668614-140668616 on Build GRCh38
rs1020601395 Chr.5: 140668510-140668510 on Build GRCh38
rs763072507 Chr.5: 140668244-140668244 on Build GRCh38
rs369331487 Chr.5: 140668285-140668285 on Build GRCh38
rs73791744 Chr.5: 140668171-140668171 on Build GRCh38
rs1031684276 Chr.5: 140668419-140668419 on Build GRCh38
rs758413132 Chr.5: 140668613-140668613 on Build GRCh38
rs201721516 Chr.5: 140668474-140668474 on Build GRCh38
rs763994589 Chr.5: 140668335-140668335 on Build GRCh38
rs754627955 Chr.5: 140668203-140668203 on Build GRCh38
rs559813671 Chr.5: 140668161-140668161 on Build GRCh38
rs765081173 Chr.5: 140668602-140668602 on Build GRCh38
rs777057228 Chr.5: 140668456-140668456 on Build GRCh38
rs115837083 Chr.5: 140668259-140668259 on Build GRCh38
rs377745187 Chr.5: 140668463-140668463 on Build GRCh38
rs781514355 Chr.5: 140668631-140668631 on Build GRCh38
rs749385762 Chr.5: 140668249-140668249 on Build GRCh38
rs773709102 Chr.5: 140668464-140668464 on Build GRCh38
rs752408724 Chr.5: 140668475-140668475 on Build GRCh38
rs748453005 Chr.5: 140668633-140668633 on Build GRCh38
rs570371409 Chr.5: 140668375-140668375 on Build GRCh38
rs988837636 Chr.5: 140668502-140668502 on Build GRCh38
rs773771462 Chr.5: 140668547-140668547 on Build GRCh38
rs760283614 Chr.5: 140668576-140668576 on Build GRCh38
rs373304228 Chr.5: 140668377-140668377 on Build GRCh38
rs1000090517 Chr.5: 140668415-140668415 on Build GRCh38
rs763367694 Chr.5: 140668403-140668403 on Build GRCh38
rs780293557 Chr.5: 140668511-140668511 on Build GRCh38
rs754573024 Chr.5: 140668363-140668363 on Build GRCh38
rs781361784 Chr.5: 140668534-140668534 on Build GRCh38

View Full Product Details