Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 139044099-139044688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZC3HAV1
Entrez Gene ID
56829
Gene Name
zinc finger CCCH-type containing, antiviral 1
Gene Aliases
ARTD13, FLB6421, PARP13, ZAP, ZC3H2, ZC3HDC2
UniGene
Hs.133512
Gene Chromosome Location
Chr.7: 139043519-139109720 on Build GRCh38
Transcripts
NM_020119.3 XM_005250501.2

SNP Detail

SNP ID Chromosome Location
rs1052976207 Chr.7: 139044370-139044370 on Build GRCh38
rs569167850 Chr.7: 139044648-139044648 on Build GRCh38
rs935177231 Chr.7: 139044574-139044574 on Build GRCh38
rs999452800 Chr.7: 139044638-139044638 on Build GRCh38
rs74606736 Chr.7: 139044279-139044279 on Build GRCh38
rs979339068 Chr.7: 139044515-139044515 on Build GRCh38
rs1023857604 Chr.7: 139044390-139044390 on Build GRCh38
rs551560660 Chr.7: 139044213-139044213 on Build GRCh38
rs972482459 Chr.7: 139044438-139044438 on Build GRCh38
rs942616863 Chr.7: 139044513-139044513 on Build GRCh38
rs896572956 Chr.7: 139044623-139044623 on Build GRCh38
rs896464244 Chr.7: 139044374-139044374 on Build GRCh38
rs1006517618 Chr.7: 139044406-139044406 on Build GRCh38
rs949532283 Chr.7: 139044634-139044634 on Build GRCh38
rs1016475631 Chr.7: 139044143-139044143 on Build GRCh38
rs925522536 Chr.7: 139044248-139044248 on Build GRCh38
rs1001347355 Chr.7: 139044183-139044183 on Build GRCh38
rs548024418 Chr.7: 139044226-139044226 on Build GRCh38
rs1025049901 Chr.7: 139044207-139044207 on Build GRCh38
rs933663241 Chr.7: 139044265-139044265 on Build GRCh38
rs762118411 Chr.7: 139044405-139044405 on Build GRCh38
rs773481850 Chr.7: 139044657-139044657 on Build GRCh38
rs1014008332 Chr.7: 139044389-139044389 on Build GRCh38
rs1014098188 Chr.7: 139044200-139044200 on Build GRCh38
rs530308718 Chr.7: 139044280-139044280 on Build GRCh38
rs548914058 Chr.7: 139044398-139044398 on Build GRCh38
rs1048324940 Chr.7: 139044138-139044138 on Build GRCh38
rs959448408 Chr.7: 139044195-139044195 on Build GRCh38
rs547117066 Chr.7: 139044499-139044499 on Build GRCh38
rs7798511 Chr.7: 139044641-139044641 on Build GRCh38
rs1016423202 Chr.7: 139044408-139044408 on Build GRCh38
rs531549315 Chr.7: 139044191-139044191 on Build GRCh38
rs910922953 Chr.7: 139044482-139044482 on Build GRCh38
rs1045470626 Chr.7: 139044642-139044642 on Build GRCh38
rs962233998 Chr.7: 139044435-139044435 on Build GRCh38
rs972501841 Chr.7: 139044221-139044221 on Build GRCh38
rs955027071 Chr.7: 139044476-139044476 on Build GRCh38
rs187358934 Chr.7: 139044512-139044512 on Build GRCh38
rs765814346 Chr.7: 139044193-139044193 on Build GRCh38
rs986879157 Chr.7: 139044481-139044481 on Build GRCh38
rs966681606 Chr.7: 139044220-139044220 on Build GRCh38
rs888391862 Chr.7: 139044147-139044147 on Build GRCh38
rs911131514 Chr.7: 139044328-139044328 on Build GRCh38
rs560020353 Chr.7: 139044278-139044278 on Build GRCh38
rs758455400 Chr.7: 139044656-139044656 on Build GRCh38
rs75375718 Chr.7: 139044605-139044605 on Build GRCh38
rs905218336 Chr.7: 139044647-139044647 on Build GRCh38
rs1025833679 Chr.7: 139044456-139044456 on Build GRCh38
rs373443867 Chr.7: 139044359-139044359 on Build GRCh38
rs925103593 Chr.7: 139044563-139044563 on Build GRCh38
rs139247573 Chr.7: 139044350-139044350 on Build GRCh38
rs943767570 Chr.7: 139044330-139044330 on Build GRCh38
rs918364083 Chr.7: 139044351-139044351 on Build GRCh38
rs535844710 Chr.7: 139044590-139044590 on Build GRCh38
rs942502174 Chr.7: 139044329-139044329 on Build GRCh38
rs903733741 Chr.7: 139044355-139044355 on Build GRCh38

View Full Product Details