Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 149166713-149167235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF398
Entrez Gene ID
57541
Gene Name
zinc finger protein 398
Gene Aliases
P51, P71, ZER6
UniGene
Hs.632032
Gene Chromosome Location
Chr.7: 149126416-149183042 on Build GRCh38
Transcripts
NM_020781.3 NM_170686.2 XM_011516439.2 XM_011516440.2 XM_011516441.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7403278 c.583A>G p.I195V Chr.7: 149166852-149166852 on Build GRCh38
3635802 c.578C>T p.S193F Chr.7: 149166847-149166847 on Build GRCh38
461663 c.586C>G p.Q196E Chr.7: 149166855-149166855 on Build GRCh38
1699547 c.643C>T p.P215S Chr.7: 149166912-149166912 on Build GRCh38
351565 c.645C>T p.P215P Chr.7: 149166914-149166914 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965213418 Chr.7: 149167050-149167050 on Build GRCh38
rs764226409 Chr.7: 149166983-149166983 on Build GRCh38
rs76143098 Chr.7: 149166747-149166747 on Build GRCh38
rs755067688 Chr.7: 149166844-149166844 on Build GRCh38
rs369700851 Chr.7: 149166976-149166976 on Build GRCh38
rs140986850 Chr.7: 149166916-149166916 on Build GRCh38
rs777143208 Chr.7: 149166884-149166884 on Build GRCh38
rs745813941 Chr.7: 149166831-149166831 on Build GRCh38
rs151094307 Chr.7: 149166981-149166981 on Build GRCh38
rs764753757 Chr.7: 149166793-149166793 on Build GRCh38
rs772479210 Chr.7: 149166857-149166857 on Build GRCh38
rs748586769 Chr.7: 149166796-149166799 on Build GRCh38
rs377069952 Chr.7: 149167027-149167027 on Build GRCh38
rs183810146 Chr.7: 149167064-149167064 on Build GRCh38
rs939044685 Chr.7: 149167033-149167033 on Build GRCh38
rs976776608 Chr.7: 149167063-149167063 on Build GRCh38
rs1036524290 Chr.7: 149167132-149167132 on Build GRCh38
rs181189181 Chr.7: 149166775-149166775 on Build GRCh38
rs896800900 Chr.7: 149166955-149166955 on Build GRCh38
rs985821366 Chr.7: 149166982-149166982 on Build GRCh38
rs751430993 Chr.7: 149166801-149166801 on Build GRCh38
rs906671340 Chr.7: 149167195-149167195 on Build GRCh38
rs771208538 Chr.7: 149166873-149166873 on Build GRCh38
rs779122063 Chr.7: 149166949-149166949 on Build GRCh38
rs760071005 Chr.7: 149166888-149166888 on Build GRCh38
rs12532104 Chr.7: 149167141-149167141 on Build GRCh38
rs762584561 Chr.7: 149166919-149166919 on Build GRCh38
rs58653603 Chr.7: 149167198-149167198 on Build GRCh38
rs763875425 Chr.7: 149166920-149166920 on Build GRCh38
rs543327506 Chr.7: 149166940-149166940 on Build GRCh38
rs747330143 Chr.7: 149166871-149166871 on Build GRCh38
rs1035449475 Chr.7: 149166900-149166900 on Build GRCh38
rs371624152 Chr.7: 149166852-149166852 on Build GRCh38
rs144380452 Chr.7: 149166820-149166820 on Build GRCh38
rs368726259 Chr.7: 149166859-149166859 on Build GRCh38
rs750354375 Chr.7: 149166942-149166942 on Build GRCh38
rs948310373 Chr.7: 149167155-149167155 on Build GRCh38
rs772390697 Chr.7: 149166963-149166963 on Build GRCh38
rs761611005 Chr.7: 149166921-149166921 on Build GRCh38
rs188183256 Chr.7: 149167115-149167115 on Build GRCh38
rs374337614 Chr.7: 149166795-149166795 on Build GRCh38
rs180725203 Chr.7: 149167120-149167120 on Build GRCh38
rs758089075 Chr.7: 149166798-149166798 on Build GRCh38
rs761071461 Chr.7: 149166776-149166776 on Build GRCh38
rs995047765 Chr.7: 149167172-149167172 on Build GRCh38
rs755567790 Chr.7: 149166769-149166769 on Build GRCh38
rs904822316 Chr.7: 149166995-149166995 on Build GRCh38
rs1044965447 Chr.7: 149166990-149166990 on Build GRCh38
rs763777932 Chr.7: 149166800-149166800 on Build GRCh38
rs866917529 Chr.7: 149167166-149167166 on Build GRCh38
rs1029251601 Chr.7: 149167206-149167206 on Build GRCh38
rs777005064 Chr.7: 149166778-149166778 on Build GRCh38
rs906717360 Chr.7: 149166840-149166840 on Build GRCh38
rs935296126 Chr.7: 149167083-149167083 on Build GRCh38
rs779173522 Chr.7: 149166850-149166850 on Build GRCh38
rs769267456 Chr.7: 149166907-149166907 on Build GRCh38
rs1018089729 Chr.7: 149167017-149167017 on Build GRCh38
rs374882084 Chr.7: 149166817-149166817 on Build GRCh38
rs758985797 Chr.7: 149166792-149166792 on Build GRCh38
rs575928152 Chr.7: 149167108-149167108 on Build GRCh38
rs377203540 Chr.7: 149166966-149166966 on Build GRCh38
rs529183157 Chr.7: 149167197-149167197 on Build GRCh38
rs779266785 Chr.7: 149166791-149166792 on Build GRCh38
rs757139929 Chr.7: 149166813-149166813 on Build GRCh38
rs894062282 Chr.7: 149166751-149166751 on Build GRCh38
rs756168832 Chr.7: 149166944-149166944 on Build GRCh38
rs766974305 Chr.7: 149166777-149166777 on Build GRCh38
rs767238632 Chr.7: 149166922-149166922 on Build GRCh38
rs752889353 Chr.7: 149166960-149166960 on Build GRCh38
rs1045754767 Chr.7: 149167173-149167173 on Build GRCh38

View Full Product Details