Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 63894809-63895363 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINB2
Entrez Gene ID
5055
Gene Name
serpin family B member 2
Gene Aliases
HsT1201, PAI, PAI-2, PAI2, PLANH2
UniGene
Hs.594481
Gene Chromosome Location
Chr.18: 63887705-63903890 on Build GRCh38
Transcripts
NM_001143818.1 NM_002575.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3891254 c.243C>T p.F81F Chr.18: 63895338-63895338 on Build GRCh38
4931714 c.245T>C p.M82T Chr.18: 63895340-63895340 on Build GRCh38
4950382 c.169G>A p.V57M Chr.18: 63895264-63895264 on Build GRCh38
6083425 c.246G>T p.M82I Chr.18: 63895341-63895341 on Build GRCh38
1711484 c.263G>A p.G88D Chr.18: 63895358-63895358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543048163 Chr.18: 63894934-63894934 on Build GRCh38
rs767454836 Chr.18: 63895245-63895246 on Build GRCh38
rs192464851 Chr.18: 63894953-63894953 on Build GRCh38
rs377690463 Chr.18: 63894942-63894942 on Build GRCh38
rs200884637 Chr.18: 63895320-63895320 on Build GRCh38
rs1050014658 Chr.18: 63895177-63895177 on Build GRCh38
rs199718055 Chr.18: 63895241-63895241 on Build GRCh38
rs138183373 Chr.18: 63895334-63895334 on Build GRCh38
rs948239805 Chr.18: 63895176-63895176 on Build GRCh38
rs368507799 Chr.18: 63895256-63895256 on Build GRCh38
rs769963428 Chr.18: 63895295-63895295 on Build GRCh38
rs754713165 Chr.18: 63895278-63895278 on Build GRCh38
rs771915183 Chr.18: 63895319-63895319 on Build GRCh38
rs746442179 Chr.18: 63895289-63895289 on Build GRCh38
rs749422897 Chr.18: 63895301-63895301 on Build GRCh38
rs530259021 Chr.18: 63895135-63895135 on Build GRCh38
rs113144148 Chr.18: 63894840-63894840 on Build GRCh38
rs868228579 Chr.18: 63895248-63895248 on Build GRCh38
rs7228512 Chr.18: 63894999-63894999 on Build GRCh38
rs777955847 Chr.18: 63895279-63895279 on Build GRCh38
rs867732518 Chr.18: 63894943-63894943 on Build GRCh38
rs7229383 Chr.18: 63895157-63895157 on Build GRCh38
rs560159671 Chr.18: 63895143-63895143 on Build GRCh38
rs145713562 Chr.18: 63895311-63895311 on Build GRCh38
rs1040712588 Chr.18: 63895200-63895200 on Build GRCh38
rs898776902 Chr.18: 63894848-63894848 on Build GRCh38
rs761615586 Chr.18: 63895247-63895247 on Build GRCh38
rs565139632 Chr.18: 63895036-63895036 on Build GRCh38
rs137994008 Chr.18: 63895272-63895272 on Build GRCh38
rs371462289 Chr.18: 63895225-63895225 on Build GRCh38
rs896931796 Chr.18: 63894924-63894924 on Build GRCh38
rs764519142 Chr.18: 63895007-63895007 on Build GRCh38
rs931461195 Chr.18: 63895161-63895161 on Build GRCh38
rs7233370 Chr.18: 63895137-63895137 on Build GRCh38
rs763773946 Chr.18: 63895237-63895237 on Build GRCh38
rs763381308 Chr.18: 63895004-63895004 on Build GRCh38
rs754822775 Chr.18: 63895105-63895105 on Build GRCh38
rs976212614 Chr.18: 63895045-63895045 on Build GRCh38
rs757644603 Chr.18: 63895286-63895286 on Build GRCh38
rs538139326 Chr.18: 63894909-63894909 on Build GRCh38
rs553457885 Chr.18: 63894950-63894950 on Build GRCh38
rs1001305772 Chr.18: 63894949-63894949 on Build GRCh38
rs1030347032 Chr.18: 63895057-63895057 on Build GRCh38
rs763276518 Chr.18: 63895233-63895233 on Build GRCh38
rs755611573 Chr.18: 63895271-63895271 on Build GRCh38
rs146281294 Chr.18: 63895195-63895195 on Build GRCh38
rs757500045 Chr.18: 63895166-63895166 on Build GRCh38
rs770056378 Chr.18: 63895227-63895227 on Build GRCh38
rs574725859 Chr.18: 63894931-63894931 on Build GRCh38
rs980445291 Chr.18: 63895079-63895079 on Build GRCh38
rs775761168 Chr.18: 63895298-63895298 on Build GRCh38
rs940087800 Chr.18: 63895134-63895134 on Build GRCh38
rs1056591216 Chr.18: 63894933-63894934 on Build GRCh38
rs781760818 Chr.18: 63895287-63895287 on Build GRCh38
rs765855871 Chr.18: 63895333-63895333 on Build GRCh38
rs927173564 Chr.18: 63895081-63895081 on Build GRCh38
rs557250182 Chr.18: 63895114-63895114 on Build GRCh38
rs568919651 Chr.18: 63895189-63895189 on Build GRCh38
rs187935580 Chr.18: 63894868-63894868 on Build GRCh38
rs141835206 Chr.18: 63895088-63895088 on Build GRCh38
rs771495272 Chr.18: 63895216-63895216 on Build GRCh38
rs1012386066 Chr.18: 63894937-63894937 on Build GRCh38
rs753702164 Chr.18: 63895073-63895073 on Build GRCh38
rs1021862319 Chr.18: 63895065-63895065 on Build GRCh38
rs552747433 Chr.18: 63895190-63895190 on Build GRCh38
rs957031178 Chr.18: 63895000-63895000 on Build GRCh38
rs749879244 Chr.18: 63895263-63895263 on Build GRCh38
rs768953477 Chr.18: 63895302-63895302 on Build GRCh38
rs144193829 Chr.18: 63894991-63894991 on Build GRCh38
rs996176802 Chr.18: 63894893-63894893 on Build GRCh38
rs377037100 Chr.18: 63895339-63895339 on Build GRCh38
rs554800742 Chr.18: 63894917-63894917 on Build GRCh38
rs532118979 Chr.18: 63895147-63895147 on Build GRCh38
rs761682552 Chr.18: 63895315-63895315 on Build GRCh38
rs1049988479 Chr.18: 63894895-63894895 on Build GRCh38
rs543125041 Chr.18: 63894939-63894939 on Build GRCh38
rs747400109 Chr.18: 63895281-63895281 on Build GRCh38
rs375613757 Chr.18: 63895133-63895133 on Build GRCh38
rs775849214 Chr.18: 63895229-63895229 on Build GRCh38

View Full Product Details