Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 82848713-82849171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEC31A
Entrez Gene ID
22872
Gene Name
SEC31 homolog A, COPII coat complex component
Gene Aliases
ABP125, ABP130, HSPC275, HSPC334, SEC31L1
UniGene
Hs.370024
Gene Chromosome Location
Chr.4: 82818509-82900571 on Build GRCh38
Transcripts
NM_001077206.3 NM_001077207.3 NM_001077208.3 NM_001191049.2 NM_001300744.2 NM_001300745.2 NM_001318119.1 NM_001318120.1 NM_016211.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6388289 c.2439G>T p.R813S Chr.4: 82848867-82848867 on Build GRCh38
1685569 c.2408delC p.P803fs*42 Chr.4: 82848898-82848898 on Build GRCh38
6330475 c.2408C>T p.P803L Chr.4: 82848898-82848898 on Build GRCh38
481599 c.2389C>A p.H797N Chr.4: 82848917-82848917 on Build GRCh38
1694615 c.2407C>T p.P803S Chr.4: 82848899-82848899 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970032310 Chr.4: 82849083-82849083 on Build GRCh38
rs376229559 Chr.4: 82849018-82849018 on Build GRCh38
rs940152832 Chr.4: 82848767-82848768 on Build GRCh38
rs145039719 Chr.4: 82848897-82848897 on Build GRCh38
rs200045009 Chr.4: 82848917-82848917 on Build GRCh38
rs367848552 Chr.4: 82848893-82848893 on Build GRCh38
rs747593971 Chr.4: 82849007-82849007 on Build GRCh38
rs767081772 Chr.4: 82849008-82849008 on Build GRCh38
rs748035305 Chr.4: 82849056-82849056 on Build GRCh38
rs373124024 Chr.4: 82849009-82849009 on Build GRCh38
rs766065708 Chr.4: 82848768-82848768 on Build GRCh38
rs981492790 Chr.4: 82848746-82848746 on Build GRCh38
rs747042104 Chr.4: 82848959-82848959 on Build GRCh38
rs768992157 Chr.4: 82848991-82848992 on Build GRCh38
rs149019274 Chr.4: 82848960-82848960 on Build GRCh38
rs748722172 Chr.4: 82848879-82848879 on Build GRCh38
rs141183010 Chr.4: 82848894-82848894 on Build GRCh38
rs1003202365 Chr.4: 82849097-82849097 on Build GRCh38
rs200755860 Chr.4: 82848874-82848874 on Build GRCh38
rs528045930 Chr.4: 82849119-82849119 on Build GRCh38
rs370331407 Chr.4: 82849025-82849025 on Build GRCh38
rs376688057 Chr.4: 82848997-82848997 on Build GRCh38
rs750135300 Chr.4: 82848936-82848936 on Build GRCh38
rs773302646 Chr.4: 82848758-82848758 on Build GRCh38
rs756889677 Chr.4: 82848858-82848858 on Build GRCh38
rs368774877 Chr.4: 82848889-82848889 on Build GRCh38
rs780743504 Chr.4: 82848862-82848862 on Build GRCh38
rs559324984 Chr.4: 82849109-82849109 on Build GRCh38
rs777405265 Chr.4: 82848836-82848836 on Build GRCh38
rs984172280 Chr.4: 82849124-82849124 on Build GRCh38
rs770928244 Chr.4: 82848967-82848967 on Build GRCh38
rs761898461 Chr.4: 82849114-82849115 on Build GRCh38
rs757547019 Chr.4: 82848776-82848776 on Build GRCh38
rs748581886 Chr.4: 82848947-82848947 on Build GRCh38
rs776977004 Chr.4: 82848974-82848974 on Build GRCh38
rs776995560 Chr.4: 82848898-82848898 on Build GRCh38
rs868858614 Chr.4: 82848941-82848941 on Build GRCh38
rs759344068 Chr.4: 82848906-82848906 on Build GRCh38
rs760997715 Chr.4: 82848760-82848760 on Build GRCh38
rs372081597 Chr.4: 82848945-82848945 on Build GRCh38
rs759434094 Chr.4: 82848790-82848790 on Build GRCh38
rs755334090 Chr.4: 82848943-82848943 on Build GRCh38
rs369214708 Chr.4: 82848990-82848990 on Build GRCh38
rs769024482 Chr.4: 82848869-82848869 on Build GRCh38
rs746623501 Chr.4: 82849026-82849026 on Build GRCh38
rs573434292 Chr.4: 82849141-82849141 on Build GRCh38
rs543639686 Chr.4: 82848924-82848924 on Build GRCh38
rs201709543 Chr.4: 82849002-82849002 on Build GRCh38
rs752673057 Chr.4: 82848914-82848914 on Build GRCh38
rs748828860 Chr.4: 82848767-82848767 on Build GRCh38
rs780548665 Chr.4: 82848988-82848988 on Build GRCh38
rs749591409 Chr.4: 82848867-82848867 on Build GRCh38
rs750849638 Chr.4: 82848838-82848838 on Build GRCh38
rs144888568 Chr.4: 82848958-82848958 on Build GRCh38
rs147804203 Chr.4: 82848878-82848878 on Build GRCh38
rs181693669 Chr.4: 82849070-82849070 on Build GRCh38
rs751137305 Chr.4: 82848843-82848843 on Build GRCh38
rs190198098 Chr.4: 82849036-82849036 on Build GRCh38
rs889443293 Chr.4: 82848939-82848939 on Build GRCh38
rs755814035 Chr.4: 82849011-82849011 on Build GRCh38
rs781376339 Chr.4: 82848816-82848816 on Build GRCh38
rs117533578 Chr.4: 82848916-82848916 on Build GRCh38
rs780767332 Chr.4: 82848930-82848930 on Build GRCh38
rs781251320 Chr.4: 82848827-82848827 on Build GRCh38
rs867303444 Chr.4: 82849130-82849130 on Build GRCh38
rs772013899 Chr.4: 82848884-82848884 on Build GRCh38
rs772581729 Chr.4: 82848954-82848954 on Build GRCh38
rs371000483 Chr.4: 82848793-82848793 on Build GRCh38
rs756726271 Chr.4: 82848928-82848928 on Build GRCh38
rs765178221 Chr.4: 82848909-82848909 on Build GRCh38
rs1023204428 Chr.4: 82849079-82849079 on Build GRCh38

View Full Product Details