Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 154275310-154275885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX36
Entrez Gene ID
170506
Gene Name
DEAH-box helicase 36
Gene Aliases
DDX36, G4R1, MLEL1, RHAU
UniGene
Hs.446270
Gene Chromosome Location
Chr.3: 154275668-154324497 on Build GRCh38
Transcripts
NM_001114397.1 NM_020865.2

SNP Detail

SNP ID Chromosome Location
rs746689850 Chr.3: 154275659-154275659 on Build GRCh38
rs900536081 Chr.3: 154275589-154275589 on Build GRCh38
rs969944379 Chr.3: 154275436-154275436 on Build GRCh38
rs922793118 Chr.3: 154275740-154275740 on Build GRCh38
rs747161984 Chr.3: 154275418-154275418 on Build GRCh38
rs1049699260 Chr.3: 154275603-154275603 on Build GRCh38
rs1034652825 Chr.3: 154275699-154275699 on Build GRCh38
rs763460864 Chr.3: 154275797-154275797 on Build GRCh38
rs920770580 Chr.3: 154275472-154275474 on Build GRCh38
rs529462021 Chr.3: 154275753-154275753 on Build GRCh38
rs969744491 Chr.3: 154275681-154275681 on Build GRCh38
rs1047923545 Chr.3: 154275516-154275516 on Build GRCh38
rs182877702 Chr.3: 154275563-154275563 on Build GRCh38
rs144399858 Chr.3: 154275700-154275700 on Build GRCh38
rs568280166 Chr.3: 154275348-154275348 on Build GRCh38
rs776565045 Chr.3: 154275538-154275538 on Build GRCh38
rs931942996 Chr.3: 154275803-154275803 on Build GRCh38
rs887999444 Chr.3: 154275849-154275849 on Build GRCh38
rs112801501 Chr.3: 154275732-154275732 on Build GRCh38
rs925767509 Chr.3: 154275443-154275443 on Build GRCh38
rs988633616 Chr.3: 154275734-154275734 on Build GRCh38
rs68002435 Chr.3: 154275474-154275475 on Build GRCh38
rs72996619 Chr.3: 154275731-154275731 on Build GRCh38
rs142215991 Chr.3: 154275678-154275678 on Build GRCh38
rs1015518697 Chr.3: 154275644-154275644 on Build GRCh38
rs1010761743 Chr.3: 154275647-154275647 on Build GRCh38
rs983649758 Chr.3: 154275783-154275783 on Build GRCh38
rs979773101 Chr.3: 154275692-154275692 on Build GRCh38
rs900463605 Chr.3: 154275592-154275593 on Build GRCh38
rs199575464 Chr.3: 154275472-154275472 on Build GRCh38
rs944793060 Chr.3: 154275760-154275760 on Build GRCh38
rs930863009 Chr.3: 154275494-154275494 on Build GRCh38
rs1040365410 Chr.3: 154275588-154275588 on Build GRCh38
rs888030669 Chr.3: 154275605-154275605 on Build GRCh38
rs762005142 Chr.3: 154275592-154275592 on Build GRCh38
rs1015302456 Chr.3: 154275359-154275359 on Build GRCh38
rs772957132 Chr.3: 154275562-154275562 on Build GRCh38
rs1040416983 Chr.3: 154275557-154275557 on Build GRCh38
rs35799657 Chr.3: 154275473-154275474 on Build GRCh38
rs1049067520 Chr.3: 154275845-154275845 on Build GRCh38
rs1003226195 Chr.3: 154275685-154275685 on Build GRCh38
rs1004981598 Chr.3: 154275632-154275632 on Build GRCh38
rs1053366188 Chr.3: 154275541-154275541 on Build GRCh38
rs1014444635 Chr.3: 154275662-154275662 on Build GRCh38
rs192391095 Chr.3: 154275433-154275433 on Build GRCh38
rs1020698730 Chr.3: 154275344-154275344 on Build GRCh38
rs1049267422 Chr.3: 154275604-154275604 on Build GRCh38
rs959910827 Chr.3: 154275386-154275386 on Build GRCh38
rs913292277 Chr.3: 154275542-154275542 on Build GRCh38
rs116253421 Chr.3: 154275352-154275352 on Build GRCh38
rs1040899900 Chr.3: 154275762-154275762 on Build GRCh38
rs769070756 Chr.3: 154275442-154275442 on Build GRCh38
rs72996616 Chr.3: 154275379-154275379 on Build GRCh38
rs916929448 Chr.3: 154275435-154275435 on Build GRCh38
rs935787073 Chr.3: 154275519-154275519 on Build GRCh38
rs201687778 Chr.3: 154275473-154275473 on Build GRCh38
rs944802315 Chr.3: 154275551-154275551 on Build GRCh38

View Full Product Details