Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 18504622-18505153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTSL1
Entrez Gene ID
92949
Gene Name
ADAMTS like 1
Gene Aliases
ADAMTSL-1, ADAMTSR1, C9orf94, PUNCTIN
UniGene
Hs.741483
Gene Chromosome Location
Chr.9: 17906574-18910950 on Build GRCh38
Transcripts
NM_001040272.5 NM_052866.4 XM_011518063.2 XM_011518064.2 XM_017015310.1 XM_017015311.1 XM_017015312.1 XM_017015313.1 XM_017015314.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
229120 c.100G>A p.G34S Chr.9: 18504865-18504865 on Build GRCh38
5932347 c.83C>T p.S28F Chr.9: 18504848-18504848 on Build GRCh38
158915 c.68C>G p.S23C Chr.9: 18504833-18504833 on Build GRCh38
4519445 c.100G>A p.G34S Chr.9: 18504865-18504865 on Build GRCh38
3728810 c.191+6T>C p.? Chr.9: 18504962-18504962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376196071 Chr.9: 18504874-18504874 on Build GRCh38
rs955500157 Chr.9: 18505085-18505085 on Build GRCh38
rs973160820 Chr.9: 18505075-18505075 on Build GRCh38
rs895247144 Chr.9: 18505018-18505018 on Build GRCh38
rs756911048 Chr.9: 18504839-18504839 on Build GRCh38
rs1016873753 Chr.9: 18505051-18505051 on Build GRCh38
rs921325649 Chr.9: 18504835-18504835 on Build GRCh38
rs530004896 Chr.9: 18505049-18505049 on Build GRCh38
rs753067752 Chr.9: 18504989-18504989 on Build GRCh38
rs776044423 Chr.9: 18504961-18504984 on Build GRCh38
rs893734843 Chr.9: 18504693-18504693 on Build GRCh38
rs755856772 Chr.9: 18504976-18504976 on Build GRCh38
rs769277257 Chr.9: 18504855-18504855 on Build GRCh38
rs755330972 Chr.9: 18504914-18504914 on Build GRCh38
rs959127248 Chr.9: 18504709-18504709 on Build GRCh38
rs73421026 Chr.9: 18504773-18504773 on Build GRCh38
rs1004457225 Chr.9: 18504667-18504667 on Build GRCh38
rs760916992 Chr.9: 18504864-18504864 on Build GRCh38
rs771011730 Chr.9: 18504930-18504930 on Build GRCh38
rs866189730 Chr.9: 18504841-18504841 on Build GRCh38
rs1003718784 Chr.9: 18504700-18504700 on Build GRCh38
rs538389320 Chr.9: 18504771-18504771 on Build GRCh38
rs772060822 Chr.9: 18504787-18504787 on Build GRCh38
rs1021947230 Chr.9: 18504736-18504736 on Build GRCh38
rs1055406692 Chr.9: 18505117-18505117 on Build GRCh38
rs182961542 Chr.9: 18504655-18504655 on Build GRCh38
rs559045248 Chr.9: 18505048-18505048 on Build GRCh38
rs776006996 Chr.9: 18504820-18504820 on Build GRCh38
rs2277160 Chr.9: 18504918-18504918 on Build GRCh38
rs1035152254 Chr.9: 18504776-18504776 on Build GRCh38
rs751071846 Chr.9: 18504888-18504888 on Build GRCh38
rs750041158 Chr.9: 18505094-18505094 on Build GRCh38
rs906783831 Chr.9: 18504881-18504881 on Build GRCh38
rs994371535 Chr.9: 18505073-18505073 on Build GRCh38
rs151197424 Chr.9: 18504656-18504656 on Build GRCh38
rs966801244 Chr.9: 18504744-18504744 on Build GRCh38
rs142532642 Chr.9: 18504956-18504956 on Build GRCh38
rs951906957 Chr.9: 18505015-18505015 on Build GRCh38
rs749737333 Chr.9: 18504850-18504850 on Build GRCh38
rs769527362 Chr.9: 18504940-18504940 on Build GRCh38
rs374230656 Chr.9: 18505004-18505004 on Build GRCh38
rs370892034 Chr.9: 18504923-18504923 on Build GRCh38
rs529911200 Chr.9: 18504659-18504659 on Build GRCh38
rs773419466 Chr.9: 18504789-18504789 on Build GRCh38
rs895343655 Chr.9: 18504912-18504912 on Build GRCh38
rs370805538 Chr.9: 18504795-18504795 on Build GRCh38
rs1033388702 Chr.9: 18504701-18504701 on Build GRCh38
rs139254572 Chr.9: 18504794-18504794 on Build GRCh38
rs765662226 Chr.9: 18504978-18504978 on Build GRCh38
rs764878763 Chr.9: 18505089-18505089 on Build GRCh38
rs144094882 Chr.9: 18504868-18504868 on Build GRCh38
rs778080712 Chr.9: 18505001-18505001 on Build GRCh38
rs770264028 Chr.9: 18504930-18504931 on Build GRCh38
rs370752330 Chr.9: 18504861-18504861 on Build GRCh38
rs780536559 Chr.9: 18504844-18504844 on Build GRCh38
rs748610470 Chr.9: 18504859-18504859 on Build GRCh38
rs779507767 Chr.9: 18504856-18504856 on Build GRCh38
rs999605643 Chr.9: 18504929-18504929 on Build GRCh38
rs759460223 Chr.9: 18504805-18504805 on Build GRCh38
rs376026489 Chr.9: 18504811-18504811 on Build GRCh38
rs543148065 Chr.9: 18504947-18504947 on Build GRCh38
rs370844113 Chr.9: 18504972-18504972 on Build GRCh38
rs928170886 Chr.9: 18505088-18505088 on Build GRCh38
rs764005243 Chr.9: 18504967-18504967 on Build GRCh38
rs539472601 Chr.9: 18504696-18504696 on Build GRCh38
rs747112161 Chr.9: 18504924-18504924 on Build GRCh38
rs776766953 Chr.9: 18504933-18504933 on Build GRCh38
rs909994016 Chr.9: 18505055-18505055 on Build GRCh38
rs367593235 Chr.9: 18504973-18504973 on Build GRCh38
rs762823178 Chr.9: 18504879-18504879 on Build GRCh38
rs548373318 Chr.9: 18504674-18504674 on Build GRCh38
rs959599350 Chr.9: 18504784-18504784 on Build GRCh38
rs200438691 Chr.9: 18504941-18504941 on Build GRCh38
rs532051274 Chr.9: 18505003-18505003 on Build GRCh38
rs867234164 Chr.9: 18504788-18504788 on Build GRCh38
rs763639322 Chr.9: 18504887-18504887 on Build GRCh38
rs200963194 Chr.9: 18504860-18504860 on Build GRCh38
rs772786872 Chr.9: 18504668-18504668 on Build GRCh38
rs751177615 Chr.9: 18504837-18504837 on Build GRCh38
rs774573107 Chr.9: 18504779-18504779 on Build GRCh38
rs752643866 Chr.9: 18504816-18504816 on Build GRCh38
rs551329951 Chr.9: 18504721-18504721 on Build GRCh38
rs370239851 Chr.9: 18504875-18504875 on Build GRCh38
rs763189325 Chr.9: 18504745-18504745 on Build GRCh38
rs760827707 Chr.9: 18504792-18504792 on Build GRCh38
rs758866537 Chr.9: 18504921-18504921 on Build GRCh38
rs368626488 Chr.9: 18504889-18504889 on Build GRCh38
rs146464834 Chr.9: 18505036-18505036 on Build GRCh38
rs746118437 Chr.9: 18504938-18504938 on Build GRCh38
rs61742718 Chr.9: 18504870-18504870 on Build GRCh38
rs373751923 Chr.9: 18504971-18504971 on Build GRCh38
rs939996017 Chr.9: 18505071-18505071 on Build GRCh38
rs777177013 Chr.9: 18504824-18504824 on Build GRCh38
rs758921747 Chr.9: 18504999-18504999 on Build GRCh38
rs566513595 Chr.9: 18504810-18504810 on Build GRCh38
rs746242288 Chr.9: 18504819-18504819 on Build GRCh38
rs776393550 Chr.9: 18504869-18504869 on Build GRCh38
rs779454317 Chr.9: 18504916-18504916 on Build GRCh38
rs4385562 Chr.9: 18504666-18504666 on Build GRCh38
rs762045337 Chr.9: 18504781-18504781 on Build GRCh38
rs898530669 Chr.9: 18505070-18505070 on Build GRCh38
rs763829691 Chr.9: 18504963-18504963 on Build GRCh38
rs143700051 Chr.9: 18504908-18504908 on Build GRCh38
rs542908248 Chr.9: 18504913-18504913 on Build GRCh38

View Full Product Details