Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 86945957-86946505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD4
Entrez Gene ID
8322
Gene Name
frizzled class receptor 4
Gene Aliases
CD344, EVR1, FEVR, FZD4S, Fz-4, Fz4, FzE4, GPCR, hFz4
UniGene
Hs.591968
Gene Chromosome Location
Chr.11: 86945675-86955398 on Build GRCh38
Transcripts
NM_012193.3 NR_120591.1 NR_120592.1

Gene Symbol
PRSS23
Entrez Gene ID
11098
Gene Name
protease, serine 23
Gene Aliases
SIG13, SPUVE, ZSIG13
UniGene
Hs.25338
Gene Chromosome Location
Chr.11: 86791059-86952910 on Build GRCh38
Transcripts
NM_012193.3 NR_120591.1 NR_120592.1

SNP Detail

SNP ID Chromosome Location
rs889556186 Chr.11: 86946437-86946437 on Build GRCh38
rs1027379610 Chr.11: 86946099-86946099 on Build GRCh38
rs1026356311 Chr.11: 86946235-86946235 on Build GRCh38
rs34344036 Chr.11: 86946125-86946126 on Build GRCh38
rs993900226 Chr.11: 86946214-86946214 on Build GRCh38
rs1043840228 Chr.11: 86946059-86946059 on Build GRCh38
rs1049486453 Chr.11: 86946426-86946426 on Build GRCh38
rs371044823 Chr.11: 86945998-86945998 on Build GRCh38
rs1011363406 Chr.11: 86946058-86946058 on Build GRCh38
rs770074267 Chr.11: 86946057-86946057 on Build GRCh38
rs535497508 Chr.11: 86946281-86946281 on Build GRCh38
rs33997579 Chr.11: 86946389-86946389 on Build GRCh38
rs1019136631 Chr.11: 86946090-86946090 on Build GRCh38
rs713065 Chr.11: 86946478-86946478 on Build GRCh38
rs985093081 Chr.11: 86946324-86946324 on Build GRCh38
rs991986308 Chr.11: 86946233-86946233 on Build GRCh38
rs936327623 Chr.11: 86946314-86946314 on Build GRCh38
rs888481629 Chr.11: 86946137-86946137 on Build GRCh38
rs888533600 Chr.11: 86946448-86946448 on Build GRCh38
rs966351583 Chr.11: 86946108-86946108 on Build GRCh38
rs928529353 Chr.11: 86946283-86946283 on Build GRCh38
rs1033558888 Chr.11: 86946190-86946190 on Build GRCh38
rs934302736 Chr.11: 86946002-86946002 on Build GRCh38
rs534530796 Chr.11: 86946042-86946042 on Build GRCh38
rs1048444045 Chr.11: 86946433-86946433 on Build GRCh38
rs1001952438 Chr.11: 86946145-86946145 on Build GRCh38
rs555500541 Chr.11: 86946333-86946333 on Build GRCh38
rs1052816823 Chr.11: 86946008-86946008 on Build GRCh38
rs934629133 Chr.11: 86946285-86946285 on Build GRCh38
rs749654984 Chr.11: 86946275-86946275 on Build GRCh38
rs925037547 Chr.11: 86946272-86946272 on Build GRCh38
rs1041205066 Chr.11: 86945978-86945978 on Build GRCh38
rs143372848 Chr.11: 86946065-86946065 on Build GRCh38
rs537418626 Chr.11: 86946373-86946373 on Build GRCh38
rs952186070 Chr.11: 86946239-86946239 on Build GRCh38
rs796761368 Chr.11: 86946105-86946106 on Build GRCh38
rs184796229 Chr.11: 86946386-86946386 on Build GRCh38
rs61903831 Chr.11: 86946367-86946367 on Build GRCh38
rs574539156 Chr.11: 86946186-86946186 on Build GRCh38
rs28413190 Chr.11: 86946442-86946444 on Build GRCh38
rs959286106 Chr.11: 86946201-86946201 on Build GRCh38
rs886048716 Chr.11: 86946464-86946464 on Build GRCh38
rs72963441 Chr.11: 86946377-86946377 on Build GRCh38
rs915716036 Chr.11: 86946268-86946268 on Build GRCh38
rs34211388 Chr.11: 86946197-86946197 on Build GRCh38
rs1024856164 Chr.11: 86946250-86946250 on Build GRCh38
rs1045404 Chr.11: 86946205-86946205 on Build GRCh38
rs969894925 Chr.11: 86946269-86946269 on Build GRCh38
rs1007741866 Chr.11: 86946072-86946072 on Build GRCh38
rs988879711 Chr.11: 86946340-86946340 on Build GRCh38
rs892869123 Chr.11: 86946025-86946025 on Build GRCh38
rs147967476 Chr.11: 86946259-86946259 on Build GRCh38

View Full Product Details