Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154956663-154957261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PYGO2
Entrez Gene ID
90780
Gene Name
pygopus family PHD finger 2
Gene Aliases
1190004M21Rik
UniGene
Hs.533597
Gene Chromosome Location
Chr.1: 154957026-154961782 on Build GRCh38
Transcripts
NM_138300.3

SNP Detail

SNP ID Chromosome Location
rs1017170233 Chr.1: 154957184-154957184 on Build GRCh38
rs929410986 Chr.1: 154956809-154956809 on Build GRCh38
rs982464992 Chr.1: 154956706-154956706 on Build GRCh38
rs144107372 Chr.1: 154956835-154956835 on Build GRCh38
rs762156276 Chr.1: 154957202-154957202 on Build GRCh38
rs1054616460 Chr.1: 154957201-154957201 on Build GRCh38
rs944710898 Chr.1: 154956848-154956848 on Build GRCh38
rs891991033 Chr.1: 154957226-154957226 on Build GRCh38
rs796208619 Chr.1: 154957073-154957073 on Build GRCh38
rs530950079 Chr.1: 154957238-154957238 on Build GRCh38
rs950513995 Chr.1: 154956985-154956985 on Build GRCh38
rs539054083 Chr.1: 154956926-154956926 on Build GRCh38
rs184642000 Chr.1: 154956715-154956715 on Build GRCh38
rs970853452 Chr.1: 154956918-154956918 on Build GRCh38
rs753360559 Chr.1: 154957114-154957114 on Build GRCh38
rs925802180 Chr.1: 154957123-154957123 on Build GRCh38
rs926572175 Chr.1: 154956708-154956708 on Build GRCh38
rs886748552 Chr.1: 154957182-154957182 on Build GRCh38
rs754731111 Chr.1: 154957120-154957120 on Build GRCh38
rs767633056 Chr.1: 154957236-154957236 on Build GRCh38
rs11542265 Chr.1: 154957090-154957090 on Build GRCh38
rs553275910 Chr.1: 154956826-154956847 on Build GRCh38
rs1043298569 Chr.1: 154956868-154956868 on Build GRCh38
rs891960466 Chr.1: 154956894-154956894 on Build GRCh38
rs764269940 Chr.1: 154956721-154956721 on Build GRCh38
rs895576405 Chr.1: 154957152-154957152 on Build GRCh38
rs936305996 Chr.1: 154957186-154957186 on Build GRCh38
rs945965134 Chr.1: 154957081-154957081 on Build GRCh38
rs867470060 Chr.1: 154956933-154956933 on Build GRCh38
rs1046736061 Chr.1: 154957170-154957170 on Build GRCh38
rs973266592 Chr.1: 154956719-154956719 on Build GRCh38
rs983564251 Chr.1: 154957004-154957004 on Build GRCh38
rs139975787 Chr.1: 154957130-154957130 on Build GRCh38
rs1009293845 Chr.1: 154956896-154956896 on Build GRCh38
rs922628811 Chr.1: 154956813-154956814 on Build GRCh38
rs35981350 Chr.1: 154957127-154957128 on Build GRCh38
rs146019286 Chr.1: 154956978-154956978 on Build GRCh38
rs920395139 Chr.1: 154956795-154956795 on Build GRCh38
rs951581931 Chr.1: 154956745-154956745 on Build GRCh38
rs911993634 Chr.1: 154957125-154957125 on Build GRCh38
rs553197456 Chr.1: 154956842-154956842 on Build GRCh38
rs1054585300 Chr.1: 154956879-154956879 on Build GRCh38
rs375150473 Chr.1: 154956980-154956980 on Build GRCh38
rs988150145 Chr.1: 154957075-154957075 on Build GRCh38
rs900755250 Chr.1: 154956876-154956876 on Build GRCh38
rs578117221 Chr.1: 154956861-154956861 on Build GRCh38
rs8454 Chr.1: 154957128-154957128 on Build GRCh38
rs1051723742 Chr.1: 154956830-154956830 on Build GRCh38
rs904324099 Chr.1: 154956889-154956889 on Build GRCh38
rs975928868 Chr.1: 154957015-154957015 on Build GRCh38
rs984704676 Chr.1: 154956763-154956763 on Build GRCh38
rs1003226963 Chr.1: 154956928-154956928 on Build GRCh38
rs964494495 Chr.1: 154956800-154956800 on Build GRCh38
rs750840214 Chr.1: 154956886-154956886 on Build GRCh38
rs951635369 Chr.1: 154956940-154956940 on Build GRCh38
rs1039417245 Chr.1: 154956857-154956857 on Build GRCh38
rs543000480 Chr.1: 154957121-154957121 on Build GRCh38
rs747983536 Chr.1: 154957183-154957183 on Build GRCh38
rs35323526 Chr.1: 154956690-154956690 on Build GRCh38
rs922681653 Chr.1: 154957061-154957061 on Build GRCh38
rs751968828 Chr.1: 154956743-154956743 on Build GRCh38
rs973310403 Chr.1: 154956966-154956966 on Build GRCh38
rs910013334 Chr.1: 154956766-154956766 on Build GRCh38
rs35122420 Chr.1: 154957181-154957182 on Build GRCh38
rs1020646452 Chr.1: 154956913-154956913 on Build GRCh38
rs964601359 Chr.1: 154957233-154957235 on Build GRCh38
rs921918953 Chr.1: 154957169-154957169 on Build GRCh38
rs970347574 Chr.1: 154956703-154956703 on Build GRCh38
rs1000821281 Chr.1: 154956878-154956878 on Build GRCh38
rs975495249 Chr.1: 154956808-154956809 on Build GRCh38
rs78431420 Chr.1: 154956932-154956932 on Build GRCh38
rs776092573 Chr.1: 154956699-154956699 on Build GRCh38

View Full Product Details