Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40793845-40794434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT28
Entrez Gene ID
162605
Gene Name
keratin 28
Gene Aliases
K25IRS4, KRT25D
UniGene
Hs.59736
Gene Chromosome Location
Chr.17: 40792196-40799959 on Build GRCh38
Transcripts
NM_181535.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
71373 c.1130T>C p.L377P Chr.17: 40793895-40793895 on Build GRCh38
7240914 c.1038G>A p.A346A Chr.17: 40793987-40793987 on Build GRCh38
6234220 c.987C>A p.S329S Chr.17: 40794038-40794038 on Build GRCh38
4952672 c.1177C>T p.L393L Chr.17: 40793848-40793848 on Build GRCh38
6080878 c.1045C>T p.Q349* Chr.17: 40793980-40793980 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765839688 Chr.17: 40793894-40793894 on Build GRCh38
rs565089115 Chr.17: 40794218-40794218 on Build GRCh38
rs142890972 Chr.17: 40794209-40794209 on Build GRCh38
rs776024934 Chr.17: 40793997-40793997 on Build GRCh38
rs779669865 Chr.17: 40794041-40794041 on Build GRCh38
rs761721997 Chr.17: 40794266-40794266 on Build GRCh38
rs781148560 Chr.17: 40794053-40794053 on Build GRCh38
rs1024313900 Chr.17: 40794306-40794306 on Build GRCh38
rs1030866662 Chr.17: 40794352-40794352 on Build GRCh38
rs749708634 Chr.17: 40794095-40794095 on Build GRCh38
rs147900752 Chr.17: 40793977-40793977 on Build GRCh38
rs910439795 Chr.17: 40794371-40794371 on Build GRCh38
rs559066865 Chr.17: 40794249-40794249 on Build GRCh38
rs750179885 Chr.17: 40793942-40793942 on Build GRCh38
rs138281139 Chr.17: 40793996-40793996 on Build GRCh38
rs530530211 Chr.17: 40794373-40794373 on Build GRCh38
rs754506957 Chr.17: 40794051-40794051 on Build GRCh38
rs574817880 Chr.17: 40794165-40794165 on Build GRCh38
rs2250681 Chr.17: 40794362-40794362 on Build GRCh38
rs28441989 Chr.17: 40794200-40794200 on Build GRCh38
rs772074286 Chr.17: 40794288-40794288 on Build GRCh38
rs760770692 Chr.17: 40793878-40793878 on Build GRCh38
rs758500873 Chr.17: 40793883-40793883 on Build GRCh38
rs144664854 Chr.17: 40793932-40793932 on Build GRCh38
rs768613522 Chr.17: 40793969-40793969 on Build GRCh38
rs374895896 Chr.17: 40793930-40793930 on Build GRCh38
rs979892457 Chr.17: 40794120-40794120 on Build GRCh38
rs925783970 Chr.17: 40794122-40794122 on Build GRCh38
rs941711874 Chr.17: 40793947-40793947 on Build GRCh38
rs751455907 Chr.17: 40794025-40794027 on Build GRCh38
rs183399309 Chr.17: 40794157-40794157 on Build GRCh38
rs749156871 Chr.17: 40793968-40793968 on Build GRCh38
rs113890114 Chr.17: 40794244-40794244 on Build GRCh38
rs267604858 Chr.17: 40794006-40794006 on Build GRCh38
rs1052868792 Chr.17: 40794127-40794127 on Build GRCh38
rs150448547 Chr.17: 40793988-40793988 on Build GRCh38
rs917141327 Chr.17: 40794073-40794073 on Build GRCh38
rs746059857 Chr.17: 40794059-40794059 on Build GRCh38
rs1013744583 Chr.17: 40794229-40794229 on Build GRCh38
rs2250671 Chr.17: 40794020-40794020 on Build GRCh38
rs2270154 Chr.17: 40793999-40793999 on Build GRCh38
rs956943653 Chr.17: 40794319-40794319 on Build GRCh38
rs565151324 Chr.17: 40793898-40793898 on Build GRCh38
rs753993009 Chr.17: 40793961-40793961 on Build GRCh38
rs779864735 Chr.17: 40793989-40793989 on Build GRCh38
rs201969522 Chr.17: 40794013-40794013 on Build GRCh38
rs986038628 Chr.17: 40794138-40794138 on Build GRCh38
rs552369769 Chr.17: 40794086-40794086 on Build GRCh38
rs1032322891 Chr.17: 40794298-40794298 on Build GRCh38
rs2270153 Chr.17: 40793987-40793987 on Build GRCh38
rs577163941 Chr.17: 40794228-40794228 on Build GRCh38
rs529718957 Chr.17: 40794282-40794282 on Build GRCh38
rs1014239492 Chr.17: 40794255-40794255 on Build GRCh38
rs763555183 Chr.17: 40793926-40793926 on Build GRCh38
rs750952330 Chr.17: 40794171-40794171 on Build GRCh38
rs985964561 Chr.17: 40794367-40794367 on Build GRCh38
rs771208891 Chr.17: 40793885-40793885 on Build GRCh38
rs186578561 Chr.17: 40794175-40794175 on Build GRCh38
rs376313806 Chr.17: 40794001-40794001 on Build GRCh38
rs769279469 Chr.17: 40794096-40794096 on Build GRCh38
rs755769017 Chr.17: 40794017-40794017 on Build GRCh38
rs746208773 Chr.17: 40793991-40793991 on Build GRCh38
rs1057311964 Chr.17: 40794213-40794213 on Build GRCh38
rs778865281 Chr.17: 40794034-40794034 on Build GRCh38
rs776981920 Chr.17: 40793990-40793990 on Build GRCh38
rs935864507 Chr.17: 40794124-40794124 on Build GRCh38
rs890685920 Chr.17: 40794176-40794176 on Build GRCh38
rs757419781 Chr.17: 40794052-40794052 on Build GRCh38
rs756835671 Chr.17: 40793933-40793933 on Build GRCh38
rs112250021 Chr.17: 40793911-40793911 on Build GRCh38
rs543722149 Chr.17: 40793897-40793897 on Build GRCh38
rs145164915 Chr.17: 40794000-40794000 on Build GRCh38
rs755994795 Chr.17: 40793948-40793948 on Build GRCh38
rs753657695 Chr.17: 40794021-40794021 on Build GRCh38
rs191458561 Chr.17: 40794310-40794310 on Build GRCh38
rs575674025 Chr.17: 40794202-40794202 on Build GRCh38
rs754889550 Chr.17: 40794032-40794032 on Build GRCh38
rs969928104 Chr.17: 40794332-40794332 on Build GRCh38
rs770158838 Chr.17: 40794067-40794067 on Build GRCh38
rs372865484 Chr.17: 40793998-40793998 on Build GRCh38
rs537219094 Chr.17: 40794036-40794036 on Build GRCh38
rs552947424 Chr.17: 40794164-40794164 on Build GRCh38
rs954732672 Chr.17: 40794132-40794132 on Build GRCh38
rs541375276 Chr.17: 40794246-40794246 on Build GRCh38
rs747164914 Chr.17: 40793980-40793980 on Build GRCh38
rs753318223 Chr.17: 40793899-40793899 on Build GRCh38
rs764460897 Chr.17: 40793928-40793928 on Build GRCh38
rs570768466 Chr.17: 40794114-40794114 on Build GRCh38
rs373502722 Chr.17: 40794024-40794024 on Build GRCh38
rs142343265 Chr.17: 40793891-40793891 on Build GRCh38
rs765118221 Chr.17: 40793869-40793870 on Build GRCh38
rs1009642949 Chr.17: 40794325-40794325 on Build GRCh38
rs758432241 Chr.17: 40793964-40793965 on Build GRCh38
rs750033985 Chr.17: 40794014-40794014 on Build GRCh38

View Full Product Details