Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 55267630-55268229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLF
Entrez Gene ID
3131
Gene Name
HLF, PAR bZIP transcription factor
Gene Aliases
-
UniGene
Hs.196952
Gene Chromosome Location
Chr.17: 55264960-55325065 on Build GRCh38
Transcripts
NM_002126.4 XM_005257269.2 XM_011524705.1 XM_017024556.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3889990 c.302C>T p.P101L Chr.17: 55267937-55267937 on Build GRCh38
5190257 c.406C>T p.P136S Chr.17: 55268041-55268041 on Build GRCh38
6680867 c.314A>C p.D105A Chr.17: 55267949-55267949 on Build GRCh38
5078135 c.369C>T p.V123V Chr.17: 55268004-55268004 on Build GRCh38
5045287 c.332C>T p.P111L Chr.17: 55267967-55267967 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201209755 Chr.17: 55267739-55267739 on Build GRCh38
rs774053574 Chr.17: 55267983-55267983 on Build GRCh38
rs778071353 Chr.17: 55267872-55267872 on Build GRCh38
rs564621419 Chr.17: 55267986-55267986 on Build GRCh38
rs769270551 Chr.17: 55267961-55267961 on Build GRCh38
rs945568063 Chr.17: 55267768-55267768 on Build GRCh38
rs183911087 Chr.17: 55267855-55267855 on Build GRCh38
rs777681481 Chr.17: 55267711-55267711 on Build GRCh38
rs755022700 Chr.17: 55267854-55267854 on Build GRCh38
rs779026540 Chr.17: 55267927-55267927 on Build GRCh38
rs948923462 Chr.17: 55268191-55268191 on Build GRCh38
rs760106013 Chr.17: 55267797-55267797 on Build GRCh38
rs747393259 Chr.17: 55267946-55267946 on Build GRCh38
rs749260477 Chr.17: 55267878-55267878 on Build GRCh38
rs760757135 Chr.17: 55267833-55267833 on Build GRCh38
rs974649458 Chr.17: 55268195-55268195 on Build GRCh38
rs760587860 Chr.17: 55268094-55268094 on Build GRCh38
rs916151819 Chr.17: 55267798-55267798 on Build GRCh38
rs780550360 Chr.17: 55268031-55268031 on Build GRCh38
rs925633892 Chr.17: 55267681-55267681 on Build GRCh38
rs181297521 Chr.17: 55267823-55267823 on Build GRCh38
rs751538271 Chr.17: 55267852-55267852 on Build GRCh38
rs964480326 Chr.17: 55268186-55268186 on Build GRCh38
rs772609266 Chr.17: 55267935-55267935 on Build GRCh38
rs145011339 Chr.17: 55268096-55268096 on Build GRCh38
rs767704148 Chr.17: 55267766-55267766 on Build GRCh38
rs968313974 Chr.17: 55268064-55268064 on Build GRCh38
rs146845700 Chr.17: 55267914-55267914 on Build GRCh38
rs770527752 Chr.17: 55268059-55268059 on Build GRCh38
rs757464335 Chr.17: 55268132-55268132 on Build GRCh38
rs775884594 Chr.17: 55267805-55267805 on Build GRCh38
rs773957916 Chr.17: 55268073-55268073 on Build GRCh38
rs766159669 Chr.17: 55267842-55267842 on Build GRCh38
rs906658784 Chr.17: 55267887-55267887 on Build GRCh38
rs772417385 Chr.17: 55267804-55267804 on Build GRCh38
rs752578837 Chr.17: 55267859-55267859 on Build GRCh38
rs767592931 Chr.17: 55267988-55267988 on Build GRCh38
rs771286163 Chr.17: 55267752-55267752 on Build GRCh38
rs977072851 Chr.17: 55268083-55268083 on Build GRCh38
rs775794711 Chr.17: 55267939-55267939 on Build GRCh38
rs577401506 Chr.17: 55267688-55267688 on Build GRCh38
rs540752789 Chr.17: 55267664-55267664 on Build GRCh38
rs761427708 Chr.17: 55267808-55267808 on Build GRCh38
rs1048639653 Chr.17: 55268209-55268209 on Build GRCh38
rs762403571 Chr.17: 55267715-55267716 on Build GRCh38
rs559122453 Chr.17: 55267665-55267665 on Build GRCh38
rs189700197 Chr.17: 55268103-55268103 on Build GRCh38
rs373604103 Chr.17: 55267970-55267970 on Build GRCh38
rs199742593 Chr.17: 55268021-55268021 on Build GRCh38
rs987750619 Chr.17: 55268156-55268156 on Build GRCh38
rs765631500 Chr.17: 55267716-55267720 on Build GRCh38
rs924364210 Chr.17: 55267694-55267694 on Build GRCh38
rs200291249 Chr.17: 55267893-55267893 on Build GRCh38
rs193920793 Chr.17: 55268071-55268072 on Build GRCh38
rs553666718 Chr.17: 55267730-55267730 on Build GRCh38
rs776962498 Chr.17: 55267716-55267716 on Build GRCh38
rs563496609 Chr.17: 55267745-55267745 on Build GRCh38
rs757474218 Chr.17: 55268020-55268020 on Build GRCh38
rs143697299 Chr.17: 55267809-55267809 on Build GRCh38
rs370981067 Chr.17: 55268000-55268000 on Build GRCh38
rs769173900 Chr.17: 55267702-55267706 on Build GRCh38
rs745982634 Chr.17: 55268024-55268024 on Build GRCh38
rs763930784 Chr.17: 55268097-55268097 on Build GRCh38
rs368847041 Chr.17: 55267779-55267779 on Build GRCh38
rs749851297 Chr.17: 55267821-55267821 on Build GRCh38
rs935656428 Chr.17: 55267683-55267683 on Build GRCh38
rs2302739 Chr.17: 55267690-55267690 on Build GRCh38
rs377459438 Chr.17: 55267934-55267934 on Build GRCh38
rs377578509 Chr.17: 55268058-55268058 on Build GRCh38
rs916103150 Chr.17: 55268129-55268129 on Build GRCh38
rs201406160 Chr.17: 55268034-55268034 on Build GRCh38
rs767497126 Chr.17: 55268082-55268082 on Build GRCh38
rs769057460 Chr.17: 55268049-55268049 on Build GRCh38
rs376843983 Chr.17: 55267989-55267989 on Build GRCh38
rs370224993 Chr.17: 55268089-55268089 on Build GRCh38
rs911845030 Chr.17: 55267657-55267657 on Build GRCh38
rs992992194 Chr.17: 55268108-55268108 on Build GRCh38
rs375082103 Chr.17: 55268001-55268001 on Build GRCh38
rs758850881 Chr.17: 55268026-55268026 on Build GRCh38
rs369992114 Chr.17: 55267968-55267968 on Build GRCh38
rs759132306 Chr.17: 55268074-55268074 on Build GRCh38
rs776917251 Chr.17: 55268057-55268057 on Build GRCh38
rs1018736087 Chr.17: 55268159-55268159 on Build GRCh38
rs560740158 Chr.17: 55267929-55267929 on Build GRCh38
rs202162317 Chr.17: 55267866-55267866 on Build GRCh38

View Full Product Details