Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 75372346-75372941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZDHHC15
Entrez Gene ID
158866
Gene Name
zinc finger DHHC-type containing 15
Gene Aliases
MRX91
UniGene
Hs.253211
Gene Chromosome Location
Chr.X: 75368427-75523626 on Build GRCh38
Transcripts
NM_001146256.1 NM_144969.2

SNP Detail

SNP ID Chromosome Location
rs920449099 Chr.X: 75372566-75372566 on Build GRCh38
rs749346642 Chr.X: 75372453-75372453 on Build GRCh38
rs943237894 Chr.X: 75372543-75372543 on Build GRCh38
rs1048309742 Chr.X: 75372626-75372626 on Build GRCh38
rs1042140927 Chr.X: 75372384-75372394 on Build GRCh38
rs55813982 Chr.X: 75372711-75372711 on Build GRCh38
rs981319759 Chr.X: 75372489-75372489 on Build GRCh38
rs948338269 Chr.X: 75372488-75372488 on Build GRCh38
rs1012477949 Chr.X: 75372691-75372691 on Build GRCh38
rs773264119 Chr.X: 75372392-75372392 on Build GRCh38
rs746836838 Chr.X: 75372517-75372517 on Build GRCh38
rs1056364354 Chr.X: 75372641-75372641 on Build GRCh38
rs918123961 Chr.X: 75372487-75372487 on Build GRCh38
rs774897598 Chr.X: 75372881-75372881 on Build GRCh38
rs952917650 Chr.X: 75372890-75372890 on Build GRCh38
rs987116141 Chr.X: 75372519-75372519 on Build GRCh38
rs770841436 Chr.X: 75372631-75372631 on Build GRCh38
rs370022125 Chr.X: 75372734-75372735 on Build GRCh38
rs968656390 Chr.X: 75372727-75372727 on Build GRCh38
rs777827532 Chr.X: 75372507-75372507 on Build GRCh38
rs999697485 Chr.X: 75372727-75372727 on Build GRCh38
rs992420789 Chr.X: 75372475-75372475 on Build GRCh38
rs984505828 Chr.X: 75372911-75372911 on Build GRCh38
rs940403369 Chr.X: 75372534-75372534 on Build GRCh38
rs192397448 Chr.X: 75372617-75372617 on Build GRCh38
rs964354994 Chr.X: 75372920-75372920 on Build GRCh38
rs772155738 Chr.X: 75372462-75372462 on Build GRCh38
rs759971458 Chr.X: 75372648-75372648 on Build GRCh38
rs1048827127 Chr.X: 75372516-75372516 on Build GRCh38
rs756558225 Chr.X: 75372840-75372840 on Build GRCh38
rs955781526 Chr.X: 75372869-75372869 on Build GRCh38
rs769423731 Chr.X: 75372876-75372876 on Build GRCh38
rs766588857 Chr.X: 75372557-75372557 on Build GRCh38
rs34483605 Chr.X: 75372579-75372580 on Build GRCh38
rs1029223838 Chr.X: 75372596-75372596 on Build GRCh38
rs769963907 Chr.X: 75372390-75372390 on Build GRCh38
rs996125573 Chr.X: 75372573-75372573 on Build GRCh38
rs1040046074 Chr.X: 75372535-75372535 on Build GRCh38
rs930478825 Chr.X: 75372583-75372583 on Build GRCh38
rs903716306 Chr.X: 75372666-75372666 on Build GRCh38
rs1000942362 Chr.X: 75372709-75372709 on Build GRCh38
rs930378299 Chr.X: 75372510-75372510 on Build GRCh38
rs1003963786 Chr.X: 75372637-75372637 on Build GRCh38
rs186999491 Chr.X: 75372869-75372869 on Build GRCh38
rs767737069 Chr.X: 75372672-75372672 on Build GRCh38
rs1024988064 Chr.X: 75372807-75372807 on Build GRCh38
rs970022191 Chr.X: 75372838-75372838 on Build GRCh38
rs1013970898 Chr.X: 75372754-75372754 on Build GRCh38
rs920112730 Chr.X: 75372491-75372491 on Build GRCh38
rs1022492074 Chr.X: 75372726-75372727 on Build GRCh38
rs1031841233 Chr.X: 75372854-75372854 on Build GRCh38
rs763224517 Chr.X: 75372503-75372503 on Build GRCh38
rs956778996 Chr.X: 75372728-75372728 on Build GRCh38
rs776407280 Chr.X: 75372669-75372669 on Build GRCh38

View Full Product Details