Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 185658125-185658605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGF2BP2
Entrez Gene ID
10644
Gene Name
insulin like growth factor 2 mRNA binding protein 2
Gene Aliases
IMP-2, IMP2, VICKZ2
UniGene
Hs.35354
Gene Chromosome Location
Chr.3: 185643739-185825051 on Build GRCh38
Transcripts
NM_001007225.1 NM_001291869.1 NM_001291872.1 NM_001291873.1 NM_001291874.1 NM_001291875.1 NM_006548.4 XM_006713477.3 XM_006713478.3 XM_017005557.1 XM_017005558.1 XM_017005559.1 XM_017005560.1 XM_017005561.1 XM_017005562.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3365109 c.1098T>G p.S366S Chr.3: 185658341-185658341 on Build GRCh38
6400054 c.1250C>T p.P417L Chr.3: 185658360-185658360 on Build GRCh38
5037879 c.1058A>C p.Y353S Chr.3: 185658381-185658381 on Build GRCh38
1042256 c.1237C>A p.H413N Chr.3: 185658373-185658373 on Build GRCh38
4444053 c.1058A>G p.Y353C Chr.3: 185658381-185658381 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760250561 Chr.3: 185658291-185658291 on Build GRCh38
rs759966950 Chr.3: 185658572-185658572 on Build GRCh38
rs927290923 Chr.3: 185658172-185658172 on Build GRCh38
rs145173270 Chr.3: 185658376-185658376 on Build GRCh38
rs780570432 Chr.3: 185658403-185658403 on Build GRCh38
rs753597875 Chr.3: 185658301-185658301 on Build GRCh38
rs930401764 Chr.3: 185658235-185658235 on Build GRCh38
rs544990914 Chr.3: 185658252-185658252 on Build GRCh38
rs527522548 Chr.3: 185658304-185658304 on Build GRCh38
rs746446411 Chr.3: 185658323-185658323 on Build GRCh38
rs760184033 Chr.3: 185658336-185658336 on Build GRCh38
rs990200591 Chr.3: 185658513-185658513 on Build GRCh38
rs748257030 Chr.3: 185658300-185658301 on Build GRCh38
rs781334310 Chr.3: 185658300-185658302 on Build GRCh38
rs748379687 Chr.3: 185658191-185658191 on Build GRCh38
rs73058847 Chr.3: 185658165-185658165 on Build GRCh38
rs1024482801 Chr.3: 185658438-185658438 on Build GRCh38
rs917405052 Chr.3: 185658153-185658153 on Build GRCh38
rs757551786 Chr.3: 185658399-185658399 on Build GRCh38
rs758852286 Chr.3: 185658402-185658402 on Build GRCh38
rs375471066 Chr.3: 185658439-185658439 on Build GRCh38
rs762693031 Chr.3: 185658445-185658445 on Build GRCh38
rs779046297 Chr.3: 185658171-185658171 on Build GRCh38
rs746178228 Chr.3: 185658401-185658401 on Build GRCh38
rs370211460 Chr.3: 185658290-185658290 on Build GRCh38
rs529304244 Chr.3: 185658377-185658377 on Build GRCh38
rs946115530 Chr.3: 185658283-185658283 on Build GRCh38
rs756262970 Chr.3: 185658389-185658389 on Build GRCh38
rs747451333 Chr.3: 185658406-185658406 on Build GRCh38
rs775998199 Chr.3: 185658417-185658417 on Build GRCh38
rs912415255 Chr.3: 185658547-185658547 on Build GRCh38
rs892316031 Chr.3: 185658341-185658341 on Build GRCh38
rs778007748 Chr.3: 185658319-185658319 on Build GRCh38
rs763745671 Chr.3: 185658296-185658296 on Build GRCh38
rs552977333 Chr.3: 185658282-185658297 on Build GRCh38
rs778041902 Chr.3: 185658354-185658354 on Build GRCh38
rs957595648 Chr.3: 185658491-185658491 on Build GRCh38
rs960668283 Chr.3: 185658428-185658428 on Build GRCh38
rs772320677 Chr.3: 185658250-185658250 on Build GRCh38
rs1039501564 Chr.3: 185658586-185658586 on Build GRCh38
rs560265674 Chr.3: 185658268-185658268 on Build GRCh38
rs764062602 Chr.3: 185658391-185658391 on Build GRCh38
rs766053290 Chr.3: 185658447-185658447 on Build GRCh38
rs762887896 Chr.3: 185658371-185658371 on Build GRCh38
rs567869182 Chr.3: 185658530-185658530 on Build GRCh38
rs761440241 Chr.3: 185658359-185658359 on Build GRCh38
rs1054844908 Chr.3: 185658215-185658215 on Build GRCh38
rs888795345 Chr.3: 185658552-185658552 on Build GRCh38
rs754175018 Chr.3: 185658398-185658398 on Build GRCh38
rs752747052 Chr.3: 185658383-185658383 on Build GRCh38
rs747801867 Chr.3: 185658425-185658425 on Build GRCh38
rs772922686 Chr.3: 185658365-185658365 on Build GRCh38
rs937056931 Chr.3: 185658386-185658386 on Build GRCh38
rs750417267 Chr.3: 185658310-185658310 on Build GRCh38
rs980367197 Chr.3: 185658443-185658443 on Build GRCh38
rs987261051 Chr.3: 185658540-185658540 on Build GRCh38
rs35311778 Chr.3: 185658484-185658485 on Build GRCh38
rs769986983 Chr.3: 185658439-185658439 on Build GRCh38
rs370761868 Chr.3: 185658451-185658451 on Build GRCh38
rs139539678 Chr.3: 185658566-185658566 on Build GRCh38
rs781134021 Chr.3: 185658317-185658317 on Build GRCh38
rs141112109 Chr.3: 185658360-185658360 on Build GRCh38
rs540094622 Chr.3: 185658539-185658539 on Build GRCh38
rs920101730 Chr.3: 185658529-185658529 on Build GRCh38
rs992359281 Chr.3: 185658168-185658168 on Build GRCh38
rs151111656 Chr.3: 185658353-185658353 on Build GRCh38
rs772642671 Chr.3: 185658325-185658325 on Build GRCh38
rs879258801 Chr.3: 185658190-185658190 on Build GRCh38
rs188457922 Chr.3: 185658315-185658315 on Build GRCh38
rs532135870 Chr.3: 185658400-185658400 on Build GRCh38
rs1040316659 Chr.3: 185658580-185658580 on Build GRCh38
rs971640127 Chr.3: 185658163-185658163 on Build GRCh38
rs769376052 Chr.3: 185658429-185658429 on Build GRCh38
rs371938233 Chr.3: 185658318-185658318 on Build GRCh38
rs199526152 Chr.3: 185658333-185658333 on Build GRCh38
rs768268823 Chr.3: 185658412-185658412 on Build GRCh38
rs761636673 Chr.3: 185658302-185658302 on Build GRCh38
rs749443500 Chr.3: 185658320-185658320 on Build GRCh38
rs529642357 Chr.3: 185658467-185658467 on Build GRCh38
rs1036116694 Chr.3: 185658209-185658209 on Build GRCh38
rs533017388 Chr.3: 185658197-185658197 on Build GRCh38

View Full Product Details