Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 169778812-169779371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYNN
Entrez Gene ID
55892
Gene Name
myoneurin
Gene Aliases
OSZF, SBBIZ1, ZBTB31, ZNF902
UniGene
Hs.507025
Gene Chromosome Location
Chr.3: 169772247-169789716 on Build GRCh38
Transcripts
NM_001185118.1 NM_001185119.1 NM_018657.4 NR_033702.1 NR_033703.1 XM_005247621.4 XM_005247622.3 XM_005247624.3 XM_011512987.1 XM_017006864.1 XM_017006865.1 XM_017006866.1 XM_017006867.1 XM_017006868.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5640921 c.778C>T p.R260C Chr.3: 169779279-169779279 on Build GRCh38
4704752 c.566C>T p.P189L Chr.3: 169779067-169779067 on Build GRCh38
4149655 c.355G>A p.A119T Chr.3: 169778856-169778856 on Build GRCh38
5978744 c.640G>A p.D214N Chr.3: 169779141-169779141 on Build GRCh38
7336665 c.565C>T p.P189S Chr.3: 169779066-169779066 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769722576 Chr.3: 169778890-169778890 on Build GRCh38
rs751743503 Chr.3: 169779210-169779210 on Build GRCh38
rs183188973 Chr.3: 169779126-169779126 on Build GRCh38
rs1007132582 Chr.3: 169779348-169779348 on Build GRCh38
rs750758184 Chr.3: 169779196-169779196 on Build GRCh38
rs191325753 Chr.3: 169779076-169779076 on Build GRCh38
rs1000135495 Chr.3: 169778972-169778972 on Build GRCh38
rs149700988 Chr.3: 169779226-169779226 on Build GRCh38
rs745645728 Chr.3: 169779264-169779264 on Build GRCh38
rs531197853 Chr.3: 169779042-169779044 on Build GRCh38
rs536906238 Chr.3: 169779207-169779207 on Build GRCh38
rs761396977 Chr.3: 169779177-169779177 on Build GRCh38
rs747216437 Chr.3: 169779038-169779038 on Build GRCh38
rs557417257 Chr.3: 169779135-169779135 on Build GRCh38
rs766858445 Chr.3: 169779296-169779296 on Build GRCh38
rs767642313 Chr.3: 169779236-169779236 on Build GRCh38
rs758960260 Chr.3: 169778959-169778959 on Build GRCh38
rs764466451 Chr.3: 169779013-169779013 on Build GRCh38
rs746880942 Chr.3: 169779273-169779273 on Build GRCh38
rs774904137 Chr.3: 169779144-169779144 on Build GRCh38
rs939278547 Chr.3: 169779160-169779160 on Build GRCh38
rs765541791 Chr.3: 169779195-169779195 on Build GRCh38
rs139521740 Chr.3: 169779074-169779074 on Build GRCh38
rs757802722 Chr.3: 169779014-169779014 on Build GRCh38
rs749066571 Chr.3: 169779051-169779051 on Build GRCh38
rs199911803 Chr.3: 169779324-169779324 on Build GRCh38
rs1017546990 Chr.3: 169779212-169779212 on Build GRCh38
rs760353149 Chr.3: 169779068-169779068 on Build GRCh38
rs139065735 Chr.3: 169778889-169778889 on Build GRCh38
rs755282927 Chr.3: 169778959-169778960 on Build GRCh38
rs778714964 Chr.3: 169779127-169779127 on Build GRCh38
rs142875546 Chr.3: 169779012-169779012 on Build GRCh38
rs752787469 Chr.3: 169779321-169779321 on Build GRCh38
rs767270693 Chr.3: 169779001-169779001 on Build GRCh38
rs775006758 Chr.3: 169779301-169779301 on Build GRCh38
rs762160662 Chr.3: 169779079-169779079 on Build GRCh38
rs760157000 Chr.3: 169779147-169779147 on Build GRCh38
rs753168355 Chr.3: 169778843-169778843 on Build GRCh38
rs745904851 Chr.3: 169778876-169778876 on Build GRCh38
rs188625148 Chr.3: 169778861-169778861 on Build GRCh38
rs375743196 Chr.3: 169779082-169779082 on Build GRCh38
rs776068540 Chr.3: 169779278-169779278 on Build GRCh38
rs767730167 Chr.3: 169779316-169779316 on Build GRCh38
rs773957029 Chr.3: 169778946-169778946 on Build GRCh38
rs918711280 Chr.3: 169779057-169779057 on Build GRCh38
rs563594296 Chr.3: 169778926-169778926 on Build GRCh38
rs781546676 Chr.3: 169779109-169779109 on Build GRCh38
rs186909924 Chr.3: 169779337-169779337 on Build GRCh38
rs767183982 Chr.3: 169779039-169779041 on Build GRCh38
rs146178757 Chr.3: 169779239-169779239 on Build GRCh38
rs750582978 Chr.3: 169779020-169779020 on Build GRCh38
rs955619915 Chr.3: 169779334-169779334 on Build GRCh38
rs150524698 Chr.3: 169779055-169779055 on Build GRCh38
rs764285926 Chr.3: 169778931-169778931 on Build GRCh38
rs372726113 Chr.3: 169779031-169779031 on Build GRCh38
rs777747775 Chr.3: 169779046-169779046 on Build GRCh38
rs957917872 Chr.3: 169779249-169779249 on Build GRCh38
rs750243131 Chr.3: 169779338-169779338 on Build GRCh38
rs779765235 Chr.3: 169779344-169779344 on Build GRCh38
rs766384929 Chr.3: 169778945-169778945 on Build GRCh38
rs778047782 Chr.3: 169778849-169778849 on Build GRCh38
rs751880527 Chr.3: 169778872-169778874 on Build GRCh38
rs200829478 Chr.3: 169778882-169778882 on Build GRCh38
rs780026202 Chr.3: 169779096-169779096 on Build GRCh38
rs147441397 Chr.3: 169779019-169779019 on Build GRCh38
rs771498796 Chr.3: 169779066-169779066 on Build GRCh38
rs773090803 Chr.3: 169778892-169778892 on Build GRCh38
rs778996179 Chr.3: 169778862-169778862 on Build GRCh38
rs758656401 Chr.3: 169778838-169778838 on Build GRCh38
rs763013313 Chr.3: 169778944-169778944 on Build GRCh38
rs780029603 Chr.3: 169779267-169779267 on Build GRCh38
rs759454956 Chr.3: 169779056-169779056 on Build GRCh38
rs750905911 Chr.3: 169779084-169779084 on Build GRCh38
rs768355559 Chr.3: 169779155-169779155 on Build GRCh38
rs1007554625 Chr.3: 169779165-169779165 on Build GRCh38
rs763705502 Chr.3: 169779073-169779073 on Build GRCh38
rs373647274 Chr.3: 169779308-169779308 on Build GRCh38
rs775019710 Chr.3: 169779067-169779067 on Build GRCh38
rs755296438 Chr.3: 169779101-169779101 on Build GRCh38
rs763359355 Chr.3: 169779286-169779286 on Build GRCh38
rs377574590 Chr.3: 169779315-169779315 on Build GRCh38
rs769247598 Chr.3: 169779190-169779192 on Build GRCh38
rs370494498 Chr.3: 169779280-169779280 on Build GRCh38
rs772396326 Chr.3: 169778879-169778879 on Build GRCh38
rs1030832101 Chr.3: 169779058-169779058 on Build GRCh38
rs752299433 Chr.3: 169779009-169779009 on Build GRCh38
rs777264035 Chr.3: 169779320-169779321 on Build GRCh38
rs780122785 Chr.3: 169779037-169779037 on Build GRCh38
rs61745452 Chr.3: 169779257-169779257 on Build GRCh38
rs763407151 Chr.3: 169779206-169779206 on Build GRCh38
rs758316953 Chr.3: 169779265-169779265 on Build GRCh38
rs756358574 Chr.3: 169779250-169779250 on Build GRCh38
rs751623996 Chr.3: 169779097-169779097 on Build GRCh38
rs148032854 Chr.3: 169779029-169779029 on Build GRCh38
rs758209090 Chr.3: 169779339-169779339 on Build GRCh38
rs201860864 Chr.3: 169779274-169779274 on Build GRCh38
rs533705179 Chr.3: 169778934-169778934 on Build GRCh38
rs35294030 Chr.3: 169779040-169779040 on Build GRCh38
rs199670531 Chr.3: 169779095-169779095 on Build GRCh38
rs756433548 Chr.3: 169778844-169778844 on Build GRCh38
rs771045925 Chr.3: 169779052-169779052 on Build GRCh38
rs576001456 Chr.3: 169779141-169779141 on Build GRCh38
rs776269893 Chr.3: 169779176-169779176 on Build GRCh38
rs747828571 Chr.3: 169779279-169779279 on Build GRCh38

View Full Product Details