Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 108208342-108208914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIG4
Entrez Gene ID
3981
Gene Name
DNA ligase 4
Gene Aliases
LIG4S
UniGene
Hs.166091
Gene Chromosome Location
Chr.13: 108207442-108218368 on Build GRCh38
Transcripts
NM_001098268.1 NM_002312.3 NM_206937.1 XM_005254056.1 XM_005254057.4 XM_005254058.3 XM_006719951.3 XM_006719952.1 XM_011521091.2 XM_011521092.2 XM_017020563.1 XM_017020564.1 XM_017020565.1 XM_017020566.1 XM_017020567.1 XM_017020568.1 XM_017020569.1 XM_017020570.1 XM_017020571.1 XM_017020572.1 XM_017020573.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4570358 c.2634T>A p.F878L Chr.13: 108208635-108208635 on Build GRCh38
2262527 c.2561C>A p.S854Y Chr.13: 108208708-108208708 on Build GRCh38
5500571 c.2624T>C p.F875S Chr.13: 108208645-108208645 on Build GRCh38
200663 c.2443C>T p.R815C Chr.13: 108208826-108208826 on Build GRCh38
4570172 c.2410T>G p.S804A Chr.13: 108208859-108208859 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752539259 Chr.13: 108208803-108208803 on Build GRCh38
rs950925996 Chr.13: 108208577-108208577 on Build GRCh38
rs753650304 Chr.13: 108208628-108208630 on Build GRCh38
rs774731069 Chr.13: 108208556-108208556 on Build GRCh38
rs368132126 Chr.13: 108208571-108208571 on Build GRCh38
rs750080610 Chr.13: 108208579-108208579 on Build GRCh38
rs758064728 Chr.13: 108208666-108208666 on Build GRCh38
rs200695712 Chr.13: 108208505-108208505 on Build GRCh38
rs757299963 Chr.13: 108208818-108208818 on Build GRCh38
rs3093767 Chr.13: 108208404-108208404 on Build GRCh38
rs386774524 Chr.13: 108208402-108208404 on Build GRCh38
rs879062972 Chr.13: 108208749-108208749 on Build GRCh38
rs903162818 Chr.13: 108208882-108208882 on Build GRCh38
rs551263548 Chr.13: 108208530-108208530 on Build GRCh38
rs146597721 Chr.13: 108208783-108208783 on Build GRCh38
rs191975795 Chr.13: 108208846-108208846 on Build GRCh38
rs3093768 Chr.13: 108208402-108208402 on Build GRCh38
rs151271748 Chr.13: 108208614-108208614 on Build GRCh38
rs762458069 Chr.13: 108208833-108208833 on Build GRCh38
rs767996539 Chr.13: 108208636-108208636 on Build GRCh38
rs371586720 Chr.13: 108208656-108208656 on Build GRCh38
rs779925057 Chr.13: 108208586-108208586 on Build GRCh38
rs746144616 Chr.13: 108208742-108208742 on Build GRCh38
rs759806691 Chr.13: 108208633-108208633 on Build GRCh38
rs959885828 Chr.13: 108208498-108208498 on Build GRCh38
rs549559633 Chr.13: 108208713-108208713 on Build GRCh38
rs201115578 Chr.13: 108208683-108208683 on Build GRCh38
rs751149500 Chr.13: 108208801-108208801 on Build GRCh38
rs765317127 Chr.13: 108208674-108208677 on Build GRCh38
rs373272193 Chr.13: 108208879-108208879 on Build GRCh38
rs749470898 Chr.13: 108208626-108208626 on Build GRCh38
rs376493360 Chr.13: 108208809-108208809 on Build GRCh38
rs368576780 Chr.13: 108208821-108208821 on Build GRCh38
rs753757622 Chr.13: 108208848-108208848 on Build GRCh38
rs759042142 Chr.13: 108208839-108208839 on Build GRCh38
rs755419375 Chr.13: 108208824-108208824 on Build GRCh38
rs574090677 Chr.13: 108208825-108208825 on Build GRCh38
rs137899041 Chr.13: 108208844-108208844 on Build GRCh38
rs886049947 Chr.13: 108208477-108208477 on Build GRCh38
rs767764977 Chr.13: 108208458-108208458 on Build GRCh38
rs201176444 Chr.13: 108208685-108208685 on Build GRCh38
rs188422094 Chr.13: 108208720-108208720 on Build GRCh38
rs199526538 Chr.13: 108208777-108208777 on Build GRCh38
rs757245734 Chr.13: 108208852-108208852 on Build GRCh38
rs104894419 Chr.13: 108208829-108208829 on Build GRCh38
rs749954101 Chr.13: 108208659-108208659 on Build GRCh38
rs2232644 Chr.13: 108208429-108208429 on Build GRCh38
rs72660870 Chr.13: 108208744-108208744 on Build GRCh38
rs200369995 Chr.13: 108208751-108208751 on Build GRCh38
rs760459358 Chr.13: 108208845-108208845 on Build GRCh38
rs974517230 Chr.13: 108208732-108208732 on Build GRCh38
rs983917564 Chr.13: 108208589-108208589 on Build GRCh38
rs757090161 Chr.13: 108208674-108208674 on Build GRCh38
rs778998772 Chr.13: 108208733-108208733 on Build GRCh38
rs138936350 Chr.13: 108208880-108208880 on Build GRCh38
rs769297575 Chr.13: 108208770-108208770 on Build GRCh38
rs766112279 Chr.13: 108208674-108208674 on Build GRCh38
rs141441003 Chr.13: 108208804-108208804 on Build GRCh38
rs2232643 Chr.13: 108208479-108208479 on Build GRCh38
rs761227485 Chr.13: 108208762-108208762 on Build GRCh38
rs750374893 Chr.13: 108208788-108208790 on Build GRCh38
rs766679114 Chr.13: 108208533-108208533 on Build GRCh38
rs578115401 Chr.13: 108208406-108208406 on Build GRCh38
rs748716395 Chr.13: 108208834-108208834 on Build GRCh38
rs142374894 Chr.13: 108208612-108208612 on Build GRCh38
rs148711190 Chr.13: 108208607-108208607 on Build GRCh38
rs553852969 Chr.13: 108208440-108208440 on Build GRCh38
rs765951039 Chr.13: 108208784-108208784 on Build GRCh38
rs758148987 Chr.13: 108208580-108208580 on Build GRCh38
rs538355612 Chr.13: 108208367-108208367 on Build GRCh38
rs1034008680 Chr.13: 108208489-108208489 on Build GRCh38
rs763975869 Chr.13: 108208589-108208589 on Build GRCh38
rs375272861 Chr.13: 108208568-108208568 on Build GRCh38
rs183928755 Chr.13: 108208657-108208657 on Build GRCh38
rs747270261 Chr.13: 108208876-108208876 on Build GRCh38
rs771056667 Chr.13: 108208723-108208723 on Build GRCh38
rs758462531 Chr.13: 108208868-108208868 on Build GRCh38
rs940934198 Chr.13: 108208427-108208427 on Build GRCh38
rs199988074 Chr.13: 108208529-108208529 on Build GRCh38
rs369320737 Chr.13: 108208499-108208499 on Build GRCh38
rs907521049 Chr.13: 108208799-108208799 on Build GRCh38
rs764883876 Chr.13: 108208573-108208573 on Build GRCh38
rs201900772 Chr.13: 108208628-108208628 on Build GRCh38
rs545309994 Chr.13: 108208865-108208865 on Build GRCh38
rs767213565 Chr.13: 108208841-108208841 on Build GRCh38
rs906839898 Chr.13: 108208477-108208477 on Build GRCh38
rs1045424544 Chr.13: 108208758-108208758 on Build GRCh38
rs930322513 Chr.13: 108208377-108208377 on Build GRCh38
rs781089675 Chr.13: 108208693-108208693 on Build GRCh38
rs772551854 Chr.13: 108208864-108208864 on Build GRCh38
rs2232642 Chr.13: 108208700-108208700 on Build GRCh38
rs756076064 Chr.13: 108208708-108208708 on Build GRCh38
rs773940828 Chr.13: 108208835-108208835 on Build GRCh38
rs779103198 Chr.13: 108208820-108208820 on Build GRCh38
rs771148408 Chr.13: 108208627-108208627 on Build GRCh38
rs773391515 Chr.13: 108208507-108208507 on Build GRCh38
rs748160595 Chr.13: 108208503-108208503 on Build GRCh38
rs773300788 Chr.13: 108208624-108208624 on Build GRCh38
rs775834343 Chr.13: 108208637-108208637 on Build GRCh38
rs573957541 Chr.13: 108208443-108208443 on Build GRCh38
rs886049948 Chr.13: 108208507-108208507 on Build GRCh38
rs375437614 Chr.13: 108208826-108208826 on Build GRCh38
rs376657998 Chr.13: 108208802-108208802 on Build GRCh38
rs1028222636 Chr.13: 108208522-108208522 on Build GRCh38
rs149457381 Chr.13: 108208870-108208870 on Build GRCh38
rs534543979 Chr.13: 108208817-108208817 on Build GRCh38
rs780983005 Chr.13: 108208610-108208610 on Build GRCh38

View Full Product Details