Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 44767607-44768062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM69
Entrez Gene ID
140691
Gene Name
tripartite motif containing 69
Gene Aliases
HSD-34, HSD34, RNF36, Trif
UniGene
Hs.745248
Gene Chromosome Location
Chr.15: 44736362-44767829 on Build GRCh38
Transcripts
NM_001301144.1 NM_001301145.1 NM_001301146.1 NM_080745.4 NM_182985.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3816184 c.1422G>A p.E474E Chr.15: 44767691-44767691 on Build GRCh38
2189188 c.1348G>T p.E450* Chr.15: 44767617-44767617 on Build GRCh38
4055108 c.1370A>G p.Y457C Chr.15: 44767639-44767639 on Build GRCh38
5540828 c.1444C>T p.P482S Chr.15: 44767713-44767713 on Build GRCh38
6077459 c.1499A>T p.Q500L Chr.15: 44767768-44767768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs955096301 Chr.15: 44767954-44767954 on Build GRCh38
rs759474007 Chr.15: 44767790-44767795 on Build GRCh38
rs1044290530 Chr.15: 44767974-44767974 on Build GRCh38
rs755592624 Chr.15: 44767774-44767774 on Build GRCh38
rs758741468 Chr.15: 44767801-44767801 on Build GRCh38
rs149919213 Chr.15: 44767655-44767655 on Build GRCh38
rs749736724 Chr.15: 44767678-44767678 on Build GRCh38
rs759270933 Chr.15: 44767748-44767748 on Build GRCh38
rs752115228 Chr.15: 44767662-44767662 on Build GRCh38
rs914503404 Chr.15: 44767940-44767940 on Build GRCh38
rs3100143 Chr.15: 44767816-44767816 on Build GRCh38
rs985109266 Chr.15: 44767996-44767996 on Build GRCh38
rs763528011 Chr.15: 44767775-44767775 on Build GRCh38
rs907223951 Chr.15: 44767702-44767702 on Build GRCh38
rs569330126 Chr.15: 44767846-44767846 on Build GRCh38
rs773291642 Chr.15: 44767729-44767729 on Build GRCh38
rs772155024 Chr.15: 44767711-44767711 on Build GRCh38
rs905905780 Chr.15: 44768013-44768013 on Build GRCh38
rs766338082 Chr.15: 44767743-44767743 on Build GRCh38
rs371429122 Chr.15: 44767780-44767780 on Build GRCh38
rs778270656 Chr.15: 44767676-44767676 on Build GRCh38
rs775355668 Chr.15: 44767896-44767896 on Build GRCh38
rs867407692 Chr.15: 44767765-44767765 on Build GRCh38
rs542375630 Chr.15: 44767891-44767891 on Build GRCh38
rs959281242 Chr.15: 44767840-44767840 on Build GRCh38
rs746251756 Chr.15: 44767796-44767796 on Build GRCh38
rs367855776 Chr.15: 44767746-44767746 on Build GRCh38
rs751326023 Chr.15: 44767634-44767634 on Build GRCh38
rs770817598 Chr.15: 44767800-44767800 on Build GRCh38
rs111830928 Chr.15: 44767950-44767950 on Build GRCh38
rs775741901 Chr.15: 44767712-44767712 on Build GRCh38
rs964433951 Chr.15: 44767855-44767855 on Build GRCh38
rs201411863 Chr.15: 44767673-44767673 on Build GRCh38
rs768802917 Chr.15: 44767714-44767714 on Build GRCh38
rs757620967 Chr.15: 44767684-44767684 on Build GRCh38
rs910848067 Chr.15: 44768022-44768022 on Build GRCh38
rs767656901 Chr.15: 44767797-44767798 on Build GRCh38
rs779291067 Chr.15: 44767794-44767794 on Build GRCh38
rs543539926 Chr.15: 44767838-44767838 on Build GRCh38
rs760183300 Chr.15: 44767935-44767935 on Build GRCh38
rs748251986 Chr.15: 44767651-44767651 on Build GRCh38
rs527963489 Chr.15: 44767998-44767998 on Build GRCh38
rs975687231 Chr.15: 44767856-44767856 on Build GRCh38
rs779243821 Chr.15: 44767693-44767693 on Build GRCh38
rs780154100 Chr.15: 44767806-44767806 on Build GRCh38
rs922976460 Chr.15: 44767864-44767864 on Build GRCh38
rs3100144 Chr.15: 44767853-44767853 on Build GRCh38
rs767103515 Chr.15: 44767761-44767761 on Build GRCh38
rs374976088 Chr.15: 44767892-44767892 on Build GRCh38
rs773206257 Chr.15: 44767763-44767763 on Build GRCh38
rs905980772 Chr.15: 44767744-44767744 on Build GRCh38
rs762982714 Chr.15: 44767733-44767733 on Build GRCh38
rs376529781 Chr.15: 44767782-44767782 on Build GRCh38
rs747081759 Chr.15: 44767713-44767713 on Build GRCh38
rs1011439676 Chr.15: 44767648-44767648 on Build GRCh38
rs994440755 Chr.15: 44767787-44767787 on Build GRCh38
rs754577212 Chr.15: 44767636-44767636 on Build GRCh38
rs377079867 Chr.15: 44767694-44767694 on Build GRCh38

View Full Product Details