Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 108019418-108019925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HADH
Entrez Gene ID
3033
Gene Name
hydroxyacyl-CoA dehydrogenase
Gene Aliases
HAD, HADH1, HADHSC, HCDH, HHF4, MSCHAD, SCHAD
UniGene
Hs.438289
Gene Chromosome Location
Chr.4: 107989714-108035175 on Build GRCh38
Transcripts
NM_001184705.2 NM_005327.4 XM_005262972.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1049884 c.464G>T p.S155I Chr.4: 108019584-108019584 on Build GRCh38
6836385 c.485G>T p.R162I Chr.4: 108019605-108019605 on Build GRCh38
5865551 c.505C>T p.L169F Chr.4: 108019625-108019625 on Build GRCh38
4942530 c.520C>T p.P174S Chr.4: 108019640-108019640 on Build GRCh38
5902694 c.449C>T p.S150F Chr.4: 108019569-108019569 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146295174 Chr.4: 108019882-108019882 on Build GRCh38
rs1051519 Chr.4: 108019576-108019576 on Build GRCh38
rs758432207 Chr.4: 108019596-108019596 on Build GRCh38
rs763529297 Chr.4: 108019523-108019523 on Build GRCh38
rs957554325 Chr.4: 108019848-108019848 on Build GRCh38
rs963292997 Chr.4: 108019805-108019805 on Build GRCh38
rs1000578549 Chr.4: 108019800-108019800 on Build GRCh38
rs1020618142 Chr.4: 108019786-108019786 on Build GRCh38
rs377521547 Chr.4: 108019606-108019606 on Build GRCh38
rs996656729 Chr.4: 108019726-108019726 on Build GRCh38
rs916181826 Chr.4: 108019852-108019852 on Build GRCh38
rs531255799 Chr.4: 108019542-108019542 on Build GRCh38
rs1045723176 Chr.4: 108019653-108019653 on Build GRCh38
rs769995771 Chr.4: 108019636-108019636 on Build GRCh38
rs1040320480 Chr.4: 108019732-108019732 on Build GRCh38
rs780252799 Chr.4: 108019599-108019599 on Build GRCh38
rs753508883 Chr.4: 108019560-108019560 on Build GRCh38
rs756959997 Chr.4: 108019574-108019574 on Build GRCh38
rs908548524 Chr.4: 108019906-108019906 on Build GRCh38
rs375358095 Chr.4: 108019766-108019766 on Build GRCh38
rs763621744 Chr.4: 108019675-108019675 on Build GRCh38
rs969590351 Chr.4: 108019788-108019790 on Build GRCh38
rs753418769 Chr.4: 108019679-108019679 on Build GRCh38
rs779675998 Chr.4: 108019754-108019754 on Build GRCh38
rs371364058 Chr.4: 108019704-108019704 on Build GRCh38
rs761537007 Chr.4: 108019683-108019683 on Build GRCh38
rs904142374 Chr.4: 108019736-108019736 on Build GRCh38
rs980670942 Chr.4: 108019788-108019788 on Build GRCh38
rs763364092 Chr.4: 108019652-108019652 on Build GRCh38
rs557161860 Chr.4: 108019521-108019521 on Build GRCh38
rs1032023328 Chr.4: 108019833-108019833 on Build GRCh38
rs770121338 Chr.4: 108019718-108019718 on Build GRCh38
rs781436064 Chr.4: 108019708-108019708 on Build GRCh38
rs770971896 Chr.4: 108019565-108019567 on Build GRCh38
rs751580078 Chr.4: 108019700-108019700 on Build GRCh38
rs768880930 Chr.4: 108019614-108019614 on Build GRCh38
rs542779365 Chr.4: 108019646-108019646 on Build GRCh38
rs141985921 Chr.4: 108019851-108019851 on Build GRCh38
rs761678277 Chr.4: 108019618-108019618 on Build GRCh38
rs768148253 Chr.4: 108019818-108019818 on Build GRCh38
rs778561311 Chr.4: 108019638-108019639 on Build GRCh38
rs140032535 Chr.4: 108019844-108019844 on Build GRCh38
rs572909989 Chr.4: 108019794-108019794 on Build GRCh38
rs1038367812 Chr.4: 108019562-108019562 on Build GRCh38
rs575507968 Chr.4: 108019501-108019501 on Build GRCh38
rs530999953 Chr.4: 108019862-108019862 on Build GRCh38
rs763783107 Chr.4: 108019553-108019553 on Build GRCh38
rs34086962 Chr.4: 108019666-108019667 on Build GRCh38
rs750275531 Chr.4: 108019691-108019691 on Build GRCh38
rs184566960 Chr.4: 108019473-108019473 on Build GRCh38
rs570860691 Chr.4: 108019774-108019774 on Build GRCh38
rs370306695 Chr.4: 108019619-108019619 on Build GRCh38
rs957501981 Chr.4: 108019519-108019519 on Build GRCh38
rs1013314403 Chr.4: 108019782-108019782 on Build GRCh38
rs912650735 Chr.4: 108019895-108019895 on Build GRCh38
rs1049695404 Chr.4: 108019746-108019746 on Build GRCh38
rs764941431 Chr.4: 108019687-108019687 on Build GRCh38
rs374179494 Chr.4: 108019664-108019664 on Build GRCh38
rs765666675 Chr.4: 108019503-108019503 on Build GRCh38
rs745784046 Chr.4: 108019580-108019580 on Build GRCh38
rs538085330 Chr.4: 108019502-108019502 on Build GRCh38
rs1007066467 Chr.4: 108019717-108019717 on Build GRCh38
rs982061128 Chr.4: 108019888-108019888 on Build GRCh38
rs559290031 Chr.4: 108019765-108019765 on Build GRCh38
rs916141493 Chr.4: 108019545-108019545 on Build GRCh38
rs774956302 Chr.4: 108019662-108019662 on Build GRCh38
rs377007645 Chr.4: 108019698-108019698 on Build GRCh38
rs755730411 Chr.4: 108019547-108019547 on Build GRCh38
rs766947866 Chr.4: 108019660-108019660 on Build GRCh38
rs1051581222 Chr.4: 108019893-108019893 on Build GRCh38
rs780093229 Chr.4: 108019699-108019699 on Build GRCh38
rs991715012 Chr.4: 108019525-108019525 on Build GRCh38

View Full Product Details