Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57546889-57547425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF550
Entrez Gene ID
162972
Gene Name
zinc finger protein 550
Gene Aliases
-
UniGene
Hs.180257
Gene Chromosome Location
Chr.19: 57541836-57561512 on Build GRCh38
Transcripts
NM_001277090.1 NM_001277091.1 NM_001277092.1 NM_001277093.1 XM_011526567.2 XM_011526568.2 XM_017026401.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1681433 c.839A>G p.E280G Chr.19: 57547282-57547282 on Build GRCh38
7122806 c.903C>A p.P301P Chr.19: 57547218-57547218 on Build GRCh38
4744215 c.996A>C p.Q332H Chr.19: 57547248-57547248 on Build GRCh38
6831156 c.1141A>G p.T381A Chr.19: 57546980-57546980 on Build GRCh38
5707239 c.1052C>G p.A351G Chr.19: 57547192-57547192 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755474829 Chr.19: 57547051-57547051 on Build GRCh38
rs767095709 Chr.19: 57547038-57547038 on Build GRCh38
rs760625496 Chr.19: 57547027-57547027 on Build GRCh38
rs150707350 Chr.19: 57547207-57547207 on Build GRCh38
rs374457112 Chr.19: 57546981-57546981 on Build GRCh38
rs866650166 Chr.19: 57547175-57547175 on Build GRCh38
rs762522853 Chr.19: 57547356-57547357 on Build GRCh38
rs866097158 Chr.19: 57547320-57547330 on Build GRCh38
rs144170900 Chr.19: 57547093-57547093 on Build GRCh38
rs142851113 Chr.19: 57546983-57546983 on Build GRCh38
rs771542995 Chr.19: 57547166-57547166 on Build GRCh38
rs772428586 Chr.19: 57547058-57547058 on Build GRCh38
rs752435329 Chr.19: 57547333-57547333 on Build GRCh38
rs778139114 Chr.19: 57547063-57547063 on Build GRCh38
rs767686192 Chr.19: 57547321-57547321 on Build GRCh38
rs755457362 Chr.19: 57547120-57547120 on Build GRCh38
rs776761844 Chr.19: 57547072-57547073 on Build GRCh38
rs770254636 Chr.19: 57547276-57547276 on Build GRCh38
rs770556618 Chr.19: 57547261-57547261 on Build GRCh38
rs562411626 Chr.19: 57547092-57547092 on Build GRCh38
rs751900658 Chr.19: 57547361-57547362 on Build GRCh38
rs376718380 Chr.19: 57546967-57546967 on Build GRCh38
rs775955668 Chr.19: 57547277-57547277 on Build GRCh38
rs770454593 Chr.19: 57547347-57547347 on Build GRCh38
rs781567767 Chr.19: 57546941-57546941 on Build GRCh38
rs768600052 Chr.19: 57546969-57546969 on Build GRCh38
rs556524699 Chr.19: 57546993-57546993 on Build GRCh38
rs750456161 Chr.19: 57547400-57547400 on Build GRCh38
rs200035742 Chr.19: 57547245-57547245 on Build GRCh38
rs868615118 Chr.19: 57547274-57547275 on Build GRCh38
rs751029825 Chr.19: 57547016-57547016 on Build GRCh38
rs757458547 Chr.19: 57547148-57547148 on Build GRCh38
rs768582640 Chr.19: 57547157-57547157 on Build GRCh38
rs752559788 Chr.19: 57547404-57547404 on Build GRCh38
rs754576099 Chr.19: 57547099-57547099 on Build GRCh38
rs761844530 Chr.19: 57547380-57547380 on Build GRCh38
rs762571206 Chr.19: 57547008-57547008 on Build GRCh38
rs529088716 Chr.19: 57547332-57547332 on Build GRCh38
rs778192065 Chr.19: 57547146-57547146 on Build GRCh38
rs373679009 Chr.19: 57547323-57547323 on Build GRCh38
rs902170635 Chr.19: 57547074-57547075 on Build GRCh38
rs188119870 Chr.19: 57546956-57546956 on Build GRCh38
rs769520622 Chr.19: 57547358-57547358 on Build GRCh38
rs879081860 Chr.19: 57546982-57546982 on Build GRCh38
rs764904302 Chr.19: 57547276-57547292 on Build GRCh38
rs769839285 Chr.19: 57546999-57546999 on Build GRCh38
rs200419118 Chr.19: 57547079-57547079 on Build GRCh38
rs201235437 Chr.19: 57547118-57547118 on Build GRCh38
rs756427224 Chr.19: 57547041-57547041 on Build GRCh38
rs750966283 Chr.19: 57547399-57547400 on Build GRCh38
rs867721180 Chr.19: 57547167-57547167 on Build GRCh38
rs779343527 Chr.19: 57547239-57547239 on Build GRCh38
rs183472177 Chr.19: 57547402-57547402 on Build GRCh38
rs111521556 Chr.19: 57546961-57546961 on Build GRCh38
rs560070583 Chr.19: 57547267-57547267 on Build GRCh38
rs771951659 Chr.19: 57547242-57547242 on Build GRCh38
rs909911452 Chr.19: 57547360-57547360 on Build GRCh38
rs750788469 Chr.19: 57547040-57547040 on Build GRCh38
rs1019434721 Chr.19: 57547134-57547134 on Build GRCh38
rs370529019 Chr.19: 57547259-57547259 on Build GRCh38
rs777761686 Chr.19: 57547335-57547335 on Build GRCh38
rs369718343 Chr.19: 57546987-57546987 on Build GRCh38
rs147432413 Chr.19: 57547064-57547064 on Build GRCh38
rs763478648 Chr.19: 57547011-57547011 on Build GRCh38
rs756174441 Chr.19: 57547403-57547404 on Build GRCh38
rs1006637953 Chr.19: 57547345-57547345 on Build GRCh38
rs532893630 Chr.19: 57547202-57547202 on Build GRCh38
rs144066628 Chr.19: 57547349-57547349 on Build GRCh38
rs747167283 Chr.19: 57547147-57547147 on Build GRCh38
rs768531686 Chr.19: 57547088-57547088 on Build GRCh38
rs757732819 Chr.19: 57547371-57547372 on Build GRCh38
rs754593404 Chr.19: 57547172-57547172 on Build GRCh38
rs776010564 Chr.19: 57547366-57547366 on Build GRCh38
rs760029323 Chr.19: 57547272-57547272 on Build GRCh38
rs761476303 Chr.19: 57547037-57547037 on Build GRCh38
rs771145259 Chr.19: 57547230-57547230 on Build GRCh38
rs138019714 Chr.19: 57547193-57547193 on Build GRCh38
rs139099223 Chr.19: 57547318-57547318 on Build GRCh38
rs780662580 Chr.19: 57547047-57547047 on Build GRCh38
rs752380837 Chr.19: 57547341-57547343 on Build GRCh38
rs774905364 Chr.19: 57547377-57547377 on Build GRCh38
rs766687703 Chr.19: 57547100-57547100 on Build GRCh38
rs780122294 Chr.19: 57547404-57547405 on Build GRCh38
rs965289129 Chr.19: 57546919-57546919 on Build GRCh38
rs984048632 Chr.19: 57547373-57547373 on Build GRCh38
rs10410631 Chr.19: 57547371-57547371 on Build GRCh38
rs141600675 Chr.19: 57546974-57546974 on Build GRCh38
rs779735903 Chr.19: 57547149-57547149 on Build GRCh38
rs370391615 Chr.19: 57547406-57547406 on Build GRCh38
rs770799650 Chr.19: 57547065-57547066 on Build GRCh38
rs191613619 Chr.19: 57547197-57547197 on Build GRCh38
rs751092867 Chr.19: 57547096-57547096 on Build GRCh38
rs779160874 Chr.19: 57547053-57547053 on Build GRCh38
rs766525533 Chr.19: 57547403-57547403 on Build GRCh38
rs749837485 Chr.19: 57547177-57547177 on Build GRCh38
rs774263656 Chr.19: 57547160-57547160 on Build GRCh38
rs751609264 Chr.19: 57547216-57547216 on Build GRCh38
rs938097899 Chr.19: 57546975-57546975 on Build GRCh38
rs762829560 Chr.19: 57547080-57547080 on Build GRCh38
rs778705753 Chr.19: 57547330-57547330 on Build GRCh38
rs751595570 Chr.19: 57547310-57547310 on Build GRCh38
rs747711957 Chr.19: 57547165-57547165 on Build GRCh38
rs781405675 Chr.19: 57547219-57547219 on Build GRCh38
rs563579417 Chr.19: 57547367-57547367 on Build GRCh38
rs377559464 Chr.19: 57547156-57547156 on Build GRCh38
rs868732627 Chr.19: 57547186-57547186 on Build GRCh38
rs766002386 Chr.19: 57547191-57547191 on Build GRCh38
rs373511509 Chr.19: 57547089-57547089 on Build GRCh38
rs767601529 Chr.19: 57547387-57547387 on Build GRCh38
rs765774236 Chr.19: 57547121-57547121 on Build GRCh38
rs764448797 Chr.19: 57547359-57547360 on Build GRCh38
rs754752444 Chr.19: 57547222-57547222 on Build GRCh38
rs1007058681 Chr.19: 57547132-57547132 on Build GRCh38
rs781527434 Chr.19: 57547374-57547377 on Build GRCh38
rs534898048 Chr.19: 57547372-57547372 on Build GRCh38
rs1042631613 Chr.19: 57547241-57547241 on Build GRCh38
rs754270930 Chr.19: 57547050-57547050 on Build GRCh38
rs202241911 Chr.19: 57547307-57547307 on Build GRCh38
rs868581129 Chr.19: 57547273-57547274 on Build GRCh38
rs1548476 Chr.19: 57547168-57547168 on Build GRCh38
rs1020395234 Chr.19: 57547299-57547299 on Build GRCh38
rs758541560 Chr.19: 57547145-57547145 on Build GRCh38
rs746560146 Chr.19: 57547336-57547336 on Build GRCh38
rs17857023 Chr.19: 57547258-57547258 on Build GRCh38
rs775278357 Chr.19: 57547000-57547000 on Build GRCh38
rs534205692 Chr.19: 57546954-57546954 on Build GRCh38
rs759089320 Chr.19: 57547073-57547074 on Build GRCh38
rs377442837 Chr.19: 57547326-57547326 on Build GRCh38
rs938045550 Chr.19: 57547209-57547209 on Build GRCh38
rs780749850 Chr.19: 57547348-57547348 on Build GRCh38
rs141163690 Chr.19: 57547195-57547195 on Build GRCh38
rs973093807 Chr.19: 57546931-57546931 on Build GRCh38
rs116624281 Chr.19: 57547077-57547077 on Build GRCh38
rs748181844 Chr.19: 57547054-57547054 on Build GRCh38
rs757433267 Chr.19: 57547218-57547218 on Build GRCh38
rs532492261 Chr.19: 57547334-57547334 on Build GRCh38
rs763575504 Chr.19: 57547288-57547288 on Build GRCh38
rs764604148 Chr.19: 57547292-57547292 on Build GRCh38
rs752891784 Chr.19: 57547143-57547143 on Build GRCh38
rs936927651 Chr.19: 57547271-57547271 on Build GRCh38
rs1000555832 Chr.19: 57547319-57547319 on Build GRCh38
rs772887743 Chr.19: 57547026-57547027 on Build GRCh38

View Full Product Details