Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 157532740-157533310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACVR1C
Entrez Gene ID
130399
Gene Name
activin A receptor type 1C
Gene Aliases
ACVRLK7, ALK7
UniGene
Hs.562901
Gene Chromosome Location
Chr.2: 157526767-157628887 on Build GRCh38
Transcripts
NM_001111031.1 NM_001111032.1 NM_001111033.1 NM_145259.2

SNP Detail

SNP ID Chromosome Location
rs955294075 Chr.2: 157532769-157532769 on Build GRCh38
rs996802178 Chr.2: 157533053-157533053 on Build GRCh38
rs1014492559 Chr.2: 157532913-157532913 on Build GRCh38
rs1044610557 Chr.2: 157533267-157533267 on Build GRCh38
rs959607101 Chr.2: 157533198-157533198 on Build GRCh38
rs749875050 Chr.2: 157533074-157533074 on Build GRCh38
rs1042702809 Chr.2: 157533031-157533031 on Build GRCh38
rs925658205 Chr.2: 157533098-157533098 on Build GRCh38
rs1021525327 Chr.2: 157532786-157532786 on Build GRCh38
rs966934645 Chr.2: 157532798-157532798 on Build GRCh38
rs371678220 Chr.2: 157533070-157533070 on Build GRCh38
rs1027694307 Chr.2: 157532956-157532956 on Build GRCh38
rs983564019 Chr.2: 157532977-157532977 on Build GRCh38
rs1014843225 Chr.2: 157533219-157533219 on Build GRCh38
rs898416363 Chr.2: 157533044-157533044 on Build GRCh38
rs550243529 Chr.2: 157532924-157532924 on Build GRCh38
rs990787469 Chr.2: 157532934-157532934 on Build GRCh38
rs896003173 Chr.2: 157532885-157532885 on Build GRCh38
rs375358786 Chr.2: 157533019-157533019 on Build GRCh38
rs1020716322 Chr.2: 157533124-157533124 on Build GRCh38
rs908351591 Chr.2: 157532983-157532983 on Build GRCh38
rs917892807 Chr.2: 157533251-157533251 on Build GRCh38
rs528945616 Chr.2: 157532869-157532869 on Build GRCh38
rs1008017902 Chr.2: 157532775-157532775 on Build GRCh38
rs970781359 Chr.2: 157533261-157533261 on Build GRCh38
rs146121930 Chr.2: 157532951-157532951 on Build GRCh38
rs796741390 Chr.2: 157533004-157533004 on Build GRCh38
rs921189200 Chr.2: 157533024-157533025 on Build GRCh38
rs959327586 Chr.2: 157532909-157532909 on Build GRCh38
rs1032523702 Chr.2: 157533178-157533178 on Build GRCh38
rs1049422463 Chr.2: 157533091-157533091 on Build GRCh38
rs895954605 Chr.2: 157533209-157533209 on Build GRCh38
rs983910826 Chr.2: 157533269-157533269 on Build GRCh38
rs991070393 Chr.2: 157533237-157533237 on Build GRCh38
rs998603104 Chr.2: 157532770-157532770 on Build GRCh38
rs1038268961 Chr.2: 157533027-157533027 on Build GRCh38
rs950770042 Chr.2: 157532964-157532964 on Build GRCh38
rs6751595 Chr.2: 157533006-157533006 on Build GRCh38
rs967257243 Chr.2: 157533142-157533142 on Build GRCh38
rs200203351 Chr.2: 157533096-157533098 on Build GRCh38
rs907969048 Chr.2: 157533287-157533287 on Build GRCh38
rs532760471 Chr.2: 157533157-157533157 on Build GRCh38
rs890823125 Chr.2: 157533092-157533092 on Build GRCh38
rs1047830680 Chr.2: 157532994-157532994 on Build GRCh38
rs375855463 Chr.2: 157532993-157532995 on Build GRCh38
rs187633049 Chr.2: 157533285-157533285 on Build GRCh38
rs979626483 Chr.2: 157532803-157532803 on Build GRCh38
rs1008382904 Chr.2: 157533117-157533117 on Build GRCh38
rs556459130 Chr.2: 157532936-157532936 on Build GRCh38
rs149589060 Chr.2: 157532937-157532937 on Build GRCh38
rs906110003 Chr.2: 157533284-157533284 on Build GRCh38
rs113596424 Chr.2: 157532859-157532859 on Build GRCh38
rs934399407 Chr.2: 157533119-157533119 on Build GRCh38
rs1023650767 Chr.2: 157532918-157532918 on Build GRCh38

View Full Product Details