Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 226983649-226984146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COQ8A
Entrez Gene ID
56997
Gene Name
coenzyme Q8A
Gene Aliases
ADCK3, ARCA2, CABC1, COQ10D4, COQ8, SCAR9
UniGene
Hs.118241
Gene Chromosome Location
Chr.1: 226939339-226987545 on Build GRCh38
Transcripts
NM_020247.4 XM_005273201.1 XM_011544238.1 XM_011544239.2 XM_011544240.2 XM_011544241.2 XM_017001852.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4764687 c.409C>T p.R137C Chr.1: 226983844-226983844 on Build GRCh38
1199153 c.349G>A p.D117N Chr.1: 226983784-226983784 on Build GRCh38
1199152 c.1186G>A p.D396N Chr.1: 226983784-226983784 on Build GRCh38
6024881 c.1207G>A p.A403T Chr.1: 226983805-226983805 on Build GRCh38
4764686 c.1246C>T p.R416C Chr.1: 226983844-226983844 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374177780 Chr.1: 226984065-226984065 on Build GRCh38
rs147097934 Chr.1: 226984095-226984095 on Build GRCh38
rs938803428 Chr.1: 226983954-226983954 on Build GRCh38
rs908147388 Chr.1: 226984012-226984013 on Build GRCh38
rs199921339 Chr.1: 226983791-226983791 on Build GRCh38
rs375918113 Chr.1: 226984036-226984038 on Build GRCh38
rs886046067 Chr.1: 226983751-226983751 on Build GRCh38
rs185185707 Chr.1: 226983870-226983870 on Build GRCh38
rs780764114 Chr.1: 226984092-226984092 on Build GRCh38
rs772918474 Chr.1: 226983866-226983866 on Build GRCh38
rs775259402 Chr.1: 226983750-226983750 on Build GRCh38
rs531785041 Chr.1: 226983707-226983707 on Build GRCh38
rs766498050 Chr.1: 226983884-226983884 on Build GRCh38
rs745821983 Chr.1: 226983736-226983736 on Build GRCh38
rs564593360 Chr.1: 226983865-226983865 on Build GRCh38
rs753254213 Chr.1: 226983826-226983826 on Build GRCh38
rs768499518 Chr.1: 226983803-226983803 on Build GRCh38
rs1021617139 Chr.1: 226983924-226983924 on Build GRCh38
rs1049464882 Chr.1: 226984035-226984035 on Build GRCh38
rs371169984 Chr.1: 226983869-226983869 on Build GRCh38
rs760238739 Chr.1: 226983814-226983814 on Build GRCh38
rs369424828 Chr.1: 226984071-226984071 on Build GRCh38
rs756830028 Chr.1: 226983671-226983671 on Build GRCh38
rs780094357 Chr.1: 226983799-226983799 on Build GRCh38
rs764541059 Chr.1: 226983834-226983834 on Build GRCh38
rs201300405 Chr.1: 226984037-226984038 on Build GRCh38
rs757644950 Chr.1: 226983671-226983672 on Build GRCh38
rs377335668 Chr.1: 226984037-226984037 on Build GRCh38
rs753152068 Chr.1: 226983717-226983717 on Build GRCh38
rs17849927 Chr.1: 226983783-226983783 on Build GRCh38
rs998937188 Chr.1: 226983873-226983873 on Build GRCh38
rs751932768 Chr.1: 226983835-226983835 on Build GRCh38
rs997124676 Chr.1: 226983686-226983686 on Build GRCh38
rs756228992 Chr.1: 226983883-226983883 on Build GRCh38
rs908262644 Chr.1: 226983991-226983991 on Build GRCh38
rs982573678 Chr.1: 226983977-226983977 on Build GRCh38
rs200624777 Chr.1: 226983727-226983727 on Build GRCh38
rs573246415 Chr.1: 226984080-226984080 on Build GRCh38
rs755434240 Chr.1: 226983681-226983681 on Build GRCh38
rs556953183 Chr.1: 226983705-226983705 on Build GRCh38
rs200965287 Chr.1: 226983890-226983890 on Build GRCh38
rs754818658 Chr.1: 226983862-226983862 on Build GRCh38
rs932411673 Chr.1: 226984035-226984036 on Build GRCh38
rs577406586 Chr.1: 226983748-226983748 on Build GRCh38
rs765235930 Chr.1: 226984073-226984073 on Build GRCh38
rs751350069 Chr.1: 226984088-226984088 on Build GRCh38
rs762341495 Chr.1: 226984053-226984053 on Build GRCh38
rs566516807 Chr.1: 226984026-226984026 on Build GRCh38
rs1574185 Chr.1: 226984038-226984038 on Build GRCh38
rs955322017 Chr.1: 226983975-226983975 on Build GRCh38
rs367834081 Chr.1: 226984055-226984055 on Build GRCh38
rs772663118 Chr.1: 226984121-226984121 on Build GRCh38
rs1029268982 Chr.1: 226983966-226983966 on Build GRCh38
rs557215916 Chr.1: 226984040-226984040 on Build GRCh38
rs541611601 Chr.1: 226983771-226983771 on Build GRCh38
rs763311061 Chr.1: 226983827-226983827 on Build GRCh38
rs375538489 Chr.1: 226984046-226984046 on Build GRCh38
rs373191958 Chr.1: 226984072-226984072 on Build GRCh38
rs778122187 Chr.1: 226983722-226983722 on Build GRCh38
rs200375779 Chr.1: 226984101-226984101 on Build GRCh38
rs747141838 Chr.1: 226984044-226984045 on Build GRCh38
rs749174686 Chr.1: 226983801-226983801 on Build GRCh38
rs754359903 Chr.1: 226983678-226983678 on Build GRCh38
rs983605561 Chr.1: 226983996-226983996 on Build GRCh38
rs769430648 Chr.1: 226984097-226984097 on Build GRCh38
rs541836846 Chr.1: 226983796-226983796 on Build GRCh38
rs745418665 Chr.1: 226984048-226984048 on Build GRCh38
rs139133094 Chr.1: 226983786-226983786 on Build GRCh38
rs769414236 Chr.1: 226984050-226984050 on Build GRCh38
rs778795641 Chr.1: 226983863-226983863 on Build GRCh38
rs767899984 Chr.1: 226983845-226983845 on Build GRCh38
rs561835134 Chr.1: 226984098-226984098 on Build GRCh38
rs535772887 Chr.1: 226984039-226984039 on Build GRCh38
rs752645843 Chr.1: 226984074-226984074 on Build GRCh38
rs750675370 Chr.1: 226983881-226983881 on Build GRCh38
rs568908090 Chr.1: 226983919-226983919 on Build GRCh38
rs3215919 Chr.1: 226983884-226983884 on Build GRCh38
rs763355267 Chr.1: 226984072-226984073 on Build GRCh38
rs760476900 Chr.1: 226983772-226983772 on Build GRCh38
rs764547662 Chr.1: 226983872-226983872 on Build GRCh38
rs761489506 Chr.1: 226983806-226983806 on Build GRCh38
rs980169375 Chr.1: 226983936-226983936 on Build GRCh38
rs563315501 Chr.1: 226983804-226983804 on Build GRCh38
rs749258013 Chr.1: 226983855-226983855 on Build GRCh38
rs546725116 Chr.1: 226983888-226983888 on Build GRCh38
rs777934964 Chr.1: 226983787-226983787 on Build GRCh38
rs900790094 Chr.1: 226984084-226984084 on Build GRCh38
rs775063681 Chr.1: 226984052-226984052 on Build GRCh38
rs746410263 Chr.1: 226983863-226983872 on Build GRCh38
rs1031831880 Chr.1: 226983669-226983669 on Build GRCh38
rs397840166 Chr.1: 226983886-226983886 on Build GRCh38
rs990150308 Chr.1: 226983911-226983911 on Build GRCh38
rs758919616 Chr.1: 226983770-226983770 on Build GRCh38
rs753166346 Chr.1: 226983761-226983761 on Build GRCh38
rs745983452 Chr.1: 226983797-226983797 on Build GRCh38
rs776316274 Chr.1: 226984047-226984047 on Build GRCh38
rs760985111 Chr.1: 226984068-226984068 on Build GRCh38
rs757052741 Chr.1: 226984089-226984089 on Build GRCh38
rs781354287 Chr.1: 226983735-226983735 on Build GRCh38
rs539434887 Chr.1: 226983960-226983960 on Build GRCh38
rs369852478 Chr.1: 226983891-226983891 on Build GRCh38
rs1574184 Chr.1: 226984036-226984036 on Build GRCh38
rs546015164 Chr.1: 226983854-226983854 on Build GRCh38
rs149505935 Chr.1: 226983894-226983894 on Build GRCh38
rs151292389 Chr.1: 226983844-226983844 on Build GRCh38
rs765825787 Chr.1: 226983784-226983784 on Build GRCh38
rs562847528 Chr.1: 226983699-226983699 on Build GRCh38
rs550615581 Chr.1: 226983897-226983897 on Build GRCh38
rs975081029 Chr.1: 226983940-226983940 on Build GRCh38
rs878923778 Chr.1: 226983708-226983708 on Build GRCh38
rs534916785 Chr.1: 226984028-226984028 on Build GRCh38
rs1038453779 Chr.1: 226984020-226984020 on Build GRCh38
rs765804167 Chr.1: 226983822-226983822 on Build GRCh38
rs751845468 Chr.1: 226983789-226983789 on Build GRCh38
rs149048093 Chr.1: 226983813-226983813 on Build GRCh38
rs149374145 Chr.1: 226984035-226984036 on Build GRCh38
rs746698209 Chr.1: 226984044-226984044 on Build GRCh38
rs774975818 Chr.1: 226983874-226983874 on Build GRCh38
rs748684298 Chr.1: 226984111-226984111 on Build GRCh38
rs970553498 Chr.1: 226983930-226983930 on Build GRCh38
rs767004123 Chr.1: 226983672-226983672 on Build GRCh38
rs758434964 Chr.1: 226983883-226983883 on Build GRCh38
rs202227926 Chr.1: 226983849-226983849 on Build GRCh38
rs6690459 Chr.1: 226983711-226983711 on Build GRCh38
rs941212672 Chr.1: 226984018-226984018 on Build GRCh38
rs748076385 Chr.1: 226983893-226983893 on Build GRCh38
rs762853525 Chr.1: 226983768-226983768 on Build GRCh38
rs771755682 Chr.1: 226983809-226983809 on Build GRCh38
rs975008867 Chr.1: 226983743-226983743 on Build GRCh38
rs200440257 Chr.1: 226984085-226984085 on Build GRCh38
rs747183826 Chr.1: 226983725-226983725 on Build GRCh38
rs757385972 Chr.1: 226983811-226983811 on Build GRCh38
rs762323369 Chr.1: 226983879-226983879 on Build GRCh38
rs551236927 Chr.1: 226984017-226984017 on Build GRCh38
rs770489444 Chr.1: 226983867-226983867 on Build GRCh38
rs758656323 Chr.1: 226983721-226983721 on Build GRCh38
rs575275341 Chr.1: 226984042-226984042 on Build GRCh38
rs1004253103 Chr.1: 226984113-226984113 on Build GRCh38
rs767986261 Chr.1: 226983880-226983880 on Build GRCh38
rs779228058 Chr.1: 226983681-226983682 on Build GRCh38

View Full Product Details