Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 6999201-6999702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX12
Entrez Gene ID
239
Gene Name
arachidonate 12-lipoxygenase, 12S type
Gene Aliases
12-LOX, 12S-LOX, LOG12
UniGene
Hs.654431
Gene Chromosome Location
Chr.17: 6993791-7010736 on Build GRCh38
Transcripts
NM_000697.2 XM_011523780.2 NR_040089.1

Gene Symbol
ALOX12-AS1
Entrez Gene ID
100506713
Gene Name
ALOX12 antisense RNA 1
Gene Aliases
-
UniGene
Hs.422967
Gene Chromosome Location
Chr.17: 6985123-7012334 on Build GRCh38
Transcripts
NM_000697.2 XM_011523780.2 NR_040089.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4555838 c.670G>A p.D224N Chr.17: 6999329-6999329 on Build GRCh38
3958700 c.669G>A p.Q223Q Chr.17: 6999328-6999328 on Build GRCh38
437256 c.795G>C p.E265D Chr.17: 6999454-6999454 on Build GRCh38
6227514 c.656G>A p.R219H Chr.17: 6999315-6999315 on Build GRCh38
1385649 c.781C>T p.Q261* Chr.17: 6999440-6999440 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767111242 Chr.17: 6999315-6999315 on Build GRCh38
rs758548271 Chr.17: 6999387-6999387 on Build GRCh38
rs770725491 Chr.17: 6999431-6999431 on Build GRCh38
rs747070515 Chr.17: 6999393-6999393 on Build GRCh38
rs987568846 Chr.17: 6999312-6999312 on Build GRCh38
rs751228782 Chr.17: 6999297-6999297 on Build GRCh38
rs1126667 Chr.17: 6999441-6999441 on Build GRCh38
rs955225480 Chr.17: 6999645-6999645 on Build GRCh38
rs1042356 Chr.17: 6999424-6999424 on Build GRCh38
rs1000538858 Chr.17: 6999248-6999248 on Build GRCh38
rs976153972 Chr.17: 6999409-6999409 on Build GRCh38
rs893379584 Chr.17: 6999622-6999622 on Build GRCh38
rs893431768 Chr.17: 6999319-6999319 on Build GRCh38
rs554047138 Chr.17: 6999672-6999672 on Build GRCh38
rs767767360 Chr.17: 6999478-6999479 on Build GRCh38
rs772210455 Chr.17: 6999267-6999267 on Build GRCh38
rs769689866 Chr.17: 6999286-6999286 on Build GRCh38
rs773419455 Chr.17: 6999637-6999637 on Build GRCh38
rs749766071 Chr.17: 6999304-6999305 on Build GRCh38
rs1045232814 Chr.17: 6999234-6999234 on Build GRCh38
rs780225081 Chr.17: 6999391-6999391 on Build GRCh38
rs567767884 Chr.17: 6999336-6999336 on Build GRCh38
rs76734990 Chr.17: 6999476-6999476 on Build GRCh38
rs11571333 Chr.17: 6999491-6999491 on Build GRCh38
rs754414269 Chr.17: 6999460-6999460 on Build GRCh38
rs950304061 Chr.17: 6999482-6999482 on Build GRCh38
rs375234511 Chr.17: 6999654-6999654 on Build GRCh38
rs771600031 Chr.17: 6999329-6999331 on Build GRCh38
rs1037971190 Chr.17: 6999564-6999564 on Build GRCh38
rs917385857 Chr.17: 6999453-6999453 on Build GRCh38
rs760280648 Chr.17: 6999448-6999448 on Build GRCh38
rs557468213 Chr.17: 6999547-6999547 on Build GRCh38
rs757723623 Chr.17: 6999370-6999370 on Build GRCh38
rs765703279 Chr.17: 6999477-6999477 on Build GRCh38
rs996735754 Chr.17: 6999620-6999620 on Build GRCh38
rs1020479235 Chr.17: 6999353-6999353 on Build GRCh38
rs1044784861 Chr.17: 6999538-6999538 on Build GRCh38
rs775421758 Chr.17: 6999271-6999271 on Build GRCh38
rs781454389 Chr.17: 6999522-6999522 on Build GRCh38
rs371090758 Chr.17: 6999390-6999390 on Build GRCh38
rs4987104 Chr.17: 6999434-6999434 on Build GRCh38
rs774510016 Chr.17: 6999335-6999336 on Build GRCh38
rs1010390853 Chr.17: 6999321-6999321 on Build GRCh38
rs376244966 Chr.17: 6999294-6999294 on Build GRCh38
rs936317214 Chr.17: 6999575-6999575 on Build GRCh38
rs778398810 Chr.17: 6999362-6999362 on Build GRCh38
rs759052135 Chr.17: 6999302-6999302 on Build GRCh38
rs146600638 Chr.17: 6999423-6999423 on Build GRCh38
rs762252945 Chr.17: 6999242-6999242 on Build GRCh38
rs1013309180 Chr.17: 6999227-6999227 on Build GRCh38
rs554996718 Chr.17: 6999436-6999436 on Build GRCh38
rs754344753 Chr.17: 6999366-6999366 on Build GRCh38
rs750762602 Chr.17: 6999386-6999386 on Build GRCh38
rs773041408 Chr.17: 6999291-6999291 on Build GRCh38
rs538431408 Chr.17: 6999433-6999433 on Build GRCh38
rs917736734 Chr.17: 6999521-6999521 on Build GRCh38
rs759547635 Chr.17: 6999353-6999353 on Build GRCh38
rs909117381 Chr.17: 6999467-6999467 on Build GRCh38
rs761880352 Chr.17: 6999276-6999276 on Build GRCh38
rs751879724 Chr.17: 6999456-6999456 on Build GRCh38
rs2307218 Chr.17: 6999295-6999295 on Build GRCh38
rs371681028 Chr.17: 6999493-6999493 on Build GRCh38
rs747192903 Chr.17: 6999494-6999494 on Build GRCh38
rs757829809 Chr.17: 6999471-6999471 on Build GRCh38
rs569498296 Chr.17: 6999352-6999352 on Build GRCh38
rs535721539 Chr.17: 6999644-6999644 on Build GRCh38
rs546166792 Chr.17: 6999581-6999581 on Build GRCh38
rs2292352 Chr.17: 6999568-6999568 on Build GRCh38
rs779092848 Chr.17: 6999371-6999371 on Build GRCh38
rs759297658 Chr.17: 6999435-6999435 on Build GRCh38

View Full Product Details