Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 544301-544841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALNT3
Entrez Gene ID
283358
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 3
Gene Aliases
-
UniGene
Hs.504416
Gene Chromosome Location
Chr.12: 460377-561892 on Build GRCh38
Transcripts
NM_173593.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5517681 c.358G>A p.G120S Chr.12: 544345-544345 on Build GRCh38
4044169 c.445C>A p.H149N Chr.12: 544432-544432 on Build GRCh38
3464235 c.416G>A p.R139K Chr.12: 544403-544403 on Build GRCh38
1184336 c.362G>A p.R121H Chr.12: 544349-544349 on Build GRCh38
6609399 c.440A>G p.Y147C Chr.12: 544427-544427 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146217798 Chr.12: 544667-544667 on Build GRCh38
rs148339739 Chr.12: 544348-544348 on Build GRCh38
rs577667898 Chr.12: 544764-544764 on Build GRCh38
rs748073844 Chr.12: 544416-544416 on Build GRCh38
rs150421237 Chr.12: 544428-544428 on Build GRCh38
rs971357458 Chr.12: 544583-544583 on Build GRCh38
rs936151950 Chr.12: 544452-544452 on Build GRCh38
rs750806329 Chr.12: 544486-544486 on Build GRCh38
rs201835778 Chr.12: 544396-544396 on Build GRCh38
rs367723610 Chr.12: 544465-544465 on Build GRCh38
rs760193214 Chr.12: 544582-544582 on Build GRCh38
rs929236734 Chr.12: 544401-544401 on Build GRCh38
rs117643914 Chr.12: 544697-544697 on Build GRCh38
rs769493394 Chr.12: 544425-544425 on Build GRCh38
rs554222360 Chr.12: 544578-544578 on Build GRCh38
rs547634433 Chr.12: 544520-544520 on Build GRCh38
rs765585044 Chr.12: 544474-544474 on Build GRCh38
rs911668924 Chr.12: 544767-544767 on Build GRCh38
rs747012361 Chr.12: 544329-544329 on Build GRCh38
rs754327263 Chr.12: 544363-544363 on Build GRCh38
rs543301629 Chr.12: 544803-544803 on Build GRCh38
rs757585018 Chr.12: 544365-544365 on Build GRCh38
rs561464096 Chr.12: 544812-544812 on Build GRCh38
rs758121985 Chr.12: 544392-544392 on Build GRCh38
rs770644869 Chr.12: 544661-544661 on Build GRCh38
rs573970193 Chr.12: 544433-544433 on Build GRCh38
rs142833414 Chr.12: 544765-544765 on Build GRCh38
rs755006134 Chr.12: 544406-544406 on Build GRCh38
rs902796233 Chr.12: 544592-544592 on Build GRCh38
rs373586871 Chr.12: 544463-544463 on Build GRCh38
rs772256546 Chr.12: 544448-544448 on Build GRCh38
rs999788117 Chr.12: 544609-544609 on Build GRCh38
rs977414681 Chr.12: 544623-544623 on Build GRCh38
rs1050642344 Chr.12: 544487-544487 on Build GRCh38
rs964440700 Chr.12: 544684-544684 on Build GRCh38
rs776539129 Chr.12: 544338-544338 on Build GRCh38
rs762313364 Chr.12: 544472-544472 on Build GRCh38
rs935305340 Chr.12: 544814-544814 on Build GRCh38
rs768702913 Chr.12: 544330-544330 on Build GRCh38
rs576561177 Chr.12: 544751-544751 on Build GRCh38
rs1036044021 Chr.12: 544778-544778 on Build GRCh38
rs746497268 Chr.12: 544398-544398 on Build GRCh38
rs759699324 Chr.12: 544346-544346 on Build GRCh38
rs1008586 Chr.12: 544791-544791 on Build GRCh38
rs976193662 Chr.12: 544719-544719 on Build GRCh38
rs765578298 Chr.12: 544386-544386 on Build GRCh38
rs141528374 Chr.12: 544349-544349 on Build GRCh38
rs1046389956 Chr.12: 544585-544585 on Build GRCh38
rs944384083 Chr.12: 544505-544505 on Build GRCh38
rs774463744 Chr.12: 544345-544345 on Build GRCh38
rs1017393400 Chr.12: 544679-544679 on Build GRCh38
rs182694033 Chr.12: 544562-544562 on Build GRCh38
rs1032555789 Chr.12: 544619-544619 on Build GRCh38
rs779638162 Chr.12: 544328-544328 on Build GRCh38
rs994645735 Chr.12: 544538-544538 on Build GRCh38
rs990602413 Chr.12: 544790-544790 on Build GRCh38
rs897765500 Chr.12: 544517-544517 on Build GRCh38
rs949487513 Chr.12: 544802-544802 on Build GRCh38
rs538815093 Chr.12: 544432-544432 on Build GRCh38
rs146905079 Chr.12: 544342-544342 on Build GRCh38
rs890123063 Chr.12: 544496-544496 on Build GRCh38
rs1008587 Chr.12: 544818-544818 on Build GRCh38
rs990687210 Chr.12: 544665-544665 on Build GRCh38
rs769595274 Chr.12: 544343-544343 on Build GRCh38
rs536555178 Chr.12: 544571-544571 on Build GRCh38
rs1031859822 Chr.12: 544654-544654 on Build GRCh38
rs370715187 Chr.12: 544322-544322 on Build GRCh38
rs778461078 Chr.12: 544321-544321 on Build GRCh38
rs138912908 Chr.12: 544723-544723 on Build GRCh38
rs761215985 Chr.12: 544355-544355 on Build GRCh38
rs746859237 Chr.12: 544390-544390 on Build GRCh38
rs553963700 Chr.12: 544782-544782 on Build GRCh38
rs1036263871 Chr.12: 544508-544508 on Build GRCh38
rs771720326 Chr.12: 544324-544324 on Build GRCh38
rs931755711 Chr.12: 544484-544484 on Build GRCh38
rs944331557 Chr.12: 544770-544770 on Build GRCh38
rs745475326 Chr.12: 544323-544323 on Build GRCh38
rs370503102 Chr.12: 544457-544457 on Build GRCh38
rs781027620 Chr.12: 544410-544410 on Build GRCh38
rs1054051615 Chr.12: 544815-544815 on Build GRCh38
rs764473836 Chr.12: 544357-544357 on Build GRCh38

View Full Product Details