Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53157646-53158144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSAD
Entrez Gene ID
51380
Gene Name
cysteine sulfinic acid decarboxylase
Gene Aliases
CSD, PCAP
UniGene
Hs.279815
Gene Chromosome Location
Chr.12: 53157663-53180909 on Build GRCh38
Transcripts
NM_001244705.1 NM_001244706.1 NM_015989.4 XM_011538442.2 XM_011538446.2 XM_011538449.1 XM_011538451.1 XM_017019416.1 XM_017019417.1 XM_017019418.1 XM_017019419.1

SNP Detail

SNP ID Chromosome Location
rs949890223 Chr.12: 53157764-53157765 on Build GRCh38
rs985302916 Chr.12: 53158003-53158003 on Build GRCh38
rs1056725639 Chr.12: 53157830-53157830 on Build GRCh38
rs991327219 Chr.12: 53158065-53158065 on Build GRCh38
rs536710723 Chr.12: 53157696-53157696 on Build GRCh38
rs905867963 Chr.12: 53157911-53157911 on Build GRCh38
rs373897115 Chr.12: 53158108-53158108 on Build GRCh38
rs868187164 Chr.12: 53158050-53158050 on Build GRCh38
rs929734527 Chr.12: 53158085-53158085 on Build GRCh38
rs955836552 Chr.12: 53157726-53157726 on Build GRCh38
rs956346154 Chr.12: 53157957-53157957 on Build GRCh38
rs1044385950 Chr.12: 53157890-53157890 on Build GRCh38
rs182141031 Chr.12: 53157917-53157917 on Build GRCh38
rs142600486 Chr.12: 53157814-53157814 on Build GRCh38
rs74883721 Chr.12: 53157986-53157986 on Build GRCh38
rs910849411 Chr.12: 53158014-53158014 on Build GRCh38
rs1007804790 Chr.12: 53157851-53157851 on Build GRCh38
rs959374544 Chr.12: 53158048-53158048 on Build GRCh38
rs966159156 Chr.12: 53157909-53157909 on Build GRCh38
rs772703141 Chr.12: 53157826-53157826 on Build GRCh38
rs540016060 Chr.12: 53158032-53158032 on Build GRCh38
rs374128406 Chr.12: 53157878-53157878 on Build GRCh38
rs765925985 Chr.12: 53157965-53157965 on Build GRCh38
rs1056272668 Chr.12: 53158111-53158111 on Build GRCh38
rs955621488 Chr.12: 53157980-53157980 on Build GRCh38
rs78903466 Chr.12: 53157773-53157773 on Build GRCh38
rs1030261030 Chr.12: 53157977-53157977 on Build GRCh38
rs760415030 Chr.12: 53157842-53157842 on Build GRCh38
rs138678289 Chr.12: 53158046-53158046 on Build GRCh38
rs17749735 Chr.12: 53157749-53157749 on Build GRCh38
rs1038087397 Chr.12: 53157811-53157811 on Build GRCh38
rs768622237 Chr.12: 53158112-53158112 on Build GRCh38
rs71455240 Chr.12: 53158076-53158076 on Build GRCh38
rs1033085251 Chr.12: 53157949-53157949 on Build GRCh38
rs962350247 Chr.12: 53158037-53158037 on Build GRCh38
rs771626789 Chr.12: 53157798-53157798 on Build GRCh38
rs1019213090 Chr.12: 53157879-53157879 on Build GRCh38
rs564026814 Chr.12: 53157722-53157722 on Build GRCh38
rs12227574 Chr.12: 53157940-53157940 on Build GRCh38
rs537843442 Chr.12: 53157770-53157773 on Build GRCh38
rs947959156 Chr.12: 53158115-53158115 on Build GRCh38
rs922865451 Chr.12: 53157725-53157725 on Build GRCh38

View Full Product Details