Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12647318-12647818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAN2B1
Entrez Gene ID
4125
Gene Name
mannosidase alpha class 2B member 1
Gene Aliases
LAMAN, MANB
UniGene
Hs.356769
Gene Chromosome Location
Chr.19: 12646508-12666777 on Build GRCh38
Transcripts
NM_000528.3 NM_001173498.1 XM_005259913.1 XM_017026818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5614603 c.2793C>A p.S931R Chr.19: 12647467-12647467 on Build GRCh38
6449633 c.2743C>T p.R915C Chr.19: 12647517-12647517 on Build GRCh38
3422438 c.2826C>T p.F942F Chr.19: 12647327-12647327 on Build GRCh38
4817252 c.2828C>G p.S943C Chr.19: 12647325-12647325 on Build GRCh38
6149609 c.2805C>G p.T935T Chr.19: 12647455-12647455 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758765126 Chr.19: 12647516-12647516 on Build GRCh38
rs1011078500 Chr.19: 12647435-12647435 on Build GRCh38
rs773210333 Chr.19: 12647551-12647551 on Build GRCh38
rs755287307 Chr.19: 12647608-12647608 on Build GRCh38
rs772771585 Chr.19: 12647452-12647459 on Build GRCh38
rs369122358 Chr.19: 12647371-12647371 on Build GRCh38
rs568250986 Chr.19: 12647714-12647714 on Build GRCh38
rs1055129495 Chr.19: 12647408-12647408 on Build GRCh38
rs139366493 Chr.19: 12647532-12647532 on Build GRCh38
rs34996464 Chr.19: 12647780-12647781 on Build GRCh38
rs755681471 Chr.19: 12647583-12647583 on Build GRCh38
rs990275905 Chr.19: 12647395-12647396 on Build GRCh38
rs574411653 Chr.19: 12647553-12647553 on Build GRCh38
rs1024929407 Chr.19: 12647363-12647363 on Build GRCh38
rs559789684 Chr.19: 12647539-12647539 on Build GRCh38
rs759163329 Chr.19: 12647605-12647606 on Build GRCh38
rs775720806 Chr.19: 12647437-12647437 on Build GRCh38
rs150184949 Chr.19: 12647717-12647717 on Build GRCh38
rs199606152 Chr.19: 12647593-12647593 on Build GRCh38
rs1047812725 Chr.19: 12647352-12647352 on Build GRCh38
rs776812208 Chr.19: 12647535-12647535 on Build GRCh38
rs895172069 Chr.19: 12647413-12647413 on Build GRCh38
rs375705953 Chr.19: 12647630-12647631 on Build GRCh38
rs377752596 Chr.19: 12647505-12647505 on Build GRCh38
rs142248782 Chr.19: 12647478-12647478 on Build GRCh38
rs761768804 Chr.19: 12647452-12647452 on Build GRCh38
rs929568109 Chr.19: 12647526-12647526 on Build GRCh38
rs864621990 Chr.19: 12647517-12647517 on Build GRCh38
rs976021105 Chr.19: 12647715-12647715 on Build GRCh38
rs1022521772 Chr.19: 12647439-12647439 on Build GRCh38
rs746289340 Chr.19: 12647414-12647414 on Build GRCh38
rs864621988 Chr.19: 12647592-12647592 on Build GRCh38
rs1004058684 Chr.19: 12647390-12647390 on Build GRCh38
rs749379616 Chr.19: 12647648-12647648 on Build GRCh38
rs1018456781 Chr.19: 12647719-12647719 on Build GRCh38
rs547320420 Chr.19: 12647647-12647647 on Build GRCh38
rs747424062 Chr.19: 12647605-12647605 on Build GRCh38
rs140949750 Chr.19: 12647672-12647672 on Build GRCh38
rs765269491 Chr.19: 12647460-12647460 on Build GRCh38
rs762841449 Chr.19: 12647466-12647466 on Build GRCh38
rs942244704 Chr.19: 12647780-12647780 on Build GRCh38
rs750952284 Chr.19: 12647587-12647587 on Build GRCh38
rs779879898 Chr.19: 12647368-12647368 on Build GRCh38
rs761870446 Chr.19: 12647340-12647340 on Build GRCh38
rs370579499 Chr.19: 12647351-12647351 on Build GRCh38
rs368951229 Chr.19: 12647589-12647589 on Build GRCh38
rs372294720 Chr.19: 12647629-12647629 on Build GRCh38
rs746319362 Chr.19: 12647499-12647499 on Build GRCh38
rs200033151 Chr.19: 12647562-12647562 on Build GRCh38
rs755553683 Chr.19: 12647496-12647496 on Build GRCh38
rs1043735839 Chr.19: 12647698-12647698 on Build GRCh38
rs906602699 Chr.19: 12647370-12647370 on Build GRCh38
rs771531945 Chr.19: 12647380-12647380 on Build GRCh38
rs777777092 Chr.19: 12647642-12647642 on Build GRCh38
rs976302366 Chr.19: 12647482-12647482 on Build GRCh38
rs879931134 Chr.19: 12647766-12647766 on Build GRCh38
rs774819942 Chr.19: 12647621-12647622 on Build GRCh38
rs202009691 Chr.19: 12647631-12647631 on Build GRCh38
rs962341276 Chr.19: 12647784-12647784 on Build GRCh38
rs373332419 Chr.19: 12647630-12647630 on Build GRCh38
rs563944474 Chr.19: 12647665-12647665 on Build GRCh38
rs1029823733 Chr.19: 12647715-12647715 on Build GRCh38
rs546914205 Chr.19: 12647713-12647713 on Build GRCh38
rs575582119 Chr.19: 12647615-12647615 on Build GRCh38
rs773821191 Chr.19: 12647604-12647625 on Build GRCh38
rs776760758 Chr.19: 12647443-12647443 on Build GRCh38
rs772974771 Chr.19: 12647461-12647461 on Build GRCh38
rs368224056 Chr.19: 12647540-12647540 on Build GRCh38
rs864621989 Chr.19: 12647588-12647588 on Build GRCh38
rs928642117 Chr.19: 12647703-12647703 on Build GRCh38
rs541712525 Chr.19: 12647521-12647521 on Build GRCh38
rs748446780 Chr.19: 12647406-12647407 on Build GRCh38
rs374417692 Chr.19: 12647463-12647463 on Build GRCh38
rs1057516864 Chr.19: 12647460-12647461 on Build GRCh38
rs781508235 Chr.19: 12647610-12647610 on Build GRCh38
rs768907989 Chr.19: 12647533-12647533 on Build GRCh38
rs780794113 Chr.19: 12647372-12647372 on Build GRCh38
rs567810171 Chr.19: 12647477-12647477 on Build GRCh38
rs548626057 Chr.19: 12647786-12647786 on Build GRCh38
rs957928289 Chr.19: 12647692-12647692 on Build GRCh38
rs768734132 Chr.19: 12647442-12647442 on Build GRCh38
rs150866794 Chr.19: 12647497-12647497 on Build GRCh38
rs764857421 Chr.19: 12647615-12647615 on Build GRCh38
rs765084966 Chr.19: 12647348-12647348 on Build GRCh38
rs752287794 Chr.19: 12647488-12647488 on Build GRCh38
rs149897798 Chr.19: 12647572-12647572 on Build GRCh38
rs774352491 Chr.19: 12647561-12647561 on Build GRCh38
rs753776387 Chr.19: 12647339-12647339 on Build GRCh38
rs559212808 Chr.19: 12647369-12647369 on Build GRCh38
rs767323371 Chr.19: 12647567-12647567 on Build GRCh38
rs762745066 Chr.19: 12647552-12647552 on Build GRCh38
rs957664522 Chr.19: 12647391-12647391 on Build GRCh38
rs772275417 Chr.19: 12647419-12647419 on Build GRCh38
rs561129352 Chr.19: 12647689-12647689 on Build GRCh38
rs374333674 Chr.19: 12647375-12647375 on Build GRCh38
rs749331220 Chr.19: 12647393-12647393 on Build GRCh38
rs750273286 Chr.19: 12647350-12647350 on Build GRCh38
rs939442511 Chr.19: 12647663-12647663 on Build GRCh38
rs370921236 Chr.19: 12647718-12647718 on Build GRCh38
rs747166225 Chr.19: 12647441-12647441 on Build GRCh38
rs754733253 Chr.19: 12647481-12647481 on Build GRCh38

View Full Product Details