Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35752541-35753042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSMP
Entrez Gene ID
692094
Gene Name
microseminoprotein, prostate associated
Gene Aliases
PSMP
UniGene
Hs.571601
Gene Chromosome Location
Chr.9: 35752990-35754277 on Build GRCh38
Transcripts
NM_001044264.2 XM_005251479.4 XM_011517890.2 XM_011517891.2 XM_011517892.2 XM_011517893.2 XM_017014745.1 XM_017014746.1 XM_017014747.1 XM_017014748.1 XM_017014749.1 XM_017014750.1 NM_001080496.2

Gene Symbol
NPR2
Entrez Gene ID
4882
Gene Name
natriuretic peptide receptor 2
Gene Aliases
AMDM, ANPRB, ANPb, ECDM, GUC2B, GUCY2B, NPRB, NPRBi, SNSK
UniGene
Hs.78518
Gene Chromosome Location
Chr.9: 35752945-35809731 on Build GRCh38
Transcripts
NM_001044264.2 XM_005251479.4 XM_011517890.2 XM_011517891.2 XM_011517892.2 XM_011517893.2 XM_017014745.1 XM_017014746.1 XM_017014747.1 XM_017014748.1 XM_017014749.1 XM_017014750.1 NM_001080496.2

Gene Symbol
RGP1
Entrez Gene ID
9827
Gene Name
RGP1 homolog, RAB6A GEF complex partner 1
Gene Aliases
KIAA0258
UniGene
Hs.493804
Gene Chromosome Location
Chr.9: 35749280-35753267 on Build GRCh38
Transcripts
NM_001044264.2 XM_005251479.4 XM_011517890.2 XM_011517891.2 XM_011517892.2 XM_011517893.2 XM_017014745.1 XM_017014746.1 XM_017014747.1 XM_017014748.1 XM_017014749.1 XM_017014750.1 NM_001080496.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6334781 c.1147G>T p.E383* Chr.9: 35752725-35752725 on Build GRCh38
1108785 c.1274G>A p.S425N Chr.9: 35752852-35752852 on Build GRCh38
7348326 c.989C>T p.T330M Chr.9: 35752687-35752687 on Build GRCh38
487441 c.1135T>A p.Y379N Chr.9: 35752833-35752833 on Build GRCh38
6316785 c.1227G>T p.V409V Chr.9: 35752805-35752805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756953349 Chr.9: 35752905-35752905 on Build GRCh38
rs185547101 Chr.9: 35752718-35752718 on Build GRCh38
rs375091454 Chr.9: 35752661-35752661 on Build GRCh38
rs768943301 Chr.9: 35752844-35752844 on Build GRCh38
rs199898637 Chr.9: 35752642-35752642 on Build GRCh38
rs765169753 Chr.9: 35752910-35752910 on Build GRCh38
rs748517703 Chr.9: 35752857-35752857 on Build GRCh38
rs368056087 Chr.9: 35752687-35752687 on Build GRCh38
rs367815815 Chr.9: 35752742-35752742 on Build GRCh38
rs765745442 Chr.9: 35752767-35752767 on Build GRCh38
rs1033482029 Chr.9: 35752967-35752967 on Build GRCh38
rs181056867 Chr.9: 35752628-35752628 on Build GRCh38
rs776659202 Chr.9: 35752855-35752855 on Build GRCh38
rs972974867 Chr.9: 35752587-35752587 on Build GRCh38
rs569827733 Chr.9: 35752884-35752884 on Build GRCh38
rs753616593 Chr.9: 35752904-35752904 on Build GRCh38
rs371177398 Chr.9: 35752991-35752991 on Build GRCh38
rs1002022904 Chr.9: 35752965-35752965 on Build GRCh38
rs772341695 Chr.9: 35752733-35752733 on Build GRCh38
rs762421620 Chr.9: 35752666-35752666 on Build GRCh38
rs935258418 Chr.9: 35752763-35752763 on Build GRCh38
rs73438778 Chr.9: 35752566-35752566 on Build GRCh38
rs532782192 Chr.9: 35752784-35752784 on Build GRCh38
rs757049602 Chr.9: 35752813-35752813 on Build GRCh38
rs78786755 Chr.9: 35752756-35752756 on Build GRCh38
rs1041279460 Chr.9: 35752723-35752723 on Build GRCh38
rs763606470 Chr.9: 35752812-35752812 on Build GRCh38
rs529233058 Chr.9: 35752954-35752954 on Build GRCh38
rs747018262 Chr.9: 35752840-35752840 on Build GRCh38
rs1002918738 Chr.9: 35752970-35752970 on Build GRCh38
rs576703394 Chr.9: 35752629-35752629 on Build GRCh38
rs755824578 Chr.9: 35752809-35752809 on Build GRCh38
rs780220773 Chr.9: 35752834-35752834 on Build GRCh38
rs775719305 Chr.9: 35752645-35752645 on Build GRCh38
rs200874131 Chr.9: 35752697-35752697 on Build GRCh38
rs765690290 Chr.9: 35752667-35752667 on Build GRCh38
rs755702455 Chr.9: 35752693-35752693 on Build GRCh38
rs752453039 Chr.9: 35752692-35752692 on Build GRCh38
rs760852828 Chr.9: 35752895-35752895 on Build GRCh38
rs934212856 Chr.9: 35752826-35752826 on Build GRCh38
rs137923089 Chr.9: 35752593-35752593 on Build GRCh38
rs767063340 Chr.9: 35752796-35752796 on Build GRCh38
rs758384477 Chr.9: 35752828-35752828 on Build GRCh38
rs371502721 Chr.9: 35752688-35752688 on Build GRCh38
rs752119294 Chr.9: 35752802-35752802 on Build GRCh38
rs542056958 Chr.9: 35752668-35752668 on Build GRCh38
rs962528088 Chr.9: 35752572-35752572 on Build GRCh38
rs764575463 Chr.9: 35752658-35752658 on Build GRCh38
rs369297245 Chr.9: 35752985-35752985 on Build GRCh38
rs563711595 Chr.9: 35752737-35752737 on Build GRCh38
rs775415345 Chr.9: 35752734-35752734 on Build GRCh38
rs750209266 Chr.9: 35752911-35752911 on Build GRCh38
rs763816126 Chr.9: 35752900-35752900 on Build GRCh38
rs777665311 Chr.9: 35752715-35752715 on Build GRCh38
rs745854562 Chr.9: 35752822-35752822 on Build GRCh38
rs900012187 Chr.9: 35752823-35752823 on Build GRCh38
rs773642593 Chr.9: 35752769-35752769 on Build GRCh38
rs778850556 Chr.9: 35752818-35752818 on Build GRCh38
rs777083903 Chr.9: 35752753-35752753 on Build GRCh38
rs1036274092 Chr.9: 35752975-35752975 on Build GRCh38
rs898270774 Chr.9: 35752945-35752946 on Build GRCh38
rs775091270 Chr.9: 35752889-35752889 on Build GRCh38
rs763493130 Chr.9: 35752877-35752877 on Build GRCh38
rs372774064 Chr.9: 35752860-35752860 on Build GRCh38
rs760049994 Chr.9: 35752896-35752896 on Build GRCh38
rs749298798 Chr.9: 35752607-35752607 on Build GRCh38
rs980637386 Chr.9: 35752576-35752576 on Build GRCh38
rs61731189 Chr.9: 35752719-35752719 on Build GRCh38
rs190776142 Chr.9: 35752613-35752613 on Build GRCh38
rs760799263 Chr.9: 35752647-35752647 on Build GRCh38
rs773544726 Chr.9: 35752862-35752862 on Build GRCh38
rs765263378 Chr.9: 35752998-35752998 on Build GRCh38

View Full Product Details